Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81194792-81195300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP131
Entrez Gene ID
22994
Gene Name
centrosomal protein 131
Gene Aliases
AZ1, AZI1, ZA1
UniGene
Hs.514578
Gene Chromosome Location
Chr.17: 81189593-81222989 on Build GRCh38
Transcripts
NM_001009811.3 NM_001319228.1 NM_001319229.1 NM_014984.3 XM_011524547.2 XM_011524548.2 XM_011524550.2 XM_011524551.2 XM_011524552.2 XM_011524553.2 XM_011524555.2 XM_017024369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1589321 c.2035G>T p.E679* Chr.17: 81194954-81194954 on Build GRCh38
7080883 c.2031C>G p.L677L Chr.17: 81194958-81194958 on Build GRCh38
4664833 c.2028delA p.K676fs*5 Chr.17: 81194961-81194961 on Build GRCh38
4534082 c.2047G>A p.A683T Chr.17: 81194942-81194942 on Build GRCh38
6842496 c.2050A>G p.T684A Chr.17: 81194939-81194939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014803908 Chr.17: 81195098-81195098 on Build GRCh38
rs928624081 Chr.17: 81195231-81195231 on Build GRCh38
rs968029496 Chr.17: 81195224-81195224 on Build GRCh38
rs200077707 Chr.17: 81194842-81194842 on Build GRCh38
rs143625194 Chr.17: 81194927-81194927 on Build GRCh38
rs752363261 Chr.17: 81194991-81194991 on Build GRCh38
rs976370750 Chr.17: 81195066-81195066 on Build GRCh38
rs1004002167 Chr.17: 81195246-81195246 on Build GRCh38
rs768712461 Chr.17: 81194958-81194958 on Build GRCh38
rs566594146 Chr.17: 81195111-81195111 on Build GRCh38
rs896524088 Chr.17: 81195137-81195137 on Build GRCh38
rs373106768 Chr.17: 81195006-81195006 on Build GRCh38
rs749803404 Chr.17: 81194835-81194835 on Build GRCh38
rs555770465 Chr.17: 81195257-81195257 on Build GRCh38
rs932190924 Chr.17: 81195107-81195107 on Build GRCh38
rs773514342 Chr.17: 81194841-81194841 on Build GRCh38
rs761865588 Chr.17: 81194952-81194955 on Build GRCh38
rs754572188 Chr.17: 81194825-81194825 on Build GRCh38
rs768787890 Chr.17: 81194864-81194864 on Build GRCh38
rs765095621 Chr.17: 81194821-81194821 on Build GRCh38
rs537468678 Chr.17: 81195188-81195188 on Build GRCh38
rs201227224 Chr.17: 81195018-81195018 on Build GRCh38
rs531749954 Chr.17: 81195029-81195029 on Build GRCh38
rs754518076 Chr.17: 81194912-81194912 on Build GRCh38
rs80268990 Chr.17: 81195279-81195279 on Build GRCh38
rs1052464600 Chr.17: 81194970-81194970 on Build GRCh38
rs909052325 Chr.17: 81195276-81195276 on Build GRCh38
rs759252320 Chr.17: 81194985-81194985 on Build GRCh38
rs368831069 Chr.17: 81194930-81194930 on Build GRCh38
rs957539426 Chr.17: 81195143-81195143 on Build GRCh38
rs375434915 Chr.17: 81194844-81194844 on Build GRCh38
rs183879490 Chr.17: 81195017-81195017 on Build GRCh38
rs780588009 Chr.17: 81195005-81195005 on Build GRCh38
rs748769994 Chr.17: 81194891-81194893 on Build GRCh38
rs529671760 Chr.17: 81195221-81195221 on Build GRCh38
rs547857894 Chr.17: 81195085-81195085 on Build GRCh38
rs146026809 Chr.17: 81195063-81195063 on Build GRCh38
rs766152494 Chr.17: 81194908-81194908 on Build GRCh38
rs755626923 Chr.17: 81194995-81194995 on Build GRCh38
rs961711519 Chr.17: 81194919-81194919 on Build GRCh38
rs767036257 Chr.17: 81194987-81194987 on Build GRCh38
rs550167012 Chr.17: 81195044-81195044 on Build GRCh38
rs1019372761 Chr.17: 81195247-81195247 on Build GRCh38
rs34085091 Chr.17: 81194954-81194955 on Build GRCh38
rs372345143 Chr.17: 81194834-81194834 on Build GRCh38
rs560866471 Chr.17: 81195124-81195124 on Build GRCh38
rs969231104 Chr.17: 81195016-81195016 on Build GRCh38
rs1027964536 Chr.17: 81195256-81195256 on Build GRCh38
rs935488727 Chr.17: 81195280-81195280 on Build GRCh38
rs777349411 Chr.17: 81194837-81194838 on Build GRCh38
rs576433056 Chr.17: 81194817-81194817 on Build GRCh38
rs558437610 Chr.17: 81195136-81195136 on Build GRCh38
rs941960089 Chr.17: 81195166-81195166 on Build GRCh38
rs772622202 Chr.17: 81194936-81194936 on Build GRCh38
rs533940502 Chr.17: 81195132-81195132 on Build GRCh38
rs975083790 Chr.17: 81195252-81195252 on Build GRCh38
rs981429487 Chr.17: 81195225-81195225 on Build GRCh38
rs375187968 Chr.17: 81194942-81194942 on Build GRCh38
rs1045002508 Chr.17: 81195177-81195177 on Build GRCh38
rs990811747 Chr.17: 81195151-81195151 on Build GRCh38
rs1055580379 Chr.17: 81195052-81195052 on Build GRCh38
rs151214434 Chr.17: 81194872-81194872 on Build GRCh38
rs148448761 Chr.17: 81194976-81194976 on Build GRCh38
rs140382436 Chr.17: 81194923-81194923 on Build GRCh38
rs750333661 Chr.17: 81194883-81194883 on Build GRCh38
rs779726124 Chr.17: 81194822-81194822 on Build GRCh38
rs574168291 Chr.17: 81194845-81194845 on Build GRCh38
rs756752594 Chr.17: 81194958-81194959 on Build GRCh38
rs774822556 Chr.17: 81195036-81195036 on Build GRCh38
rs760166346 Chr.17: 81195053-81195053 on Build GRCh38
rs769931717 Chr.17: 81195019-81195019 on Build GRCh38
rs778256519 Chr.17: 81195014-81195014 on Build GRCh38
rs370909487 Chr.17: 81195113-81195114 on Build GRCh38
rs202132358 Chr.17: 81194846-81194846 on Build GRCh38
rs760975823 Chr.17: 81194862-81194862 on Build GRCh38
rs755342037 Chr.17: 81195014-81195014 on Build GRCh38
rs202100395 Chr.17: 81195110-81195111 on Build GRCh38
rs746352568 Chr.17: 81194931-81194931 on Build GRCh38
rs980074077 Chr.17: 81195262-81195262 on Build GRCh38
rs752096772 Chr.17: 81194926-81194926 on Build GRCh38
rs373114995 Chr.17: 81194937-81194937 on Build GRCh38
rs748220076 Chr.17: 81194966-81194966 on Build GRCh38
rs1026846288 Chr.17: 81195208-81195208 on Build GRCh38
rs1018469222 Chr.17: 81195140-81195140 on Build GRCh38
rs774071311 Chr.17: 81194982-81194982 on Build GRCh38
rs144100268 Chr.17: 81195049-81195049 on Build GRCh38
rs762787361 Chr.17: 81194892-81194892 on Build GRCh38
rs949382821 Chr.17: 81195141-81195141 on Build GRCh38
rs375891072 Chr.17: 81195127-81195127 on Build GRCh38
rs761833623 Chr.17: 81194881-81194881 on Build GRCh38
rs995808269 Chr.17: 81195196-81195196 on Build GRCh38
rs897492051 Chr.17: 81195108-81195108 on Build GRCh38
rs758919456 Chr.17: 81195003-81195003 on Build GRCh38
rs574079897 Chr.17: 81195261-81195261 on Build GRCh38
rs143947172 Chr.17: 81195112-81195113 on Build GRCh38
rs570377784 Chr.17: 81195146-81195146 on Build GRCh38
rs922183004 Chr.17: 81195083-81195083 on Build GRCh38
rs766164854 Chr.17: 81194979-81194979 on Build GRCh38
rs369549587 Chr.17: 81195001-81195001 on Build GRCh38
rs762875732 Chr.17: 81194977-81194977 on Build GRCh38
rs965086387 Chr.17: 81195251-81195251 on Build GRCh38
rs776803535 Chr.17: 81194964-81194964 on Build GRCh38
rs564725975 Chr.17: 81195028-81195028 on Build GRCh38
rs997225558 Chr.17: 81195119-81195119 on Build GRCh38
rs1006121837 Chr.17: 81194867-81194867 on Build GRCh38
rs763683244 Chr.17: 81195000-81195000 on Build GRCh38
rs925419665 Chr.17: 81195131-81195131 on Build GRCh38

View Full Product Details