Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81539764-81540269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAAP100
Entrez Gene ID
80233
Gene Name
Fanconi anemia core complex associated protein 100
Gene Aliases
C17orf70
UniGene
Hs.313905
Gene Chromosome Location
Chr.17: 81539885-81552403 on Build GRCh38
Transcripts
NM_025161.5 NR_033338.1 XM_006722111.2 XM_006722112.3 XM_017025162.1 XM_017025163.1

SNP Detail

SNP ID Chromosome Location
rs11552302 Chr.17: 81540033-81540033 on Build GRCh38
rs941101181 Chr.17: 81540131-81540131 on Build GRCh38
rs12675 Chr.17: 81540246-81540246 on Build GRCh38
rs536487526 Chr.17: 81540082-81540082 on Build GRCh38
rs544578195 Chr.17: 81540185-81540185 on Build GRCh38
rs930390371 Chr.17: 81539955-81539955 on Build GRCh38
rs1013708174 Chr.17: 81540160-81540160 on Build GRCh38
rs755052620 Chr.17: 81540186-81540186 on Build GRCh38
rs1016488231 Chr.17: 81539855-81539855 on Build GRCh38
rs989495891 Chr.17: 81540050-81540050 on Build GRCh38
rs538953853 Chr.17: 81539797-81539797 on Build GRCh38
rs1022256719 Chr.17: 81540051-81540051 on Build GRCh38
rs931939695 Chr.17: 81539985-81539985 on Build GRCh38
rs145233365 Chr.17: 81539878-81539878 on Build GRCh38
rs1034968403 Chr.17: 81540209-81540209 on Build GRCh38
rs142464441 Chr.17: 81539910-81539910 on Build GRCh38
rs903944958 Chr.17: 81539831-81539831 on Build GRCh38
rs920128597 Chr.17: 81539952-81539952 on Build GRCh38
rs77650837 Chr.17: 81539956-81539956 on Build GRCh38
rs1003245749 Chr.17: 81539787-81539787 on Build GRCh38
rs1002477991 Chr.17: 81540075-81540075 on Build GRCh38
rs965077328 Chr.17: 81540050-81540051 on Build GRCh38
rs200954420 Chr.17: 81540234-81540235 on Build GRCh38
rs115617790 Chr.17: 81539822-81539822 on Build GRCh38
rs183467338 Chr.17: 81539895-81539895 on Build GRCh38
rs548386354 Chr.17: 81540023-81540023 on Build GRCh38
rs558091279 Chr.17: 81540171-81540171 on Build GRCh38
rs954851367 Chr.17: 81540036-81540036 on Build GRCh38
rs534287295 Chr.17: 81539937-81539937 on Build GRCh38
rs922912351 Chr.17: 81540233-81540233 on Build GRCh38
rs984533260 Chr.17: 81540079-81540079 on Build GRCh38
rs1017651906 Chr.17: 81540045-81540047 on Build GRCh38
rs576434767 Chr.17: 81540172-81540172 on Build GRCh38
rs1050199938 Chr.17: 81540022-81540022 on Build GRCh38
rs533658692 Chr.17: 81540170-81540170 on Build GRCh38
rs918294743 Chr.17: 81540232-81540232 on Build GRCh38
rs773662080 Chr.17: 81539796-81539796 on Build GRCh38
rs776190650 Chr.17: 81539896-81539896 on Build GRCh38
rs966606208 Chr.17: 81540057-81540057 on Build GRCh38
rs767584170 Chr.17: 81540122-81540122 on Build GRCh38
rs1000929161 Chr.17: 81540029-81540029 on Build GRCh38
rs973798122 Chr.17: 81540226-81540226 on Build GRCh38
rs927802117 Chr.17: 81540068-81540068 on Build GRCh38
rs368225992 Chr.17: 81540119-81540119 on Build GRCh38
rs941058153 Chr.17: 81540073-81540073 on Build GRCh38
rs762541920 Chr.17: 81539852-81539852 on Build GRCh38
rs7405641 Chr.17: 81539864-81539864 on Build GRCh38
rs114871016 Chr.17: 81539879-81539879 on Build GRCh38
rs771428862 Chr.17: 81539786-81539786 on Build GRCh38
rs969623726 Chr.17: 81540074-81540074 on Build GRCh38
rs975833330 Chr.17: 81540225-81540225 on Build GRCh38
rs1050319000 Chr.17: 81540140-81540140 on Build GRCh38
rs961276060 Chr.17: 81540044-81540056 on Build GRCh38
rs1013604267 Chr.17: 81539838-81539838 on Build GRCh38
rs190264970 Chr.17: 81539983-81539983 on Build GRCh38
rs7405532 Chr.17: 81539785-81539785 on Build GRCh38
rs961701742 Chr.17: 81540088-81540088 on Build GRCh38
rs1034901436 Chr.17: 81540098-81540098 on Build GRCh38
rs910560353 Chr.17: 81539899-81539899 on Build GRCh38
rs71675424 Chr.17: 81540082-81540082 on Build GRCh38
rs925095464 Chr.17: 81540141-81540141 on Build GRCh38
rs962082201 Chr.17: 81540198-81540198 on Build GRCh38
rs187044215 Chr.17: 81539945-81539945 on Build GRCh38
rs1005178796 Chr.17: 81539988-81539989 on Build GRCh38
rs899415902 Chr.17: 81540008-81540008 on Build GRCh38
rs182511071 Chr.17: 81540154-81540154 on Build GRCh38
rs973767067 Chr.17: 81540134-81540134 on Build GRCh38
rs200502458 Chr.17: 81540081-81540082 on Build GRCh38
rs952871371 Chr.17: 81540183-81540183 on Build GRCh38
rs8055 Chr.17: 81540144-81540144 on Build GRCh38
rs960641821 Chr.17: 81540217-81540217 on Build GRCh38
rs532633782 Chr.17: 81539959-81539959 on Build GRCh38
rs374694419 Chr.17: 81540078-81540078 on Build GRCh38
rs567664102 Chr.17: 81539980-81539980 on Build GRCh38
rs940368231 Chr.17: 81539975-81539975 on Build GRCh38
rs894817307 Chr.17: 81539986-81539986 on Build GRCh38
rs978017006 Chr.17: 81540091-81540091 on Build GRCh38
rs557223552 Chr.17: 81540077-81540077 on Build GRCh38
rs1042165266 Chr.17: 81539784-81539784 on Build GRCh38
rs563240290 Chr.17: 81540205-81540205 on Build GRCh38
rs113868702 Chr.17: 81540063-81540063 on Build GRCh38
rs960862398 Chr.17: 81540105-81540105 on Build GRCh38
rs939170642 Chr.17: 81540071-81540071 on Build GRCh38
rs995969260 Chr.17: 81539819-81539819 on Build GRCh38
rs538515605 Chr.17: 81540076-81540076 on Build GRCh38
rs542310947 Chr.17: 81539865-81539865 on Build GRCh38
rs544901525 Chr.17: 81539974-81539974 on Build GRCh38
rs552575209 Chr.17: 81540072-81540072 on Build GRCh38
rs113223332 Chr.17: 81539844-81539844 on Build GRCh38
rs199534993 Chr.17: 81539845-81539846 on Build GRCh38
rs907619298 Chr.17: 81540083-81540083 on Build GRCh38
rs990660315 Chr.17: 81540115-81540115 on Build GRCh38
rs376004163 Chr.17: 81540145-81540145 on Build GRCh38
rs1025235436 Chr.17: 81539936-81539936 on Build GRCh38
rs567003258 Chr.17: 81540030-81540030 on Build GRCh38
rs1038399107 Chr.17: 81539994-81539994 on Build GRCh38
rs922964621 Chr.17: 81540108-81540108 on Build GRCh38
rs1027258152 Chr.17: 81540179-81540179 on Build GRCh38
rs975718240 Chr.17: 81540092-81540092 on Build GRCh38
rs916330076 Chr.17: 81540072-81540073 on Build GRCh38
rs570873116 Chr.17: 81540073-81540073 on Build GRCh38
rs199737728 Chr.17: 81540081-81540081 on Build GRCh38

View Full Product Details