Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81672753-81673265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC137
Entrez Gene ID
339230
Gene Name
coiled-coil domain containing 137
Gene Aliases
RaRF
UniGene
Hs.744125
Gene Chromosome Location
Chr.17: 81666697-81673907 on Build GRCh38
Transcripts
NM_199287.2 XM_011524738.2

SNP Detail

SNP ID Chromosome Location
rs546104807 Chr.17: 81672855-81672855 on Build GRCh38
rs1137358 Chr.17: 81673090-81673090 on Build GRCh38
rs192168570 Chr.17: 81673172-81673172 on Build GRCh38
rs372530090 Chr.17: 81672872-81672872 on Build GRCh38
rs531819791 Chr.17: 81672889-81672889 on Build GRCh38
rs921750541 Chr.17: 81672910-81672910 on Build GRCh38
rs1041480589 Chr.17: 81673091-81673091 on Build GRCh38
rs904312919 Chr.17: 81673120-81673120 on Build GRCh38
rs560097723 Chr.17: 81672800-81672800 on Build GRCh38
rs948119994 Chr.17: 81673177-81673177 on Build GRCh38
rs974712193 Chr.17: 81672909-81672909 on Build GRCh38
rs950421179 Chr.17: 81673057-81673057 on Build GRCh38
rs995635581 Chr.17: 81673024-81673024 on Build GRCh38
rs942375722 Chr.17: 81672835-81672835 on Build GRCh38
rs923731920 Chr.17: 81673221-81673221 on Build GRCh38
rs1024203599 Chr.17: 81672827-81672827 on Build GRCh38
rs966295955 Chr.17: 81673231-81673231 on Build GRCh38
rs1137357 Chr.17: 81672986-81672986 on Build GRCh38
rs879616120 Chr.17: 81673157-81673157 on Build GRCh38
rs1004655814 Chr.17: 81673066-81673066 on Build GRCh38
rs900885979 Chr.17: 81672853-81672853 on Build GRCh38
rs111614108 Chr.17: 81672974-81672974 on Build GRCh38
rs910763962 Chr.17: 81673056-81673056 on Build GRCh38
rs566078599 Chr.17: 81672996-81672996 on Build GRCh38
rs946608138 Chr.17: 81672918-81672918 on Build GRCh38
rs921831326 Chr.17: 81673116-81673116 on Build GRCh38
rs186801107 Chr.17: 81673086-81673086 on Build GRCh38
rs1014607817 Chr.17: 81673149-81673149 on Build GRCh38
rs979203446 Chr.17: 81673127-81673127 on Build GRCh38
rs59853503 Chr.17: 81672953-81672953 on Build GRCh38
rs976720816 Chr.17: 81673240-81673240 on Build GRCh38
rs962080895 Chr.17: 81672976-81672976 on Build GRCh38
rs142313053 Chr.17: 81672904-81672904 on Build GRCh38
rs756358510 Chr.17: 81673190-81673190 on Build GRCh38
rs1027445801 Chr.17: 81672841-81672841 on Build GRCh38
rs553457215 Chr.17: 81673134-81673134 on Build GRCh38
rs775898794 Chr.17: 81672830-81672830 on Build GRCh38
rs772656123 Chr.17: 81672919-81672919 on Build GRCh38
rs562087712 Chr.17: 81672905-81672905 on Build GRCh38
rs898713123 Chr.17: 81673200-81673200 on Build GRCh38
rs1137360 Chr.17: 81673189-81673189 on Build GRCh38
rs546071919 Chr.17: 81673140-81673140 on Build GRCh38
rs1000064904 Chr.17: 81672896-81672896 on Build GRCh38
rs1043542175 Chr.17: 81672950-81672950 on Build GRCh38
rs34153944 Chr.17: 81672844-81672844 on Build GRCh38
rs146159002 Chr.17: 81673006-81673006 on Build GRCh38
rs993491665 Chr.17: 81673159-81673159 on Build GRCh38
rs1056366257 Chr.17: 81672979-81672979 on Build GRCh38
rs1025826384 Chr.17: 81673026-81673026 on Build GRCh38
rs1019009989 Chr.17: 81673080-81673080 on Build GRCh38
rs910724021 Chr.17: 81673229-81673229 on Build GRCh38
rs935400470 Chr.17: 81673156-81673156 on Build GRCh38
rs866103349 Chr.17: 81672834-81672834 on Build GRCh38
rs373730985 Chr.17: 81672898-81672898 on Build GRCh38
rs968375060 Chr.17: 81673126-81673126 on Build GRCh38
rs999991453 Chr.17: 81673129-81673129 on Build GRCh38
rs114857292 Chr.17: 81673020-81673020 on Build GRCh38
rs1012898442 Chr.17: 81672998-81672998 on Build GRCh38
rs535547408 Chr.17: 81673132-81673132 on Build GRCh38
rs538081797 Chr.17: 81673083-81673083 on Build GRCh38
rs547431765 Chr.17: 81672969-81672969 on Build GRCh38
rs972106622 Chr.17: 81672982-81672982 on Build GRCh38
rs931567476 Chr.17: 81673228-81673228 on Build GRCh38
rs923794056 Chr.17: 81673130-81673130 on Build GRCh38
rs571735412 Chr.17: 81673135-81673135 on Build GRCh38
rs3088016 Chr.17: 81673019-81673019 on Build GRCh38
rs766173816 Chr.17: 81672991-81672991 on Build GRCh38

View Full Product Details