Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81683605-81684109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL16
Entrez Gene ID
339231
Gene Name
ADP ribosylation factor like GTPase 16
Gene Aliases
-
UniGene
Hs.658789
Gene Chromosome Location
Chr.17: 81681194-81683924 on Build GRCh38
Transcripts
NM_001040025.1 NM_004712.4 XM_011525463.2 XM_017025297.1 XM_017025298.1

Gene Symbol
HGS
Entrez Gene ID
9146
Gene Name
hepatocyte growth factor-regulated tyrosine kinase substrate
Gene Aliases
HRS
UniGene
Hs.661056
Gene Chromosome Location
Chr.17: 81683932-81702121 on Build GRCh38
Transcripts
NM_001040025.1 NM_004712.4 XM_011525463.2 XM_017025297.1 XM_017025298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3388303 c.105G>T p.G35G Chr.17: 81683721-81683721 on Build GRCh38
318777 c.103G>T p.G35W Chr.17: 81683723-81683723 on Build GRCh38
318776 c.37G>C p.A13P Chr.17: 81683789-81683789 on Build GRCh38
437792 c.120G>C p.V40V Chr.17: 81683706-81683706 on Build GRCh38
3756031 c.28A>C p.S10R Chr.17: 81683798-81683798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768437046 Chr.17: 81683672-81683672 on Build GRCh38
rs763124555 Chr.17: 81683777-81683777 on Build GRCh38
rs543953971 Chr.17: 81683746-81683746 on Build GRCh38
rs201570553 Chr.17: 81684052-81684052 on Build GRCh38
rs747407664 Chr.17: 81684048-81684048 on Build GRCh38
rs1051699219 Chr.17: 81683949-81683949 on Build GRCh38
rs759776530 Chr.17: 81683726-81683726 on Build GRCh38
rs368213492 Chr.17: 81684063-81684063 on Build GRCh38
rs781356419 Chr.17: 81683761-81683761 on Build GRCh38
rs778619798 Chr.17: 81683711-81683711 on Build GRCh38
rs1003825021 Chr.17: 81684009-81684009 on Build GRCh38
rs760296765 Chr.17: 81683674-81683675 on Build GRCh38
rs547084241 Chr.17: 81683634-81683634 on Build GRCh38
rs767555748 Chr.17: 81683652-81683652 on Build GRCh38
rs753903291 Chr.17: 81684038-81684038 on Build GRCh38
rs763688042 Chr.17: 81683838-81683838 on Build GRCh38
rs747461039 Chr.17: 81684020-81684020 on Build GRCh38
rs564291833 Chr.17: 81683841-81683841 on Build GRCh38
rs935484830 Chr.17: 81684060-81684060 on Build GRCh38
rs983431155 Chr.17: 81683891-81683891 on Build GRCh38
rs200303242 Chr.17: 81683770-81683770 on Build GRCh38
rs764416489 Chr.17: 81683737-81683737 on Build GRCh38
rs938470045 Chr.17: 81683760-81683760 on Build GRCh38
rs545808679 Chr.17: 81683815-81683815 on Build GRCh38
rs931978243 Chr.17: 81683939-81683939 on Build GRCh38
rs201801115 Chr.17: 81683683-81683683 on Build GRCh38
rs745484976 Chr.17: 81683638-81683638 on Build GRCh38
rs774173615 Chr.17: 81683673-81683673 on Build GRCh38
rs907035536 Chr.17: 81683633-81683633 on Build GRCh38
rs1039730498 Chr.17: 81683902-81683902 on Build GRCh38
rs760836941 Chr.17: 81683680-81683680 on Build GRCh38
rs374310994 Chr.17: 81683852-81683852 on Build GRCh38
rs749854038 Chr.17: 81683698-81683698 on Build GRCh38
rs1031317577 Chr.17: 81683806-81683806 on Build GRCh38
rs1013914996 Chr.17: 81683957-81683957 on Build GRCh38
rs920704857 Chr.17: 81683843-81683843 on Build GRCh38
rs771738169 Chr.17: 81683645-81683646 on Build GRCh38
rs767407000 Chr.17: 81684027-81684027 on Build GRCh38
rs753220870 Chr.17: 81683659-81683659 on Build GRCh38
rs756178766 Chr.17: 81683854-81683854 on Build GRCh38
rs745977428 Chr.17: 81683768-81683768 on Build GRCh38
rs745413123 Chr.17: 81683663-81683663 on Build GRCh38
rs761207421 Chr.17: 81683828-81683828 on Build GRCh38
rs772008913 Chr.17: 81683676-81683676 on Build GRCh38
rs200781947 Chr.17: 81683747-81683747 on Build GRCh38
rs8066889 Chr.17: 81683798-81683798 on Build GRCh38
rs775184338 Chr.17: 81683648-81683649 on Build GRCh38
rs749167182 Chr.17: 81683641-81683642 on Build GRCh38
rs537595032 Chr.17: 81684039-81684039 on Build GRCh38
rs750978759 Chr.17: 81683734-81683735 on Build GRCh38
rs768014114 Chr.17: 81683790-81683790 on Build GRCh38
rs771381626 Chr.17: 81684049-81684049 on Build GRCh38
rs376204703 Chr.17: 81683872-81683872 on Build GRCh38
rs755186339 Chr.17: 81683865-81683865 on Build GRCh38
rs774259481 Chr.17: 81683646-81683646 on Build GRCh38
rs774709058 Chr.17: 81683811-81683811 on Build GRCh38
rs202102010 Chr.17: 81683627-81683627 on Build GRCh38
rs1023863962 Chr.17: 81684003-81684004 on Build GRCh38
rs922197449 Chr.17: 81683913-81683913 on Build GRCh38
rs149517153 Chr.17: 81683839-81683839 on Build GRCh38
rs1018981392 Chr.17: 81684025-81684025 on Build GRCh38
rs774636076 Chr.17: 81683788-81683788 on Build GRCh38
rs771043178 Chr.17: 81683723-81683723 on Build GRCh38
rs768046521 Chr.17: 81684059-81684059 on Build GRCh38
rs928306461 Chr.17: 81683881-81683881 on Build GRCh38
rs371738056 Chr.17: 81683691-81683691 on Build GRCh38
rs1052775295 Chr.17: 81683952-81683952 on Build GRCh38
rs749512219 Chr.17: 81683804-81683804 on Build GRCh38
rs754088778 Chr.17: 81683655-81683655 on Build GRCh38
rs371175655 Chr.17: 81683840-81683840 on Build GRCh38
rs758186447 Chr.17: 81683750-81683750 on Build GRCh38
rs372861397 Chr.17: 81683766-81683766 on Build GRCh38
rs780516941 Chr.17: 81683661-81683661 on Build GRCh38
rs769760975 Chr.17: 81683640-81683641 on Build GRCh38
rs959925790 Chr.17: 81683682-81683682 on Build GRCh38
rs189203673 Chr.17: 81683909-81683909 on Build GRCh38
rs370007887 Chr.17: 81683688-81683688 on Build GRCh38
rs377471928 Chr.17: 81683801-81683801 on Build GRCh38
rs764695007 Chr.17: 81683693-81683693 on Build GRCh38
rs561402446 Chr.17: 81683921-81683921 on Build GRCh38
rs755219272 Chr.17: 81683637-81683638 on Build GRCh38
rs200982711 Chr.17: 81684061-81684061 on Build GRCh38
rs748934687 Chr.17: 81683702-81683702 on Build GRCh38
rs114274845 Chr.17: 81684018-81684018 on Build GRCh38
rs763650859 Chr.17: 81683700-81683700 on Build GRCh38
rs376106458 Chr.17: 81683791-81683791 on Build GRCh38
rs541395981 Chr.17: 81683795-81683795 on Build GRCh38
rs907737327 Chr.17: 81683775-81683775 on Build GRCh38
rs999590227 Chr.17: 81683818-81683818 on Build GRCh38
rs577562062 Chr.17: 81684033-81684033 on Build GRCh38
rs765563002 Chr.17: 81683727-81683727 on Build GRCh38
rs146058677 Chr.17: 81684066-81684066 on Build GRCh38
rs766716389 Chr.17: 81683770-81683770 on Build GRCh38
rs766242507 Chr.17: 81684077-81684077 on Build GRCh38
rs747939330 Chr.17: 81683713-81683713 on Build GRCh38
rs998544874 Chr.17: 81683802-81683802 on Build GRCh38
rs755376294 Chr.17: 81683657-81683657 on Build GRCh38
rs775833125 Chr.17: 81683773-81683773 on Build GRCh38
rs995098984 Chr.17: 81683972-81683972 on Build GRCh38
rs907697419 Chr.17: 81683911-81683911 on Build GRCh38
rs374574980 Chr.17: 81683954-81683954 on Build GRCh38
rs759168951 Chr.17: 81683642-81683643 on Build GRCh38
rs764772265 Chr.17: 81683792-81683792 on Build GRCh38
rs776911783 Chr.17: 81683725-81683725 on Build GRCh38
rs750256484 Chr.17: 81684030-81684030 on Build GRCh38
rs780223715 Chr.17: 81683856-81683856 on Build GRCh38
rs944978933 Chr.17: 81683900-81683900 on Build GRCh38
rs749331897 Chr.17: 81683858-81683858 on Build GRCh38
rs759908499 Chr.17: 81683687-81683687 on Build GRCh38
rs570187520 Chr.17: 81683741-81683741 on Build GRCh38
rs369417256 Chr.17: 81683730-81683730 on Build GRCh38
rs752075005 Chr.17: 81683740-81683740 on Build GRCh38
rs934366135 Chr.17: 81683950-81683950 on Build GRCh38
rs958298335 Chr.17: 81683697-81683697 on Build GRCh38
rs761722942 Chr.17: 81683674-81683674 on Build GRCh38
rs752874903 Chr.17: 81684051-81684055 on Build GRCh38
rs531444654 Chr.17: 81683850-81683850 on Build GRCh38
rs767282458 Chr.17: 81684064-81684064 on Build GRCh38
rs528968669 Chr.17: 81683935-81683935 on Build GRCh38
rs764717373 Chr.17: 81683829-81683829 on Build GRCh38
rs746964801 Chr.17: 81683721-81683721 on Build GRCh38
rs184786330 Chr.17: 81683707-81683707 on Build GRCh38
rs998513914 Chr.17: 81683639-81683639 on Build GRCh38
rs753682847 Chr.17: 81684086-81684086 on Build GRCh38
rs987383854 Chr.17: 81684072-81684072 on Build GRCh38
rs746956311 Chr.17: 81683755-81683755 on Build GRCh38
rs755203910 Chr.17: 81683871-81683871 on Build GRCh38
rs748998326 Chr.17: 81683667-81683667 on Build GRCh38
rs900626402 Chr.17: 81683782-81683782 on Build GRCh38
rs769507275 Chr.17: 81683641-81683641 on Build GRCh38
rs756047044 Chr.17: 81684037-81684037 on Build GRCh38
rs779022975 Chr.17: 81683866-81683866 on Build GRCh38
rs775106375 Chr.17: 81683642-81683642 on Build GRCh38
rs746428589 Chr.17: 81684054-81684054 on Build GRCh38
rs748203230 Chr.17: 81683640-81683646 on Build GRCh38
rs863223352 Chr.17: 81683794-81683794 on Build GRCh38
rs768772097 Chr.17: 81683810-81683810 on Build GRCh38
rs757771013 Chr.17: 81683745-81683745 on Build GRCh38
rs780791306 Chr.17: 81683635-81683635 on Build GRCh38
rs760567020 Chr.17: 81684067-81684067 on Build GRCh38
rs747885808 Chr.17: 81683749-81683749 on Build GRCh38
rs979839467 Chr.17: 81683733-81683733 on Build GRCh38
rs946965084 Chr.17: 81684042-81684042 on Build GRCh38
rs533659840 Chr.17: 81683979-81683979 on Build GRCh38
rs941859694 Chr.17: 81683914-81683914 on Build GRCh38
rs551826459 Chr.17: 81684010-81684010 on Build GRCh38
rs770789495 Chr.17: 81683756-81683756 on Build GRCh38
rs773220555 Chr.17: 81683716-81683716 on Build GRCh38
rs375089359 Chr.17: 81683700-81683700 on Build GRCh38
rs760762564 Chr.17: 81683654-81683654 on Build GRCh38
rs773628468 Chr.17: 81684019-81684019 on Build GRCh38
rs769059840 Chr.17: 81683786-81683786 on Build GRCh38
rs772159723 Chr.17: 81683714-81683714 on Build GRCh38
rs758636311 Chr.17: 81684043-81684043 on Build GRCh38
rs770557358 Chr.17: 81683628-81683628 on Build GRCh38
rs905763770 Chr.17: 81683966-81683966 on Build GRCh38
rs763710819 Chr.17: 81684024-81684024 on Build GRCh38
rs769963539 Chr.17: 81683799-81683799 on Build GRCh38
rs905422628 Chr.17: 81684003-81684003 on Build GRCh38
rs761806625 Chr.17: 81683647-81683647 on Build GRCh38
rs911858153 Chr.17: 81683895-81683895 on Build GRCh38
rs769262666 Chr.17: 81683666-81683666 on Build GRCh38
rs778049575 Chr.17: 81684045-81684045 on Build GRCh38
rs762603530 Chr.17: 81683835-81683835 on Build GRCh38
rs762321709 Chr.17: 81683789-81683789 on Build GRCh38
rs751934742 Chr.17: 81683804-81683805 on Build GRCh38
rs950808659 Chr.17: 81683982-81683982 on Build GRCh38
rs776885646 Chr.17: 81683685-81683685 on Build GRCh38
rs752166339 Chr.17: 81683830-81683830 on Build GRCh38
rs371698739 Chr.17: 81683980-81683981 on Build GRCh38
rs574184462 Chr.17: 81683787-81683787 on Build GRCh38
rs1043899335 Chr.17: 81683928-81683928 on Build GRCh38
rs780348444 Chr.17: 81683803-81683803 on Build GRCh38
rs890486976 Chr.17: 81683959-81683959 on Build GRCh38
rs779483077 Chr.17: 81683658-81683658 on Build GRCh38
rs983232580 Chr.17: 81683906-81683906 on Build GRCh38
rs777499499 Chr.17: 81683752-81683752 on Build GRCh38
rs77221411 Chr.17: 81683814-81683814 on Build GRCh38
rs756982506 Chr.17: 81683842-81683842 on Build GRCh38
rs763447437 Chr.17: 81683744-81683745 on Build GRCh38
rs369598646 Chr.17: 81683797-81683797 on Build GRCh38
rs1011501366 Chr.17: 81683981-81683981 on Build GRCh38
rs77511372 Chr.17: 81683962-81683962 on Build GRCh38
rs773844357 Chr.17: 81684026-81684026 on Build GRCh38
rs781361216 Chr.17: 81684042-81684043 on Build GRCh38
rs9890692 Chr.17: 81683980-81683980 on Build GRCh38
rs8080624 Chr.17: 81683645-81683645 on Build GRCh38

View Full Product Details