Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81809755-81810262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCGR
Entrez Gene ID
2642
Gene Name
glucagon receptor
Gene Aliases
GGR, GL-R
UniGene
Hs.208
Gene Chromosome Location
Chr.17: 81804078-81814013 on Build GRCh38
Transcripts
NM_000160.4 XM_006722277.1 XM_011523539.1 XM_017024446.1 XM_017024447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1589309 c.66G>T p.Q22H Chr.17: 81809787-81809787 on Build GRCh38
4747541 c.151delC p.P52fs*92 Chr.17: 81809872-81809872 on Build GRCh38
5741190 c.163G>A p.E55K Chr.17: 81809884-81809884 on Build GRCh38
3388307 c.150G>T p.L50L Chr.17: 81809871-81809871 on Build GRCh38
4650921 c.88G>C p.D30H Chr.17: 81809809-81809809 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758274454 Chr.17: 81810010-81810010 on Build GRCh38
rs570064871 Chr.17: 81809838-81809838 on Build GRCh38
rs760232106 Chr.17: 81809844-81809844 on Build GRCh38
rs980081455 Chr.17: 81809865-81809865 on Build GRCh38
rs1042501838 Chr.17: 81810211-81810211 on Build GRCh38
rs987433506 Chr.17: 81809992-81809992 on Build GRCh38
rs370781519 Chr.17: 81810128-81810128 on Build GRCh38
rs1005724940 Chr.17: 81810085-81810085 on Build GRCh38
rs906566713 Chr.17: 81810057-81810057 on Build GRCh38
rs984272223 Chr.17: 81809854-81809854 on Build GRCh38
rs1037892197 Chr.17: 81809960-81809960 on Build GRCh38
rs939422845 Chr.17: 81810234-81810234 on Build GRCh38
rs1017028958 Chr.17: 81809873-81809873 on Build GRCh38
rs1020960200 Chr.17: 81809800-81809800 on Build GRCh38
rs1042862392 Chr.17: 81810044-81810044 on Build GRCh38
rs890717084 Chr.17: 81810210-81810210 on Build GRCh38
rs574040622 Chr.17: 81810023-81810023 on Build GRCh38
rs770131289 Chr.17: 81809797-81809797 on Build GRCh38
rs564275276 Chr.17: 81810147-81810147 on Build GRCh38
rs749600573 Chr.17: 81810212-81810212 on Build GRCh38
rs972186244 Chr.17: 81809944-81809944 on Build GRCh38
rs895416034 Chr.17: 81810084-81810084 on Build GRCh38
rs868520740 Chr.17: 81809887-81809887 on Build GRCh38
rs1038260454 Chr.17: 81810161-81810161 on Build GRCh38
rs778317575 Chr.17: 81810182-81810182 on Build GRCh38
rs893609124 Chr.17: 81810031-81810031 on Build GRCh38
rs368731011 Chr.17: 81809990-81809990 on Build GRCh38
rs781270743 Chr.17: 81810092-81810092 on Build GRCh38
rs915116900 Chr.17: 81810204-81810204 on Build GRCh38
rs971096752 Chr.17: 81809835-81809835 on Build GRCh38
rs776227223 Chr.17: 81809883-81809883 on Build GRCh38
rs879515311 Chr.17: 81810068-81810068 on Build GRCh38
rs375299891 Chr.17: 81810203-81810203 on Build GRCh38
rs555478414 Chr.17: 81809959-81809959 on Build GRCh38
rs779357170 Chr.17: 81809890-81809890 on Build GRCh38
rs1031351124 Chr.17: 81810034-81810034 on Build GRCh38
rs945194285 Chr.17: 81810205-81810205 on Build GRCh38
rs149803850 Chr.17: 81810127-81810127 on Build GRCh38
rs1055057090 Chr.17: 81810240-81810240 on Build GRCh38
rs151045413 Chr.17: 81810026-81810026 on Build GRCh38
rs1047803341 Chr.17: 81809928-81809928 on Build GRCh38
rs4796842 Chr.17: 81810078-81810078 on Build GRCh38
rs987151047 Chr.17: 81810157-81810157 on Build GRCh38
rs938741500 Chr.17: 81809895-81809895 on Build GRCh38
rs184752900 Chr.17: 81809950-81809950 on Build GRCh38
rs747611271 Chr.17: 81809807-81809807 on Build GRCh38
rs558314955 Chr.17: 81809784-81809784 on Build GRCh38
rs370693011 Chr.17: 81809895-81809895 on Build GRCh38
rs959556496 Chr.17: 81809847-81809847 on Build GRCh38
rs746310016 Chr.17: 81809785-81809785 on Build GRCh38
rs910188928 Chr.17: 81810169-81810169 on Build GRCh38
rs891521412 Chr.17: 81810215-81810215 on Build GRCh38
rs907927334 Chr.17: 81810128-81810129 on Build GRCh38
rs770253303 Chr.17: 81810176-81810176 on Build GRCh38
rs780718624 Chr.17: 81809802-81809802 on Build GRCh38
rs531331425 Chr.17: 81810179-81810179 on Build GRCh38
rs942151069 Chr.17: 81810152-81810152 on Build GRCh38
rs961843348 Chr.17: 81810107-81810107 on Build GRCh38
rs1801483 Chr.17: 81809839-81809839 on Build GRCh38
rs554158366 Chr.17: 81810029-81810029 on Build GRCh38
rs751811869 Chr.17: 81810035-81810035 on Build GRCh38
rs893913005 Chr.17: 81809796-81809796 on Build GRCh38
rs373371585 Chr.17: 81809882-81809882 on Build GRCh38
rs759393331 Chr.17: 81809947-81809947 on Build GRCh38
rs971874837 Chr.17: 81810125-81810125 on Build GRCh38
rs910451351 Chr.17: 81810016-81810016 on Build GRCh38

View Full Product Details