Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81811470-81811967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCGR
Entrez Gene ID
2642
Gene Name
glucagon receptor
Gene Aliases
GGR, GL-R
UniGene
Hs.208
Gene Chromosome Location
Chr.17: 81804078-81814013 on Build GRCh38
Transcripts
NM_000160.4 XM_006722277.1 XM_011523539.1 XM_017024446.1 XM_017024447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4655105 c.810C>T p.I270I Chr.17: 81811803-81811803 on Build GRCh38
5343059 c.766G>A p.A256T Chr.17: 81811759-81811759 on Build GRCh38
1480174 c.873C>A p.N291K Chr.17: 81811941-81811941 on Build GRCh38
5095113 c.592C>A p.L198I Chr.17: 81811495-81811495 on Build GRCh38
7213752 c.873C>T p.N291N Chr.17: 81811941-81811941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567081976 Chr.17: 81811574-81811574 on Build GRCh38
rs753414714 Chr.17: 81811654-81811654 on Build GRCh38
rs1047414399 Chr.17: 81811688-81811688 on Build GRCh38
rs749623311 Chr.17: 81811746-81811746 on Build GRCh38
rs975953807 Chr.17: 81811712-81811712 on Build GRCh38
rs758992810 Chr.17: 81811793-81811793 on Build GRCh38
rs969117373 Chr.17: 81811870-81811870 on Build GRCh38
rs541382645 Chr.17: 81811817-81811817 on Build GRCh38
rs1013763978 Chr.17: 81811626-81811626 on Build GRCh38
rs931286195 Chr.17: 81811683-81811683 on Build GRCh38
rs768869871 Chr.17: 81811750-81811750 on Build GRCh38
rs758161776 Chr.17: 81811676-81811676 on Build GRCh38
rs368505353 Chr.17: 81811856-81811856 on Build GRCh38
rs545023907 Chr.17: 81811621-81811621 on Build GRCh38
rs769998685 Chr.17: 81811734-81811734 on Build GRCh38
rs540828545 Chr.17: 81811689-81811689 on Build GRCh38
rs367775051 Chr.17: 81811728-81811728 on Build GRCh38
rs762571273 Chr.17: 81811566-81811566 on Build GRCh38
rs576029834 Chr.17: 81811674-81811674 on Build GRCh38
rs886976203 Chr.17: 81811694-81811694 on Build GRCh38
rs995806123 Chr.17: 81811804-81811804 on Build GRCh38
rs775595105 Chr.17: 81811738-81811738 on Build GRCh38
rs866637241 Chr.17: 81811935-81811935 on Build GRCh38
rs77303632 Chr.17: 81811938-81811938 on Build GRCh38
rs969006663 Chr.17: 81811608-81811608 on Build GRCh38
rs999148337 Chr.17: 81811771-81811771 on Build GRCh38
rs918016123 Chr.17: 81811775-81811775 on Build GRCh38
rs887401793 Chr.17: 81811827-81811827 on Build GRCh38
rs377493062 Chr.17: 81811627-81811627 on Build GRCh38
rs867191607 Chr.17: 81811714-81811714 on Build GRCh38
rs776486195 Chr.17: 81811764-81811764 on Build GRCh38
rs774745742 Chr.17: 81811752-81811752 on Build GRCh38
rs772983197 Chr.17: 81811889-81811889 on Build GRCh38
rs758121220 Chr.17: 81811814-81811814 on Build GRCh38
rs569107738 Chr.17: 81811835-81811835 on Build GRCh38
rs775852486 Chr.17: 81811614-81811614 on Build GRCh38
rs917647944 Chr.17: 81811628-81811628 on Build GRCh38
rs778766138 Chr.17: 81811557-81811557 on Build GRCh38
rs371217388 Chr.17: 81811667-81811667 on Build GRCh38
rs762346408 Chr.17: 81811652-81811652 on Build GRCh38
rs945340953 Chr.17: 81811589-81811589 on Build GRCh38
rs555145298 Chr.17: 81811569-81811569 on Build GRCh38
rs879157191 Chr.17: 81811817-81811817 on Build GRCh38
rs756177909 Chr.17: 81811941-81811941 on Build GRCh38
rs781413720 Chr.17: 81811699-81811699 on Build GRCh38
rs371111992 Chr.17: 81811577-81811577 on Build GRCh38
rs769975124 Chr.17: 81811609-81811609 on Build GRCh38
rs980487146 Chr.17: 81811899-81811899 on Build GRCh38
rs1027349740 Chr.17: 81811632-81811632 on Build GRCh38
rs997738885 Chr.17: 81811917-81811917 on Build GRCh38
rs371527112 Chr.17: 81811770-81811770 on Build GRCh38
rs964065567 Chr.17: 81811575-81811575 on Build GRCh38
rs867827368 Chr.17: 81811576-81811576 on Build GRCh38
rs947782180 Chr.17: 81811641-81811641 on Build GRCh38
rs200674527 Chr.17: 81811936-81811936 on Build GRCh38
rs1037221263 Chr.17: 81811801-81811801 on Build GRCh38
rs373969028 Chr.17: 81811776-81811776 on Build GRCh38
rs147520731 Chr.17: 81811902-81811902 on Build GRCh38
rs754634656 Chr.17: 81811528-81811528 on Build GRCh38
rs773836077 Chr.17: 81811821-81811822 on Build GRCh38
rs779642007 Chr.17: 81811524-81811524 on Build GRCh38
rs781285550 Chr.17: 81811928-81811928 on Build GRCh38
rs536915989 Chr.17: 81811527-81811527 on Build GRCh38
rs550972775 Chr.17: 81811805-81811805 on Build GRCh38
rs768423359 Chr.17: 81811722-81811722 on Build GRCh38
rs752264380 Chr.17: 81811812-81811812 on Build GRCh38
rs992975466 Chr.17: 81811831-81811831 on Build GRCh38
rs528738808 Chr.17: 81811684-81811684 on Build GRCh38
rs763972117 Chr.17: 81811845-81811845 on Build GRCh38
rs561479377 Chr.17: 81811678-81811678 on Build GRCh38
rs759823117 Chr.17: 81811607-81811607 on Build GRCh38
rs771987875 Chr.17: 81811570-81811570 on Build GRCh38
rs368152053 Chr.17: 81811803-81811803 on Build GRCh38
rs746261957 Chr.17: 81811653-81811653 on Build GRCh38
rs28454947 Chr.17: 81811590-81811590 on Build GRCh38
rs776062273 Chr.17: 81811779-81811779 on Build GRCh38
rs774870069 Chr.17: 81811629-81811629 on Build GRCh38
rs577644525 Chr.17: 81811620-81811620 on Build GRCh38
rs543300179 Chr.17: 81811677-81811677 on Build GRCh38
rs373598736 Chr.17: 81811675-81811675 on Build GRCh38
rs908475371 Chr.17: 81811942-81811942 on Build GRCh38
rs1007185593 Chr.17: 81811872-81811872 on Build GRCh38
rs899933288 Chr.17: 81811749-81811749 on Build GRCh38
rs903917665 Chr.17: 81811592-81811592 on Build GRCh38
rs1046896088 Chr.17: 81811563-81811563 on Build GRCh38
rs776903275 Chr.17: 81811597-81811597 on Build GRCh38
rs548905669 Chr.17: 81811906-81811906 on Build GRCh38
rs550072079 Chr.17: 81811691-81811691 on Build GRCh38

View Full Product Details