Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81869066-81869557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGDIA
Entrez Gene ID
396
Gene Name
Rho GDP dissociation inhibitor alpha
Gene Aliases
GDIA1, HEL-S-47e, NPHS8, RHOGDI, RHOGDI-1
UniGene
Hs.159161
Gene Chromosome Location
Chr.17: 81867719-81871406 on Build GRCh38
Transcripts
NM_001185077.2 NM_001185078.2 NM_001301240.1 NM_001301241.1 NM_001301242.1 NM_001301243.1 NM_004309.5 NR_125441.1 XM_011523574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1387356 c.337A>C p.K113Q Chr.17: 81869344-81869344 on Build GRCh38
4955940 c.270G>T p.L90L Chr.17: 81869546-81869546 on Build GRCh38
3523691 c.357C>T p.N119N Chr.17: 81869231-81869231 on Build GRCh38
1610985 c.340A>G p.I114V Chr.17: 81869341-81869341 on Build GRCh38
6603756 c.327G>A p.E109E Chr.17: 81869354-81869354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983256374 Chr.17: 81869220-81869220 on Build GRCh38
rs768557957 Chr.17: 81869151-81869151 on Build GRCh38
rs747484778 Chr.17: 81869520-81869520 on Build GRCh38
rs770938892 Chr.17: 81869306-81869306 on Build GRCh38
rs774457316 Chr.17: 81869446-81869446 on Build GRCh38
rs780734602 Chr.17: 81869094-81869094 on Build GRCh38
rs772364864 Chr.17: 81869440-81869441 on Build GRCh38
rs764587753 Chr.17: 81869274-81869274 on Build GRCh38
rs374482246 Chr.17: 81869139-81869139 on Build GRCh38
rs199932528 Chr.17: 81869422-81869422 on Build GRCh38
rs760805237 Chr.17: 81869442-81869459 on Build GRCh38
rs775178310 Chr.17: 81869458-81869458 on Build GRCh38
rs544853396 Chr.17: 81869156-81869156 on Build GRCh38
rs542259070 Chr.17: 81869192-81869192 on Build GRCh38
rs771914015 Chr.17: 81869246-81869246 on Build GRCh38
rs1038500522 Chr.17: 81869423-81869423 on Build GRCh38
rs761900115 Chr.17: 81869117-81869117 on Build GRCh38
rs750339619 Chr.17: 81869499-81869499 on Build GRCh38
rs765561186 Chr.17: 81869189-81869189 on Build GRCh38
rs747030110 Chr.17: 81869262-81869262 on Build GRCh38
rs1055034469 Chr.17: 81869340-81869340 on Build GRCh38
rs202219842 Chr.17: 81869153-81869153 on Build GRCh38
rs370801159 Chr.17: 81869131-81869131 on Build GRCh38
rs201706773 Chr.17: 81869275-81869275 on Build GRCh38
rs372877726 Chr.17: 81869116-81869116 on Build GRCh38
rs577843824 Chr.17: 81869459-81869459 on Build GRCh38
rs556871814 Chr.17: 81869473-81869473 on Build GRCh38
rs955615883 Chr.17: 81869259-81869259 on Build GRCh38
rs764453464 Chr.17: 81869195-81869195 on Build GRCh38
rs764293700 Chr.17: 81869504-81869505 on Build GRCh38
rs767269457 Chr.17: 81869531-81869531 on Build GRCh38
rs368747500 Chr.17: 81869145-81869145 on Build GRCh38
rs772550899 Chr.17: 81869516-81869516 on Build GRCh38
rs758120563 Chr.17: 81869298-81869298 on Build GRCh38
rs762398266 Chr.17: 81869491-81869491 on Build GRCh38
rs932073483 Chr.17: 81869106-81869106 on Build GRCh38
rs369031018 Chr.17: 81869447-81869447 on Build GRCh38
rs778379704 Chr.17: 81869140-81869140 on Build GRCh38
rs1056817094 Chr.17: 81869533-81869533 on Build GRCh38
rs757075945 Chr.17: 81869494-81869494 on Build GRCh38
rs199798201 Chr.17: 81869266-81869266 on Build GRCh38
rs780157115 Chr.17: 81869443-81869443 on Build GRCh38
rs553259609 Chr.17: 81869430-81869430 on Build GRCh38
rs138551115 Chr.17: 81869270-81869270 on Build GRCh38
rs775694555 Chr.17: 81869441-81869442 on Build GRCh38
rs779123568 Chr.17: 81869511-81869511 on Build GRCh38
rs541288466 Chr.17: 81869119-81869119 on Build GRCh38
rs143378131 Chr.17: 81869401-81869401 on Build GRCh38
rs746895642 Chr.17: 81869305-81869305 on Build GRCh38
rs560565688 Chr.17: 81869524-81869524 on Build GRCh38
rs761503758 Chr.17: 81869530-81869530 on Build GRCh38
rs991176308 Chr.17: 81869526-81869526 on Build GRCh38
rs963283196 Chr.17: 81869242-81869242 on Build GRCh38
rs779674387 Chr.17: 81869207-81869207 on Build GRCh38
rs754555534 Chr.17: 81869283-81869283 on Build GRCh38
rs932025446 Chr.17: 81869253-81869253 on Build GRCh38
rs370209119 Chr.17: 81869300-81869300 on Build GRCh38
rs759048894 Chr.17: 81869402-81869402 on Build GRCh38
rs1051340659 Chr.17: 81869277-81869277 on Build GRCh38
rs377159229 Chr.17: 81869109-81869109 on Build GRCh38
rs769295286 Chr.17: 81869096-81869096 on Build GRCh38
rs373807389 Chr.17: 81869514-81869514 on Build GRCh38
rs749341262 Chr.17: 81869509-81869509 on Build GRCh38
rs761853814 Chr.17: 81869535-81869535 on Build GRCh38
rs767361688 Chr.17: 81869495-81869495 on Build GRCh38
rs759807623 Chr.17: 81869163-81869163 on Build GRCh38
rs747901067 Chr.17: 81869243-81869243 on Build GRCh38
rs375714223 Chr.17: 81869444-81869444 on Build GRCh38
rs939284909 Chr.17: 81869319-81869319 on Build GRCh38
rs990337929 Chr.17: 81869475-81869475 on Build GRCh38
rs573987770 Chr.17: 81869114-81869114 on Build GRCh38
rs755356784 Chr.17: 81869091-81869091 on Build GRCh38
rs767684802 Chr.17: 81869118-81869118 on Build GRCh38
rs754100711 Chr.17: 81869126-81869126 on Build GRCh38
rs144351801 Chr.17: 81869291-81869291 on Build GRCh38
rs200795980 Chr.17: 81869293-81869293 on Build GRCh38
rs753842528 Chr.17: 81869505-81869505 on Build GRCh38
rs759646191 Chr.17: 81869269-81869269 on Build GRCh38
rs752380567 Chr.17: 81869416-81869416 on Build GRCh38
rs574995862 Chr.17: 81869486-81869486 on Build GRCh38
rs769871113 Chr.17: 81869331-81869331 on Build GRCh38
rs201049488 Chr.17: 81869159-81869159 on Build GRCh38
rs765300609 Chr.17: 81869538-81869538 on Build GRCh38
rs200606006 Chr.17: 81869162-81869162 on Build GRCh38
rs772569168 Chr.17: 81869449-81869449 on Build GRCh38
rs112190652 Chr.17: 81869252-81869252 on Build GRCh38
rs566979183 Chr.17: 81869277-81869278 on Build GRCh38
rs560498599 Chr.17: 81869197-81869197 on Build GRCh38
rs781133009 Chr.17: 81869309-81869309 on Build GRCh38
rs751178380 Chr.17: 81869493-81869493 on Build GRCh38
rs1017004857 Chr.17: 81869239-81869239 on Build GRCh38
rs372602369 Chr.17: 81869426-81869426 on Build GRCh38
rs1029682325 Chr.17: 81869436-81869436 on Build GRCh38
rs780000161 Chr.17: 81869505-81869505 on Build GRCh38
rs753357203 Chr.17: 81869279-81869279 on Build GRCh38
rs755667117 Chr.17: 81869146-81869146 on Build GRCh38
rs749038625 Chr.17: 81869147-81869147 on Build GRCh38
rs1055384334 Chr.17: 81869175-81869175 on Build GRCh38
rs777396664 Chr.17: 81869424-81869424 on Build GRCh38
rs773849890 Chr.17: 81869453-81869453 on Build GRCh38
rs149223274 Chr.17: 81869399-81869399 on Build GRCh38
rs371966890 Chr.17: 81869231-81869231 on Build GRCh38
rs1018520979 Chr.17: 81869468-81869468 on Build GRCh38
rs542316722 Chr.17: 81869485-81869485 on Build GRCh38
rs202224232 Chr.17: 81869518-81869518 on Build GRCh38
rs113301975 Chr.17: 81869287-81869287 on Build GRCh38
rs543875480 Chr.17: 81869219-81869219 on Build GRCh38
rs763747796 Chr.17: 81869492-81869492 on Build GRCh38
rs748656815 Chr.17: 81869093-81869093 on Build GRCh38
rs759924169 Chr.17: 81869412-81869414 on Build GRCh38
rs367718991 Chr.17: 81869216-81869216 on Build GRCh38
rs765777308 Chr.17: 81869276-81869276 on Build GRCh38
rs897244624 Chr.17: 81869425-81869425 on Build GRCh38
rs774821533 Chr.17: 81869363-81869363 on Build GRCh38
rs1021312970 Chr.17: 81869174-81869174 on Build GRCh38
rs775232711 Chr.17: 81869098-81869098 on Build GRCh38
rs1007575764 Chr.17: 81869457-81869457 on Build GRCh38
rs762117986 Chr.17: 81869394-81869394 on Build GRCh38
rs528768290 Chr.17: 81869204-81869204 on Build GRCh38
rs545229548 Chr.17: 81869465-81869465 on Build GRCh38
rs745506650 Chr.17: 81869095-81869095 on Build GRCh38
rs200342668 Chr.17: 81869129-81869129 on Build GRCh38
rs1011125421 Chr.17: 81869166-81869166 on Build GRCh38
rs745875773 Chr.17: 81869437-81869437 on Build GRCh38
rs768880419 Chr.17: 81869350-81869350 on Build GRCh38
rs935382178 Chr.17: 81869273-81869273 on Build GRCh38
rs1029628347 Chr.17: 81869256-81869256 on Build GRCh38
rs566838646 Chr.17: 81869322-81869322 on Build GRCh38
rs764906274 Chr.17: 81869404-81869404 on Build GRCh38
rs181424923 Chr.17: 81869528-81869528 on Build GRCh38
rs775688385 Chr.17: 81869337-81869337 on Build GRCh38
rs370172286 Chr.17: 81869155-81869155 on Build GRCh38
rs756174247 Chr.17: 81869439-81869439 on Build GRCh38
rs941234852 Chr.17: 81869534-81869534 on Build GRCh38
rs762600270 Chr.17: 81869110-81869110 on Build GRCh38
rs763294984 Chr.17: 81869339-81869339 on Build GRCh38
rs372484327 Chr.17: 81869286-81869286 on Build GRCh38
rs755926987 Chr.17: 81869532-81869532 on Build GRCh38
rs907672631 Chr.17: 81869228-81869228 on Build GRCh38
rs760612373 Chr.17: 81869161-81869161 on Build GRCh38
rs587777553 Chr.17: 81869230-81869230 on Build GRCh38

View Full Product Details