Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81869212-81869707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGDIA
Entrez Gene ID
396
Gene Name
Rho GDP dissociation inhibitor alpha
Gene Aliases
GDIA1, HEL-S-47e, NPHS8, RHOGDI, RHOGDI-1
UniGene
Hs.159161
Gene Chromosome Location
Chr.17: 81867719-81871406 on Build GRCh38
Transcripts
NM_001185077.2 NM_001185078.2 NM_001301240.1 NM_001301241.1 NM_001301242.1 NM_001301243.1 NM_004309.5 NR_125441.1 XM_011523574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1387356 c.337A>C p.K113Q Chr.17: 81869344-81869344 on Build GRCh38
4955940 c.270G>T p.L90L Chr.17: 81869546-81869546 on Build GRCh38
3523691 c.357C>T p.N119N Chr.17: 81869231-81869231 on Build GRCh38
3523692 c.253C>T p.P85S Chr.17: 81869563-81869563 on Build GRCh38
1610985 c.340A>G p.I114V Chr.17: 81869341-81869341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747484778 Chr.17: 81869520-81869520 on Build GRCh38
rs770938892 Chr.17: 81869306-81869306 on Build GRCh38
rs774457316 Chr.17: 81869446-81869446 on Build GRCh38
rs772364864 Chr.17: 81869440-81869441 on Build GRCh38
rs764587753 Chr.17: 81869274-81869274 on Build GRCh38
rs368364854 Chr.17: 81869676-81869676 on Build GRCh38
rs199932528 Chr.17: 81869422-81869422 on Build GRCh38
rs760805237 Chr.17: 81869442-81869459 on Build GRCh38
rs775178310 Chr.17: 81869458-81869458 on Build GRCh38
rs771914015 Chr.17: 81869246-81869246 on Build GRCh38
rs1038500522 Chr.17: 81869423-81869423 on Build GRCh38
rs750339619 Chr.17: 81869499-81869499 on Build GRCh38
rs758730597 Chr.17: 81869568-81869568 on Build GRCh38
rs867948642 Chr.17: 81869608-81869608 on Build GRCh38
rs375109586 Chr.17: 81869664-81869664 on Build GRCh38
rs747030110 Chr.17: 81869262-81869262 on Build GRCh38
rs1055034469 Chr.17: 81869340-81869340 on Build GRCh38
rs369582831 Chr.17: 81869669-81869669 on Build GRCh38
rs773774745 Chr.17: 81869579-81869579 on Build GRCh38
rs201706773 Chr.17: 81869275-81869275 on Build GRCh38
rs755080375 Chr.17: 81869563-81869563 on Build GRCh38
rs200283878 Chr.17: 81869617-81869617 on Build GRCh38
rs766153339 Chr.17: 81869621-81869621 on Build GRCh38
rs577843824 Chr.17: 81869459-81869459 on Build GRCh38
rs556871814 Chr.17: 81869473-81869473 on Build GRCh38
rs955615883 Chr.17: 81869259-81869259 on Build GRCh38
rs764293700 Chr.17: 81869504-81869505 on Build GRCh38
rs767269457 Chr.17: 81869531-81869531 on Build GRCh38
rs772550899 Chr.17: 81869516-81869516 on Build GRCh38
rs758120563 Chr.17: 81869298-81869298 on Build GRCh38
rs762398266 Chr.17: 81869491-81869491 on Build GRCh38
rs369031018 Chr.17: 81869447-81869447 on Build GRCh38
rs1029211053 Chr.17: 81869655-81869655 on Build GRCh38
rs759440871 Chr.17: 81869647-81869647 on Build GRCh38
rs1056817094 Chr.17: 81869533-81869533 on Build GRCh38
rs757075945 Chr.17: 81869494-81869494 on Build GRCh38
rs780157115 Chr.17: 81869443-81869443 on Build GRCh38
rs553259609 Chr.17: 81869430-81869430 on Build GRCh38
rs199798201 Chr.17: 81869266-81869266 on Build GRCh38
rs138551115 Chr.17: 81869270-81869270 on Build GRCh38
rs775694555 Chr.17: 81869441-81869442 on Build GRCh38
rs779123568 Chr.17: 81869511-81869511 on Build GRCh38
rs143378131 Chr.17: 81869401-81869401 on Build GRCh38
rs575313894 Chr.17: 81869637-81869637 on Build GRCh38
rs201081362 Chr.17: 81869577-81869577 on Build GRCh38
rs746895642 Chr.17: 81869305-81869305 on Build GRCh38
rs560565688 Chr.17: 81869524-81869524 on Build GRCh38
rs761503758 Chr.17: 81869530-81869530 on Build GRCh38
rs991176308 Chr.17: 81869526-81869526 on Build GRCh38
rs963283196 Chr.17: 81869242-81869242 on Build GRCh38
rs754555534 Chr.17: 81869283-81869283 on Build GRCh38
rs752937765 Chr.17: 81869564-81869564 on Build GRCh38
rs370209119 Chr.17: 81869300-81869300 on Build GRCh38
rs932025446 Chr.17: 81869253-81869253 on Build GRCh38
rs759048894 Chr.17: 81869402-81869402 on Build GRCh38
rs1051340659 Chr.17: 81869277-81869277 on Build GRCh38
rs373807389 Chr.17: 81869514-81869514 on Build GRCh38
rs749341262 Chr.17: 81869509-81869509 on Build GRCh38
rs767361688 Chr.17: 81869495-81869495 on Build GRCh38
rs761853814 Chr.17: 81869535-81869535 on Build GRCh38
rs1004492781 Chr.17: 81869587-81869587 on Build GRCh38
rs375714223 Chr.17: 81869444-81869444 on Build GRCh38
rs747901067 Chr.17: 81869243-81869243 on Build GRCh38
rs939284909 Chr.17: 81869319-81869319 on Build GRCh38
rs990337929 Chr.17: 81869475-81869475 on Build GRCh38
rs201919353 Chr.17: 81869656-81869656 on Build GRCh38
rs144351801 Chr.17: 81869291-81869291 on Build GRCh38
rs200795980 Chr.17: 81869293-81869293 on Build GRCh38
rs761344643 Chr.17: 81869594-81869594 on Build GRCh38
rs753842528 Chr.17: 81869505-81869505 on Build GRCh38
rs956712978 Chr.17: 81869679-81869679 on Build GRCh38
rs759646191 Chr.17: 81869269-81869269 on Build GRCh38
rs752380567 Chr.17: 81869416-81869416 on Build GRCh38
rs769871113 Chr.17: 81869331-81869331 on Build GRCh38
rs574995862 Chr.17: 81869486-81869486 on Build GRCh38
rs758277491 Chr.17: 81869606-81869614 on Build GRCh38
rs765300609 Chr.17: 81869538-81869538 on Build GRCh38
rs112190652 Chr.17: 81869252-81869252 on Build GRCh38
rs772569168 Chr.17: 81869449-81869449 on Build GRCh38
rs566979183 Chr.17: 81869277-81869278 on Build GRCh38
rs781133009 Chr.17: 81869309-81869309 on Build GRCh38
rs751178380 Chr.17: 81869493-81869493 on Build GRCh38
rs1017004857 Chr.17: 81869239-81869239 on Build GRCh38
rs372602369 Chr.17: 81869426-81869426 on Build GRCh38
rs1029682325 Chr.17: 81869436-81869436 on Build GRCh38
rs780000161 Chr.17: 81869505-81869505 on Build GRCh38
rs753357203 Chr.17: 81869279-81869279 on Build GRCh38
rs777396664 Chr.17: 81869424-81869424 on Build GRCh38
rs773849890 Chr.17: 81869453-81869453 on Build GRCh38
rs149223274 Chr.17: 81869399-81869399 on Build GRCh38
rs779924883 Chr.17: 81869672-81869682 on Build GRCh38
rs11546986 Chr.17: 81869573-81869573 on Build GRCh38
rs1018520979 Chr.17: 81869468-81869468 on Build GRCh38
rs542316722 Chr.17: 81869485-81869485 on Build GRCh38
rs202224232 Chr.17: 81869518-81869518 on Build GRCh38
rs113301975 Chr.17: 81869287-81869287 on Build GRCh38
rs532695235 Chr.17: 81869619-81869619 on Build GRCh38
rs763747796 Chr.17: 81869492-81869492 on Build GRCh38
rs759924169 Chr.17: 81869412-81869414 on Build GRCh38
rs765777308 Chr.17: 81869276-81869276 on Build GRCh38
rs371636830 Chr.17: 81869618-81869618 on Build GRCh38
rs897244624 Chr.17: 81869425-81869425 on Build GRCh38
rs774821533 Chr.17: 81869363-81869363 on Build GRCh38
rs750341988 Chr.17: 81869540-81869540 on Build GRCh38
rs762117986 Chr.17: 81869394-81869394 on Build GRCh38
rs1007575764 Chr.17: 81869457-81869457 on Build GRCh38
rs545229548 Chr.17: 81869465-81869465 on Build GRCh38
rs753799299 Chr.17: 81869641-81869641 on Build GRCh38
rs745875773 Chr.17: 81869437-81869437 on Build GRCh38
rs747325364 Chr.17: 81869571-81869571 on Build GRCh38
rs768880419 Chr.17: 81869350-81869350 on Build GRCh38
rs935382178 Chr.17: 81869273-81869273 on Build GRCh38
rs1029628347 Chr.17: 81869256-81869256 on Build GRCh38
rs566838646 Chr.17: 81869322-81869322 on Build GRCh38
rs1038216708 Chr.17: 81869539-81869539 on Build GRCh38
rs764906274 Chr.17: 81869404-81869404 on Build GRCh38
rs181424923 Chr.17: 81869528-81869528 on Build GRCh38
rs775688385 Chr.17: 81869337-81869337 on Build GRCh38
rs11546992 Chr.17: 81869560-81869560 on Build GRCh38
rs756174247 Chr.17: 81869439-81869439 on Build GRCh38
rs941234852 Chr.17: 81869534-81869534 on Build GRCh38
rs763294984 Chr.17: 81869339-81869339 on Build GRCh38
rs771522183 Chr.17: 81869572-81869572 on Build GRCh38
rs563752119 Chr.17: 81869574-81869574 on Build GRCh38
rs372484327 Chr.17: 81869286-81869286 on Build GRCh38
rs778169808 Chr.17: 81869569-81869569 on Build GRCh38
rs755926987 Chr.17: 81869532-81869532 on Build GRCh38
rs771665592 Chr.17: 81869610-81869610 on Build GRCh38

View Full Product Details