Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82239420-82239918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1D
Entrez Gene ID
1453
Gene Name
casein kinase 1 delta
Gene Aliases
ASPS, CKIdelta, FASPS2, HCKID
UniGene
Hs.631725
Gene Chromosome Location
Chr.17: 82239317-82273742 on Build GRCh38
Transcripts
XM_005256337.4 NM_001042422.2 NM_001042423.2 NM_001206950.1 NM_001206951.1 NM_001206952.1 NM_004207.3 XM_011523607.1

Gene Symbol
SLC16A3
Entrez Gene ID
9123
Gene Name
solute carrier family 16 member 3
Gene Aliases
MCT 3, MCT 4, MCT-3, MCT-4, MCT3, MCT4
UniGene
Hs.500761
Gene Chromosome Location
Chr.17: 82228406-82239499 on Build GRCh38
Transcripts
XM_005256337.4 NM_001042422.2 NM_001042423.2 NM_001206950.1 NM_001206951.1 NM_001206952.1 NM_004207.3 XM_011523607.1

SNP Detail

SNP ID Chromosome Location
rs779670163 Chr.17: 82239758-82239758 on Build GRCh38
rs115739233 Chr.17: 82239774-82239774 on Build GRCh38
rs983916004 Chr.17: 82239794-82239794 on Build GRCh38
rs974740496 Chr.17: 82239812-82239812 on Build GRCh38
rs938247310 Chr.17: 82239623-82239623 on Build GRCh38
rs551310818 Chr.17: 82239500-82239500 on Build GRCh38
rs549487338 Chr.17: 82239579-82239579 on Build GRCh38
rs1013862472 Chr.17: 82239728-82239728 on Build GRCh38
rs183811963 Chr.17: 82239533-82239533 on Build GRCh38
rs767911665 Chr.17: 82239487-82239487 on Build GRCh38
rs911840882 Chr.17: 82239624-82239624 on Build GRCh38
rs545820765 Chr.17: 82239508-82239508 on Build GRCh38
rs112994596 Chr.17: 82239629-82239629 on Build GRCh38
rs113201258 Chr.17: 82239803-82239803 on Build GRCh38
rs976297828 Chr.17: 82239735-82239735 on Build GRCh38
rs745552148 Chr.17: 82239597-82239597 on Build GRCh38
rs996692732 Chr.17: 82239490-82239490 on Build GRCh38
rs113940765 Chr.17: 82239853-82239853 on Build GRCh38
rs925755101 Chr.17: 82239878-82239878 on Build GRCh38
rs920800701 Chr.17: 82239608-82239608 on Build GRCh38
rs951914420 Chr.17: 82239560-82239560 on Build GRCh38
rs778682723 Chr.17: 82239574-82239574 on Build GRCh38
rs571451542 Chr.17: 82239872-82239872 on Build GRCh38
rs911991471 Chr.17: 82239796-82239796 on Build GRCh38
rs373342211 Chr.17: 82239773-82239773 on Build GRCh38
rs9895071 Chr.17: 82239523-82239523 on Build GRCh38
rs899462301 Chr.17: 82239541-82239552 on Build GRCh38
rs567878426 Chr.17: 82239616-82239616 on Build GRCh38
rs929372853 Chr.17: 82239609-82239609 on Build GRCh38
rs752007012 Chr.17: 82239443-82239443 on Build GRCh38
rs962570905 Chr.17: 82239732-82239732 on Build GRCh38
rs991488494 Chr.17: 82239840-82239840 on Build GRCh38
rs370020643 Chr.17: 82239477-82239477 on Build GRCh38
rs921963516 Chr.17: 82239830-82239830 on Build GRCh38
rs998973774 Chr.17: 82239705-82239705 on Build GRCh38
rs757974913 Chr.17: 82239662-82239662 on Build GRCh38
rs949547268 Chr.17: 82239630-82239630 on Build GRCh38
rs895443341 Chr.17: 82239719-82239719 on Build GRCh38
rs113780791 Chr.17: 82239511-82239512 on Build GRCh38
rs777286404 Chr.17: 82239485-82239485 on Build GRCh38
rs904365421 Chr.17: 82239696-82239696 on Build GRCh38
rs756569112 Chr.17: 82239556-82239556 on Build GRCh38
rs929022558 Chr.17: 82239886-82239886 on Build GRCh38
rs993177440 Chr.17: 82239702-82239702 on Build GRCh38
rs1037176322 Chr.17: 82239874-82239874 on Build GRCh38
rs918138112 Chr.17: 82239850-82239850 on Build GRCh38
rs959737681 Chr.17: 82239539-82239539 on Build GRCh38
rs1007422888 Chr.17: 82239564-82239564 on Build GRCh38
rs1017838881 Chr.17: 82239569-82239570 on Build GRCh38
rs925266523 Chr.17: 82239820-82239820 on Build GRCh38
rs887718572 Chr.17: 82239769-82239769 on Build GRCh38
rs1028203039 Chr.17: 82239787-82239787 on Build GRCh38
rs895548800 Chr.17: 82239897-82239897 on Build GRCh38
rs112716395 Chr.17: 82239481-82239481 on Build GRCh38
rs1043963505 Chr.17: 82239496-82239496 on Build GRCh38
rs879668263 Chr.17: 82239677-82239678 on Build GRCh38
rs1023187070 Chr.17: 82239563-82239563 on Build GRCh38
rs919038163 Chr.17: 82239883-82239883 on Build GRCh38
rs904074482 Chr.17: 82239498-82239499 on Build GRCh38
rs1131633 Chr.17: 82239473-82239473 on Build GRCh38
rs1053392098 Chr.17: 82239892-82239892 on Build GRCh38
rs1021494540 Chr.17: 82239495-82239495 on Build GRCh38
rs1042905428 Chr.17: 82239694-82239694 on Build GRCh38
rs542120117 Chr.17: 82239460-82239460 on Build GRCh38
rs1040952751 Chr.17: 82239631-82239631 on Build GRCh38
rs1031721451 Chr.17: 82239536-82239536 on Build GRCh38
rs746517344 Chr.17: 82239881-82239881 on Build GRCh38
rs970280743 Chr.17: 82239765-82239765 on Build GRCh38
rs894252020 Chr.17: 82239526-82239526 on Build GRCh38
rs938197883 Chr.17: 82239511-82239512 on Build GRCh38
rs976563812 Chr.17: 82239596-82239596 on Build GRCh38
rs976134120 Chr.17: 82239475-82239475 on Build GRCh38
rs966734102 Chr.17: 82239811-82239811 on Build GRCh38
rs932597259 Chr.17: 82239666-82239666 on Build GRCh38
rs368852326 Chr.17: 82239793-82239793 on Build GRCh38
rs908124603 Chr.17: 82239718-82239718 on Build GRCh38
rs1048745638 Chr.17: 82239742-82239742 on Build GRCh38
rs1048189010 Chr.17: 82239459-82239459 on Build GRCh38
rs970313148 Chr.17: 82239569-82239569 on Build GRCh38
rs899642506 Chr.17: 82239486-82239486 on Build GRCh38
rs966194324 Chr.17: 82239575-82239575 on Build GRCh38
rs979429000 Chr.17: 82239816-82239816 on Build GRCh38
rs929429774 Chr.17: 82239446-82239446 on Build GRCh38
rs1031439166 Chr.17: 82239711-82239711 on Build GRCh38

View Full Product Details