Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82389018-82389514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OGFOD3
Entrez Gene ID
79701
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 3
Gene Aliases
C17orf101
UniGene
Hs.187505
Gene Chromosome Location
Chr.17: 82389210-82418637 on Build GRCh38
Transcripts
NM_024648.2 NM_175902.4 NR_033265.1

SNP Detail

SNP ID Chromosome Location
rs138996814 Chr.17: 82389173-82389173 on Build GRCh38
rs997107465 Chr.17: 82389081-82389081 on Build GRCh38
rs1021052791 Chr.17: 82389356-82389356 on Build GRCh38
rs923897571 Chr.17: 82389074-82389074 on Build GRCh38
rs1026368924 Chr.17: 82389037-82389037 on Build GRCh38
rs1009684359 Chr.17: 82389108-82389108 on Build GRCh38
rs982069945 Chr.17: 82389128-82389128 on Build GRCh38
rs933104828 Chr.17: 82389463-82389463 on Build GRCh38
rs928420941 Chr.17: 82389340-82389340 on Build GRCh38
rs919014822 Chr.17: 82389439-82389439 on Build GRCh38
rs1021021663 Chr.17: 82389109-82389109 on Build GRCh38
rs532132529 Chr.17: 82389289-82389289 on Build GRCh38
rs938538165 Chr.17: 82389347-82389347 on Build GRCh38
rs889334464 Chr.17: 82389083-82389083 on Build GRCh38
rs184092562 Chr.17: 82389212-82389212 on Build GRCh38
rs893988560 Chr.17: 82389326-82389326 on Build GRCh38
rs558925581 Chr.17: 82389085-82389085 on Build GRCh38
rs114749072 Chr.17: 82389382-82389382 on Build GRCh38
rs113618368 Chr.17: 82389284-82389284 on Build GRCh38
rs1051337791 Chr.17: 82389291-82389291 on Build GRCh38
rs554768379 Chr.17: 82389058-82389058 on Build GRCh38
rs557329058 Chr.17: 82389154-82389154 on Build GRCh38
rs991448843 Chr.17: 82389392-82389392 on Build GRCh38
rs1008148880 Chr.17: 82389084-82389084 on Build GRCh38
rs577961430 Chr.17: 82389107-82389107 on Build GRCh38
rs1023566976 Chr.17: 82389462-82389462 on Build GRCh38
rs1009716248 Chr.17: 82389339-82389339 on Build GRCh38
rs897523007 Chr.17: 82389069-82389069 on Build GRCh38
rs970768714 Chr.17: 82389365-82389365 on Build GRCh38
rs76038896 Chr.17: 82389219-82389219 on Build GRCh38
rs937174385 Chr.17: 82389046-82389046 on Build GRCh38
rs1005927579 Chr.17: 82389129-82389129 on Build GRCh38
rs959619452 Chr.17: 82389389-82389389 on Build GRCh38
rs144047275 Chr.17: 82389082-82389082 on Build GRCh38
rs971830208 Chr.17: 82389437-82389437 on Build GRCh38
rs778748914 Chr.17: 82389239-82389239 on Build GRCh38
rs752190797 Chr.17: 82389045-82389045 on Build GRCh38
rs754182592 Chr.17: 82389489-82389489 on Build GRCh38
rs909092478 Chr.17: 82389403-82389403 on Build GRCh38
rs1003623877 Chr.17: 82389371-82389371 on Build GRCh38
rs978713524 Chr.17: 82389225-82389225 on Build GRCh38
rs1047645576 Chr.17: 82389128-82389130 on Build GRCh38
rs777595518 Chr.17: 82389055-82389055 on Build GRCh38
rs941844641 Chr.17: 82389209-82389209 on Build GRCh38
rs886398615 Chr.17: 82389357-82389357 on Build GRCh38
rs916648567 Chr.17: 82389310-82389310 on Build GRCh38
rs999738231 Chr.17: 82389397-82389397 on Build GRCh38
rs77157205 Chr.17: 82389130-82389130 on Build GRCh38
rs116916712 Chr.17: 82389313-82389313 on Build GRCh38
rs1030088312 Chr.17: 82389391-82389391 on Build GRCh38
rs549362525 Chr.17: 82389090-82389090 on Build GRCh38
rs565280470 Chr.17: 82389287-82389287 on Build GRCh38
rs989702281 Chr.17: 82389441-82389441 on Build GRCh38
rs755610248 Chr.17: 82389053-82389053 on Build GRCh38
rs566654553 Chr.17: 82389455-82389455 on Build GRCh38
rs115300345 Chr.17: 82389411-82389411 on Build GRCh38
rs74001662 Chr.17: 82389187-82389187 on Build GRCh38
rs958308451 Chr.17: 82389414-82389414 on Build GRCh38
rs550263795 Chr.17: 82389312-82389312 on Build GRCh38
rs1048100294 Chr.17: 82389349-82389349 on Build GRCh38
rs1037400801 Chr.17: 82389372-82389372 on Build GRCh38
rs900183629 Chr.17: 82389381-82389381 on Build GRCh38
rs916702435 Chr.17: 82389086-82389086 on Build GRCh38
rs753753087 Chr.17: 82389065-82389065 on Build GRCh38
rs1036081495 Chr.17: 82389268-82389268 on Build GRCh38
rs958028861 Chr.17: 82389293-82389293 on Build GRCh38
rs888770699 Chr.17: 82389294-82389294 on Build GRCh38
rs116606308 Chr.17: 82389064-82389064 on Build GRCh38
rs928513777 Chr.17: 82389040-82389040 on Build GRCh38
rs970736249 Chr.17: 82389110-82389110 on Build GRCh38
rs780288283 Chr.17: 82389325-82389325 on Build GRCh38
rs1007849078 Chr.17: 82389303-82389303 on Build GRCh38

View Full Product Details