Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82409226-82409643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OGFOD3
Entrez Gene ID
79701
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 3
Gene Aliases
C17orf101
UniGene
Hs.187505
Gene Chromosome Location
Chr.17: 82389210-82418637 on Build GRCh38
Transcripts
NM_024648.2 NM_175902.4 NR_033265.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5955763 c.417C>A p.D139E Chr.17: 82409374-82409374 on Build GRCh38
4545902 c.388G>A p.E130K Chr.17: 82409403-82409403 on Build GRCh38
1242047 c.387T>C p.A129A Chr.17: 82409404-82409404 on Build GRCh38
4547309 c.419G>A p.G140E Chr.17: 82409372-82409372 on Build GRCh38
4547438 c.422G>A p.G141E Chr.17: 82409369-82409369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs62080965 Chr.17: 82409330-82409330 on Build GRCh38
rs537561732 Chr.17: 82409507-82409507 on Build GRCh38
rs764858384 Chr.17: 82409443-82409443 on Build GRCh38
rs532280769 Chr.17: 82409449-82409449 on Build GRCh38
rs918486226 Chr.17: 82409284-82409284 on Build GRCh38
rs745900788 Chr.17: 82409602-82409602 on Build GRCh38
rs750880502 Chr.17: 82409363-82409363 on Build GRCh38
rs770914794 Chr.17: 82409363-82409363 on Build GRCh38
rs907272298 Chr.17: 82409251-82409251 on Build GRCh38
rs556928356 Chr.17: 82409587-82409587 on Build GRCh38
rs942455380 Chr.17: 82409508-82409508 on Build GRCh38
rs764581519 Chr.17: 82409346-82409346 on Build GRCh38
rs141382926 Chr.17: 82409338-82409338 on Build GRCh38
rs947075408 Chr.17: 82409552-82409552 on Build GRCh38
rs553294740 Chr.17: 82409271-82409271 on Build GRCh38
rs796832947 Chr.17: 82409340-82409341 on Build GRCh38
rs914287689 Chr.17: 82409551-82409551 on Build GRCh38
rs538930896 Chr.17: 82409586-82409586 on Build GRCh38
rs991902085 Chr.17: 82409266-82409266 on Build GRCh38
rs775789220 Chr.17: 82409336-82409336 on Build GRCh38
rs143751196 Chr.17: 82409395-82409395 on Build GRCh38
rs973992487 Chr.17: 82409279-82409279 on Build GRCh38
rs542320999 Chr.17: 82409374-82409374 on Build GRCh38
rs774365029 Chr.17: 82409366-82409367 on Build GRCh38
rs767991456 Chr.17: 82409357-82409357 on Build GRCh38
rs749594027 Chr.17: 82409416-82409416 on Build GRCh38
rs898435872 Chr.17: 82409332-82409333 on Build GRCh38
rs762189090 Chr.17: 82409430-82409430 on Build GRCh38
rs773372479 Chr.17: 82409340-82409341 on Build GRCh38
rs767829194 Chr.17: 82409438-82409438 on Build GRCh38
rs757971603 Chr.17: 82409389-82409389 on Build GRCh38
rs182478202 Chr.17: 82409545-82409545 on Build GRCh38
rs765985805 Chr.17: 82409277-82409277 on Build GRCh38
rs376815146 Chr.17: 82409529-82409529 on Build GRCh38
rs779097339 Chr.17: 82409601-82409601 on Build GRCh38
rs199683954 Chr.17: 82409329-82409329 on Build GRCh38
rs553131157 Chr.17: 82409580-82409580 on Build GRCh38
rs534921898 Chr.17: 82409565-82409565 on Build GRCh38
rs762348800 Chr.17: 82409356-82409356 on Build GRCh38
rs147643450 Chr.17: 82409607-82409607 on Build GRCh38
rs769803318 Chr.17: 82409334-82409334 on Build GRCh38
rs573854897 Chr.17: 82409254-82409254 on Build GRCh38
rs145224873 Chr.17: 82409556-82409556 on Build GRCh38
rs774747744 Chr.17: 82409428-82409428 on Build GRCh38
rs1050875584 Chr.17: 82409315-82409315 on Build GRCh38
rs117643494 Chr.17: 82409259-82409259 on Build GRCh38
rs774696299 Chr.17: 82409355-82409355 on Build GRCh38
rs7211306 Chr.17: 82409477-82409477 on Build GRCh38
rs201971168 Chr.17: 82409386-82409386 on Build GRCh38
rs112823833 Chr.17: 82409455-82409455 on Build GRCh38
rs981588848 Chr.17: 82409249-82409249 on Build GRCh38
rs185665735 Chr.17: 82409581-82409581 on Build GRCh38
rs907959908 Chr.17: 82409534-82409534 on Build GRCh38
rs777774451 Chr.17: 82409391-82409391 on Build GRCh38
rs929852790 Chr.17: 82409294-82409294 on Build GRCh38
rs183658728 Chr.17: 82409414-82409414 on Build GRCh38
rs371562893 Chr.17: 82409516-82409516 on Build GRCh38
rs140153882 Chr.17: 82409367-82409367 on Build GRCh38
rs769710455 Chr.17: 82409333-82409333 on Build GRCh38
rs777975351 Chr.17: 82409412-82409412 on Build GRCh38
rs1053459798 Chr.17: 82409253-82409253 on Build GRCh38
rs143458096 Chr.17: 82409410-82409410 on Build GRCh38
rs768731209 Chr.17: 82409424-82409424 on Build GRCh38
rs112176293 Chr.17: 82409573-82409573 on Build GRCh38
rs372617992 Chr.17: 82409459-82409459 on Build GRCh38
rs928665409 Chr.17: 82409452-82409452 on Build GRCh38
rs759472839 Chr.17: 82409423-82409425 on Build GRCh38
rs111690356 Chr.17: 82409399-82409399 on Build GRCh38
rs972038135 Chr.17: 82409411-82409411 on Build GRCh38
rs148998477 Chr.17: 82409409-82409409 on Build GRCh38
rs200591786 Chr.17: 82409439-82409439 on Build GRCh38

View Full Product Details