Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82446539-82447039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf62
Entrez Gene ID
79415
Gene Name
chromosome 17 open reading frame 62
Gene Aliases
-
UniGene
Hs.163113
Gene Chromosome Location
Chr.17: 82442586-82450831 on Build GRCh38
Transcripts
NM_001033046.3 NM_001100407.2 NM_001100408.2 NM_001193653.1 NM_001193654.1 NM_001193655.1 NM_001193657.1 NR_036514.1 NR_036516.1 NR_036517.1 NR_036518.1 XM_006722293.2 XM_011523606.2 XM_017025072.1 XM_017025073.1 XM_017025074.1 XM_017025075.1 XM_017025076.1 XM_017025077.1 XM_017025078.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
291525 c.193G>C p.D65H Chr.17: 82446631-82446631 on Build GRCh38
5983749 c.183G>T p.Q61H Chr.17: 82446641-82446641 on Build GRCh38
7352470 c.145C>T p.L49F Chr.17: 82446679-82446679 on Build GRCh38
1387508 c.160G>T p.G54C Chr.17: 82446664-82446664 on Build GRCh38
6614674 c.153C>T p.Y51Y Chr.17: 82446671-82446671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs72857481 Chr.17: 82446679-82446679 on Build GRCh38
rs762955082 Chr.17: 82446724-82446724 on Build GRCh38
rs774623467 Chr.17: 82446742-82446742 on Build GRCh38
rs952721200 Chr.17: 82446780-82446780 on Build GRCh38
rs146453694 Chr.17: 82446900-82446900 on Build GRCh38
rs1027382805 Chr.17: 82446772-82446772 on Build GRCh38
rs769836121 Chr.17: 82446643-82446643 on Build GRCh38
rs563246870 Chr.17: 82446864-82446864 on Build GRCh38
rs905303531 Chr.17: 82446965-82446965 on Build GRCh38
rs575337426 Chr.17: 82446870-82446870 on Build GRCh38
rs923168129 Chr.17: 82446873-82446873 on Build GRCh38
rs767970541 Chr.17: 82446686-82446686 on Build GRCh38
rs972878530 Chr.17: 82446593-82446593 on Build GRCh38
rs1025106125 Chr.17: 82446757-82446757 on Build GRCh38
rs537141947 Chr.17: 82446716-82446716 on Build GRCh38
rs546476068 Chr.17: 82446575-82446575 on Build GRCh38
rs769671501 Chr.17: 82446721-82446721 on Build GRCh38
rs764345206 Chr.17: 82446587-82446587 on Build GRCh38
rs528237726 Chr.17: 82446938-82446938 on Build GRCh38
rs552550919 Chr.17: 82446779-82446779 on Build GRCh38
rs984632189 Chr.17: 82446862-82446862 on Build GRCh38
rs113105756 Chr.17: 82446846-82446846 on Build GRCh38
rs776577551 Chr.17: 82446573-82446573 on Build GRCh38
rs1053122702 Chr.17: 82446918-82446918 on Build GRCh38
rs761292944 Chr.17: 82446747-82446747 on Build GRCh38
rs982998840 Chr.17: 82446968-82446968 on Build GRCh38
rs544938746 Chr.17: 82446863-82446863 on Build GRCh38
rs969382150 Chr.17: 82446603-82446603 on Build GRCh38
rs1025032580 Chr.17: 82446594-82446594 on Build GRCh38
rs201461354 Chr.17: 82446610-82446610 on Build GRCh38
rs758804089 Chr.17: 82446582-82446582 on Build GRCh38
rs112450525 Chr.17: 82446604-82446604 on Build GRCh38
rs1025251085 Chr.17: 82446958-82446958 on Build GRCh38
rs561164088 Chr.17: 82446921-82446921 on Build GRCh38
rs140822873 Chr.17: 82446670-82446670 on Build GRCh38
rs763184892 Chr.17: 82446659-82446659 on Build GRCh38
rs1042416994 Chr.17: 82446901-82446901 on Build GRCh38
rs1042162725 Chr.17: 82446729-82446729 on Build GRCh38
rs952776377 Chr.17: 82446998-82446998 on Build GRCh38
rs1012494191 Chr.17: 82446774-82446774 on Build GRCh38
rs770787192 Chr.17: 82446632-82446632 on Build GRCh38
rs750963264 Chr.17: 82446688-82446688 on Build GRCh38
rs775578053 Chr.17: 82446723-82446723 on Build GRCh38
rs777425774 Chr.17: 82446714-82446714 on Build GRCh38
rs985126290 Chr.17: 82446793-82446793 on Build GRCh38
rs1027042953 Chr.17: 82446974-82446974 on Build GRCh38
rs539421554 Chr.17: 82446608-82446608 on Build GRCh38
rs375257998 Chr.17: 82446614-82446614 on Build GRCh38
rs148042402 Chr.17: 82446640-82446640 on Build GRCh38
rs775739450 Chr.17: 82446656-82446656 on Build GRCh38
rs750726343 Chr.17: 82446721-82446721 on Build GRCh38
rs750765186 Chr.17: 82446746-82446746 on Build GRCh38
rs746836449 Chr.17: 82446715-82446715 on Build GRCh38
rs1046590012 Chr.17: 82446944-82446944 on Build GRCh38
rs746747466 Chr.17: 82446618-82446618 on Build GRCh38
rs372955130 Chr.17: 82446743-82446743 on Build GRCh38
rs961854518 Chr.17: 82446844-82446844 on Build GRCh38
rs199597100 Chr.17: 82446615-82446615 on Build GRCh38
rs760501935 Chr.17: 82446595-82446596 on Build GRCh38
rs908694759 Chr.17: 82446972-82446972 on Build GRCh38
rs754581937 Chr.17: 82446690-82446690 on Build GRCh38
rs757987645 Chr.17: 82446711-82446711 on Build GRCh38
rs944142739 Chr.17: 82446977-82446977 on Build GRCh38
rs758364341 Chr.17: 82446572-82446572 on Build GRCh38
rs370004931 Chr.17: 82446577-82446577 on Build GRCh38
rs973123266 Chr.17: 82446794-82446794 on Build GRCh38
rs535372187 Chr.17: 82446770-82446770 on Build GRCh38
rs907751408 Chr.17: 82446813-82446813 on Build GRCh38
rs922984401 Chr.17: 82446683-82446683 on Build GRCh38
rs372816681 Chr.17: 82446738-82446738 on Build GRCh38
rs555976449 Chr.17: 82446756-82446756 on Build GRCh38
rs953056808 Chr.17: 82446606-82446606 on Build GRCh38
rs931687635 Chr.17: 82446695-82446695 on Build GRCh38
rs1025743447 Chr.17: 82446795-82446795 on Build GRCh38
rs778584919 Chr.17: 82446707-82446707 on Build GRCh38
rs923056380 Chr.17: 82446856-82446856 on Build GRCh38
rs150179197 Chr.17: 82446671-82446671 on Build GRCh38
rs969455411 Chr.17: 82446762-82446762 on Build GRCh38
rs1002718054 Chr.17: 82446967-82446967 on Build GRCh38
rs902267787 Chr.17: 82446907-82446907 on Build GRCh38
rs752210927 Chr.17: 82446710-82446710 on Build GRCh38
rs763298051 Chr.17: 82446865-82446865 on Build GRCh38
rs574264440 Chr.17: 82446768-82446768 on Build GRCh38
rs1026033435 Chr.17: 82446982-82446982 on Build GRCh38
rs924957422 Chr.17: 82446887-82446887 on Build GRCh38
rs373722472 Chr.17: 82446589-82446589 on Build GRCh38
rs370383989 Chr.17: 82446609-82446609 on Build GRCh38

View Full Product Details