Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82449296-82449792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf62
Entrez Gene ID
79415
Gene Name
chromosome 17 open reading frame 62
Gene Aliases
-
UniGene
Hs.163113
Gene Chromosome Location
Chr.17: 82442586-82450831 on Build GRCh38
Transcripts
NM_001033046.3 NM_001100407.2 NM_001100408.2 NM_001193653.1 NM_001193654.1 NM_001193655.1 NM_001193657.1 NR_036514.1 NR_036516.1 NR_036517.1 NR_036518.1 NR_036519.1 XM_006722293.2 XM_011523606.2 XM_017025072.1 XM_017025073.1 XM_017025074.1 XM_017025075.1 XM_017025076.1 XM_017025077.1 XM_017025078.1

SNP Detail

SNP ID Chromosome Location
rs530198980 Chr.17: 82449383-82449383 on Build GRCh38
rs746418895 Chr.17: 82449544-82449544 on Build GRCh38
rs73999904 Chr.17: 82449541-82449541 on Build GRCh38
rs112690672 Chr.17: 82449548-82449548 on Build GRCh38
rs11870582 Chr.17: 82449749-82449749 on Build GRCh38
rs918365005 Chr.17: 82449511-82449511 on Build GRCh38
rs373856077 Chr.17: 82449354-82449354 on Build GRCh38
rs190628394 Chr.17: 82449726-82449726 on Build GRCh38
rs73999905 Chr.17: 82449542-82449542 on Build GRCh38
rs903289788 Chr.17: 82449576-82449576 on Build GRCh38
rs958705968 Chr.17: 82449414-82449414 on Build GRCh38
rs111962223 Chr.17: 82449343-82449343 on Build GRCh38
rs962904444 Chr.17: 82449472-82449472 on Build GRCh38
rs894471661 Chr.17: 82449618-82449618 on Build GRCh38
rs60529510 Chr.17: 82449604-82449604 on Build GRCh38
rs772807951 Chr.17: 82449697-82449697 on Build GRCh38
rs919854626 Chr.17: 82449613-82449613 on Build GRCh38
rs929710494 Chr.17: 82449519-82449519 on Build GRCh38
rs562977049 Chr.17: 82449360-82449360 on Build GRCh38
rs951188156 Chr.17: 82449719-82449719 on Build GRCh38
rs554119438 Chr.17: 82449669-82449669 on Build GRCh38
rs986520097 Chr.17: 82449730-82449730 on Build GRCh38
rs919825049 Chr.17: 82449381-82449381 on Build GRCh38
rs527298762 Chr.17: 82449504-82449504 on Build GRCh38
rs960064178 Chr.17: 82449554-82449554 on Build GRCh38
rs991153636 Chr.17: 82449418-82449418 on Build GRCh38
rs143628628 Chr.17: 82449695-82449695 on Build GRCh38
rs771062764 Chr.17: 82449449-82449449 on Build GRCh38
rs985146596 Chr.17: 82449655-82449655 on Build GRCh38
rs376798123 Chr.17: 82449441-82449441 on Build GRCh38
rs775931975 Chr.17: 82449565-82449565 on Build GRCh38
rs551923893 Chr.17: 82449527-82449527 on Build GRCh38
rs1024007683 Chr.17: 82449687-82449687 on Build GRCh38
rs1002972480 Chr.17: 82449585-82449585 on Build GRCh38
rs1025451474 Chr.17: 82449717-82449717 on Build GRCh38
rs544397900 Chr.17: 82449731-82449731 on Build GRCh38
rs568275910 Chr.17: 82449579-82449579 on Build GRCh38
rs769055360 Chr.17: 82449627-82449627 on Build GRCh38
rs548495660 Chr.17: 82449384-82449384 on Build GRCh38
rs1043170395 Chr.17: 82449757-82449760 on Build GRCh38
rs960363648 Chr.17: 82449326-82449326 on Build GRCh38
rs560365681 Chr.17: 82449431-82449431 on Build GRCh38
rs558252244 Chr.17: 82449484-82449484 on Build GRCh38
rs890887216 Chr.17: 82449477-82449477 on Build GRCh38
rs1052824187 Chr.17: 82449701-82449701 on Build GRCh38
rs182861963 Chr.17: 82449748-82449748 on Build GRCh38
rs558709185 Chr.17: 82449707-82449707 on Build GRCh38
rs947567719 Chr.17: 82449562-82449562 on Build GRCh38
rs954063883 Chr.17: 82449636-82449636 on Build GRCh38
rs908412449 Chr.17: 82449349-82449349 on Build GRCh38
rs762636267 Chr.17: 82449734-82449734 on Build GRCh38
rs138352835 Chr.17: 82449553-82449553 on Build GRCh38
rs1005006004 Chr.17: 82449572-82449572 on Build GRCh38
rs572411055 Chr.17: 82449686-82449686 on Build GRCh38
rs974017882 Chr.17: 82449375-82449375 on Build GRCh38
rs1043306868 Chr.17: 82449526-82449526 on Build GRCh38
rs893383278 Chr.17: 82449561-82449561 on Build GRCh38
rs540219618 Chr.17: 82449654-82449654 on Build GRCh38
rs912273778 Chr.17: 82449735-82449735 on Build GRCh38
rs986490593 Chr.17: 82449529-82449529 on Build GRCh38
rs912960268 Chr.17: 82449718-82449718 on Build GRCh38
rs1015005390 Chr.17: 82449581-82449581 on Build GRCh38
rs961226365 Chr.17: 82449595-82449595 on Build GRCh38

View Full Product Details