Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82563262-82563741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXK2
Entrez Gene ID
3607
Gene Name
forkhead box K2
Gene Aliases
ILF, ILF-1, ILF1
UniGene
Hs.591140
Gene Chromosome Location
Chr.17: 82519718-82604607 on Build GRCh38
Transcripts
NM_004514.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1471140 c.523G>A p.V175I Chr.17: 82563457-82563457 on Build GRCh38
3523881 c.425C>T p.T142I Chr.17: 82563359-82563359 on Build GRCh38
4614474 c.481_482insAG p.K164fs*10 Chr.17: 82563415-82563416 on Build GRCh38
4070925 c.522C>T p.A174A Chr.17: 82563456-82563456 on Build GRCh38
4685401 c.571G>A p.A191T Chr.17: 82563505-82563505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746207142 Chr.17: 82563347-82563347 on Build GRCh38
rs780826737 Chr.17: 82563565-82563565 on Build GRCh38
rs780583952 Chr.17: 82563537-82563537 on Build GRCh38
rs756557187 Chr.17: 82563441-82563441 on Build GRCh38
rs370657954 Chr.17: 82563594-82563594 on Build GRCh38
rs776280220 Chr.17: 82563480-82563480 on Build GRCh38
rs781725364 Chr.17: 82563341-82563341 on Build GRCh38
rs965922950 Chr.17: 82563627-82563627 on Build GRCh38
rs762806609 Chr.17: 82563587-82563587 on Build GRCh38
rs752637649 Chr.17: 82563411-82563411 on Build GRCh38
rs986449613 Chr.17: 82563671-82563671 on Build GRCh38
rs772733435 Chr.17: 82563372-82563372 on Build GRCh38
rs374308998 Chr.17: 82563567-82563567 on Build GRCh38
rs747226726 Chr.17: 82563334-82563334 on Build GRCh38
rs200451137 Chr.17: 82563473-82563473 on Build GRCh38
rs758421844 Chr.17: 82563325-82563325 on Build GRCh38
rs764910135 Chr.17: 82563455-82563455 on Build GRCh38
rs1054914322 Chr.17: 82563489-82563489 on Build GRCh38
rs1023853191 Chr.17: 82563302-82563302 on Build GRCh38
rs367705050 Chr.17: 82563474-82563474 on Build GRCh38
rs576770102 Chr.17: 82563641-82563641 on Build GRCh38
rs866802169 Chr.17: 82563392-82563392 on Build GRCh38
rs760266857 Chr.17: 82563382-82563382 on Build GRCh38
rs749751515 Chr.17: 82563369-82563369 on Build GRCh38
rs150523977 Chr.17: 82563434-82563434 on Build GRCh38
rs913269077 Chr.17: 82563390-82563390 on Build GRCh38
rs535971348 Chr.17: 82563528-82563528 on Build GRCh38
rs758315545 Chr.17: 82563412-82563412 on Build GRCh38
rs1019835437 Chr.17: 82563608-82563608 on Build GRCh38
rs759320912 Chr.17: 82563405-82563405 on Build GRCh38
rs1025921918 Chr.17: 82563615-82563615 on Build GRCh38
rs746295222 Chr.17: 82563439-82563439 on Build GRCh38
rs201972160 Chr.17: 82563534-82563534 on Build GRCh38
rs149606107 Chr.17: 82563576-82563576 on Build GRCh38
rs775192234 Chr.17: 82563578-82563578 on Build GRCh38
rs377626967 Chr.17: 82563350-82563350 on Build GRCh38
rs757456255 Chr.17: 82563335-82563335 on Build GRCh38
rs766033253 Chr.17: 82563384-82563384 on Build GRCh38
rs369818272 Chr.17: 82563308-82563308 on Build GRCh38
rs944328742 Chr.17: 82563326-82563326 on Build GRCh38
rs953735021 Chr.17: 82563281-82563281 on Build GRCh38
rs1041675524 Chr.17: 82563543-82563543 on Build GRCh38
rs148118470 Chr.17: 82563435-82563435 on Build GRCh38
rs1039516598 Chr.17: 82563448-82563448 on Build GRCh38
rs139363774 Chr.17: 82563572-82563572 on Build GRCh38
rs985642240 Chr.17: 82563306-82563306 on Build GRCh38
rs751659158 Chr.17: 82563523-82563523 on Build GRCh38
rs912276210 Chr.17: 82563692-82563692 on Build GRCh38
rs754575677 Chr.17: 82563549-82563550 on Build GRCh38
rs762851811 Chr.17: 82563507-82563507 on Build GRCh38
rs996140799 Chr.17: 82563602-82563602 on Build GRCh38
rs776484939 Chr.17: 82563331-82563332 on Build GRCh38
rs765125671 Chr.17: 82563315-82563315 on Build GRCh38
rs761835619 Chr.17: 82563547-82563548 on Build GRCh38
rs771484161 Chr.17: 82563371-82563371 on Build GRCh38
rs200132853 Chr.17: 82563329-82563329 on Build GRCh38
rs764065641 Chr.17: 82563514-82563514 on Build GRCh38
rs375197792 Chr.17: 82563591-82563591 on Build GRCh38
rs572639784 Chr.17: 82563553-82563553 on Build GRCh38
rs754207160 Chr.17: 82563550-82563550 on Build GRCh38
rs370351115 Chr.17: 82563562-82563562 on Build GRCh38
rs754956035 Chr.17: 82563309-82563309 on Build GRCh38
rs746556436 Chr.17: 82563557-82563557 on Build GRCh38
rs371316297 Chr.17: 82563456-82563456 on Build GRCh38
rs752833927 Chr.17: 82563321-82563321 on Build GRCh38
rs988962933 Chr.17: 82563677-82563677 on Build GRCh38
rs913479776 Chr.17: 82563701-82563701 on Build GRCh38
rs776448719 Chr.17: 82563393-82563393 on Build GRCh38
rs780496048 Chr.17: 82563443-82563443 on Build GRCh38
rs762780332 Chr.17: 82563548-82563549 on Build GRCh38
rs141879160 Chr.17: 82563502-82563502 on Build GRCh38
rs1012438335 Chr.17: 82563287-82563287 on Build GRCh38
rs776196809 Chr.17: 82563304-82563304 on Build GRCh38
rs963775628 Chr.17: 82563702-82563702 on Build GRCh38
rs752098927 Chr.17: 82563571-82563572 on Build GRCh38
rs755477837 Chr.17: 82563552-82563552 on Build GRCh38
rs543459125 Chr.17: 82563713-82563713 on Build GRCh38
rs769608161 Chr.17: 82563488-82563488 on Build GRCh38
rs769153791 Chr.17: 82563469-82563469 on Build GRCh38

View Full Product Details