Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82720183-82720678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FN3KRP
Entrez Gene ID
79672
Gene Name
fructosamine 3 kinase related protein
Gene Aliases
FN3KL
UniGene
Hs.31431
Gene Chromosome Location
Chr.17: 82716706-82728017 on Build GRCh38
Transcripts
NM_024619.3 NR_046408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6665820 c.377G>A p.G126D Chr.17: 82720355-82720355 on Build GRCh38
1387529 c.362G>A p.R121H Chr.17: 82720340-82720340 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs553145951 Chr.17: 82720436-82720436 on Build GRCh38
rs980572598 Chr.17: 82720614-82720614 on Build GRCh38
rs201627539 Chr.17: 82720359-82720359 on Build GRCh38
rs982690355 Chr.17: 82720446-82720446 on Build GRCh38
rs776780095 Chr.17: 82720248-82720248 on Build GRCh38
rs746359416 Chr.17: 82720220-82720220 on Build GRCh38
rs996750941 Chr.17: 82720328-82720328 on Build GRCh38
rs752625606 Chr.17: 82720316-82720317 on Build GRCh38
rs759352409 Chr.17: 82720367-82720367 on Build GRCh38
rs995516089 Chr.17: 82720653-82720653 on Build GRCh38
rs776584529 Chr.17: 82720360-82720360 on Build GRCh38
rs917676565 Chr.17: 82720409-82720409 on Build GRCh38
rs896602476 Chr.17: 82720417-82720417 on Build GRCh38
rs760440439 Chr.17: 82720245-82720245 on Build GRCh38
rs756439956 Chr.17: 82720392-82720392 on Build GRCh38
rs747651961 Chr.17: 82720351-82720351 on Build GRCh38
rs771589335 Chr.17: 82720352-82720352 on Build GRCh38
rs896555678 Chr.17: 82720588-82720588 on Build GRCh38
rs985838659 Chr.17: 82720278-82720278 on Build GRCh38
rs112572268 Chr.17: 82720585-82720585 on Build GRCh38
rs780949550 Chr.17: 82720407-82720407 on Build GRCh38
rs770841368 Chr.17: 82720247-82720247 on Build GRCh38
rs1037138295 Chr.17: 82720602-82720602 on Build GRCh38
rs781135315 Chr.17: 82720506-82720506 on Build GRCh38
rs1013268340 Chr.17: 82720543-82720543 on Build GRCh38
rs752740663 Chr.17: 82720258-82720258 on Build GRCh38
rs781530963 Chr.17: 82720310-82720310 on Build GRCh38
rs750718633 Chr.17: 82720339-82720339 on Build GRCh38
rs750049231 Chr.17: 82720377-82720377 on Build GRCh38
rs73361132 Chr.17: 82720494-82720494 on Build GRCh38
rs573326297 Chr.17: 82720633-82720633 on Build GRCh38
rs1050936685 Chr.17: 82720518-82720518 on Build GRCh38
rs375112043 Chr.17: 82720394-82720394 on Build GRCh38
rs764259433 Chr.17: 82720275-82720275 on Build GRCh38
rs781700313 Chr.17: 82720219-82720219 on Build GRCh38
rs757583889 Chr.17: 82720209-82720209 on Build GRCh38
rs951825970 Chr.17: 82720465-82720465 on Build GRCh38
rs746462807 Chr.17: 82720403-82720403 on Build GRCh38
rs771654065 Chr.17: 82720235-82720235 on Build GRCh38
rs897109885 Chr.17: 82720628-82720628 on Build GRCh38
rs574096968 Chr.17: 82720376-82720376 on Build GRCh38
rs778301562 Chr.17: 82720225-82720225 on Build GRCh38
rs756638100 Chr.17: 82720340-82720340 on Build GRCh38
rs768921898 Chr.17: 82720412-82720412 on Build GRCh38
rs754945903 Chr.17: 82720432-82720432 on Build GRCh38
rs1024682153 Chr.17: 82720567-82720567 on Build GRCh38
rs777343562 Chr.17: 82720402-82720402 on Build GRCh38
rs768220660 Chr.17: 82720221-82720221 on Build GRCh38
rs368761106 Chr.17: 82720395-82720395 on Build GRCh38
rs910301578 Chr.17: 82720491-82720491 on Build GRCh38
rs763045513 Chr.17: 82720265-82720265 on Build GRCh38
rs951878526 Chr.17: 82720273-82720273 on Build GRCh38
rs766845832 Chr.17: 82720393-82720393 on Build GRCh38
rs557244633 Chr.17: 82720301-82720301 on Build GRCh38
rs975762727 Chr.17: 82720495-82720495 on Build GRCh38
rs947900125 Chr.17: 82720229-82720229 on Build GRCh38
rs373904593 Chr.17: 82720353-82720353 on Build GRCh38
rs539671230 Chr.17: 82720416-82720416 on Build GRCh38
rs528215600 Chr.17: 82720595-82720595 on Build GRCh38
rs1027576567 Chr.17: 82720462-82720462 on Build GRCh38
rs901910450 Chr.17: 82720280-82720280 on Build GRCh38
rs142819200 Chr.17: 82720384-82720384 on Build GRCh38
rs767498648 Chr.17: 82720273-82720274 on Build GRCh38
rs779492087 Chr.17: 82720408-82720408 on Build GRCh38
rs750819175 Chr.17: 82720390-82720390 on Build GRCh38
rs774501404 Chr.17: 82720379-82720379 on Build GRCh38
rs545758821 Chr.17: 82720461-82720461 on Build GRCh38
rs370920900 Chr.17: 82720385-82720385 on Build GRCh38
rs764220024 Chr.17: 82720378-82720378 on Build GRCh38
rs952586812 Chr.17: 82720639-82720639 on Build GRCh38
rs370498197 Chr.17: 82720320-82720320 on Build GRCh38
rs1013569148 Chr.17: 82720422-82720422 on Build GRCh38
rs1043921229 Chr.17: 82720537-82720537 on Build GRCh38
rs1044867542 Chr.17: 82720233-82720233 on Build GRCh38
rs376106465 Chr.17: 82720542-82720542 on Build GRCh38
rs932119691 Chr.17: 82720504-82720504 on Build GRCh38
rs377585773 Chr.17: 82720228-82720228 on Build GRCh38
rs1040313086 Chr.17: 82720466-82720466 on Build GRCh38
rs751783183 Chr.17: 82720300-82720300 on Build GRCh38
rs369778435 Chr.17: 82720451-82720451 on Build GRCh38
rs542621698 Chr.17: 82720529-82720529 on Build GRCh38
rs201866362 Chr.17: 82720412-82720412 on Build GRCh38
rs1001006191 Chr.17: 82720539-82720539 on Build GRCh38
rs980923326 Chr.17: 82720419-82720419 on Build GRCh38
rs769647464 Chr.17: 82720375-82720375 on Build GRCh38
rs748025532 Chr.17: 82720515-82720515 on Build GRCh38
rs961413277 Chr.17: 82720439-82720439 on Build GRCh38
rs577104797 Chr.17: 82720344-82720344 on Build GRCh38
rs765206483 Chr.17: 82720257-82720257 on Build GRCh38
rs908437193 Chr.17: 82720450-82720450 on Build GRCh38
rs757676099 Chr.17: 82720302-82720302 on Build GRCh38
rs531067271 Chr.17: 82720522-82720522 on Build GRCh38
rs759554457 Chr.17: 82720253-82720253 on Build GRCh38
rs931118954 Chr.17: 82720433-82720433 on Build GRCh38
rs115523885 Chr.17: 82720541-82720541 on Build GRCh38

View Full Product Details