Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 2847711-2848209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMILIN2
Entrez Gene ID
84034
Gene Name
elastin microfibril interfacer 2
Gene Aliases
EMILIN-2, FOAP-10
UniGene
Hs.532815
Gene Chromosome Location
Chr.18: 2847030-2914092 on Build GRCh38
Transcripts
NM_032048.2 XM_017026038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6148950 c.205C>A p.Q69K Chr.18: 2847879-2847879 on Build GRCh38
3403493 c.240C>T p.P80P Chr.18: 2847914-2847914 on Build GRCh38
3524764 c.188G>A p.G63E Chr.18: 2847862-2847862 on Build GRCh38
4596465 c.215A>G p.Y72C Chr.18: 2847889-2847889 on Build GRCh38
6422185 c.245C>T p.P82L Chr.18: 2847919-2847919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949789081 Chr.18: 2848096-2848096 on Build GRCh38
rs2289686 Chr.18: 2847746-2847746 on Build GRCh38
rs1022221170 Chr.18: 2848043-2848043 on Build GRCh38
rs200114953 Chr.18: 2847892-2847892 on Build GRCh38
rs755062782 Chr.18: 2847883-2847883 on Build GRCh38
rs760413843 Chr.18: 2847941-2847941 on Build GRCh38
rs767570344 Chr.18: 2847927-2847927 on Build GRCh38
rs147479179 Chr.18: 2847803-2847803 on Build GRCh38
rs940355717 Chr.18: 2847989-2847989 on Build GRCh38
rs1009012629 Chr.18: 2848019-2848019 on Build GRCh38
rs747377068 Chr.18: 2847898-2847898 on Build GRCh38
rs1057269181 Chr.18: 2847976-2847976 on Build GRCh38
rs756296021 Chr.18: 2847844-2847844 on Build GRCh38
rs772640105 Chr.18: 2847803-2847803 on Build GRCh38
rs750207112 Chr.18: 2847930-2847930 on Build GRCh38
rs775028474 Chr.18: 2847911-2847911 on Build GRCh38
rs368312797 Chr.18: 2848090-2848090 on Build GRCh38
rs779024497 Chr.18: 2847962-2847962 on Build GRCh38
rs952824334 Chr.18: 2847993-2847993 on Build GRCh38
rs565267833 Chr.18: 2848156-2848156 on Build GRCh38
rs769170051 Chr.18: 2847760-2847760 on Build GRCh38
rs760887267 Chr.18: 2848119-2848119 on Build GRCh38
rs922753437 Chr.18: 2848023-2848023 on Build GRCh38
rs769447842 Chr.18: 2847899-2847899 on Build GRCh38
rs956694315 Chr.18: 2847830-2847830 on Build GRCh38
rs947280293 Chr.18: 2847915-2847915 on Build GRCh38
rs1027547546 Chr.18: 2847988-2847988 on Build GRCh38
rs573227557 Chr.18: 2848125-2848125 on Build GRCh38
rs766170092 Chr.18: 2848064-2848064 on Build GRCh38
rs762499245 Chr.18: 2847916-2847916 on Build GRCh38
rs575376753 Chr.18: 2847889-2847889 on Build GRCh38
rs750260260 Chr.18: 2847842-2847842 on Build GRCh38
rs968644015 Chr.18: 2848044-2848044 on Build GRCh38
rs897680079 Chr.18: 2848124-2848124 on Build GRCh38
rs887688479 Chr.18: 2848181-2848181 on Build GRCh38
rs748491137 Chr.18: 2847773-2847773 on Build GRCh38
rs762544547 Chr.18: 2847956-2847957 on Build GRCh38
rs904200547 Chr.18: 2847791-2847791 on Build GRCh38
rs74795212 Chr.18: 2848127-2848127 on Build GRCh38
rs978096814 Chr.18: 2848041-2848041 on Build GRCh38
rs984103283 Chr.18: 2848003-2848003 on Build GRCh38
rs1027991982 Chr.18: 2848132-2848132 on Build GRCh38
rs996391507 Chr.18: 2848004-2848004 on Build GRCh38
rs778282756 Chr.18: 2847971-2847971 on Build GRCh38
rs764039184 Chr.18: 2847818-2847818 on Build GRCh38
rs112576328 Chr.18: 2848070-2848070 on Build GRCh38
rs767484647 Chr.18: 2847836-2847836 on Build GRCh38
rs897773358 Chr.18: 2847923-2847923 on Build GRCh38
rs111725137 Chr.18: 2847891-2847891 on Build GRCh38
rs766606235 Chr.18: 2847946-2847946 on Build GRCh38
rs758834591 Chr.18: 2847970-2847970 on Build GRCh38
rs935675044 Chr.18: 2848085-2848085 on Build GRCh38
rs938477587 Chr.18: 2847969-2847969 on Build GRCh38
rs140338789 Chr.18: 2847919-2847919 on Build GRCh38
rs988714257 Chr.18: 2847861-2847861 on Build GRCh38
rs2289687 Chr.18: 2847732-2847732 on Build GRCh38
rs762400510 Chr.18: 2847805-2847805 on Build GRCh38
rs559141789 Chr.18: 2847767-2847767 on Build GRCh38
rs549193140 Chr.18: 2848018-2848018 on Build GRCh38
rs191540793 Chr.18: 2847985-2847985 on Build GRCh38
rs769344079 Chr.18: 2847797-2847797 on Build GRCh38
rs542373285 Chr.18: 2848067-2848067 on Build GRCh38
rs761666897 Chr.18: 2847832-2847832 on Build GRCh38
rs751592359 Chr.18: 2847828-2847828 on Build GRCh38
rs772488342 Chr.18: 2847924-2847924 on Build GRCh38
rs577545567 Chr.18: 2847974-2847974 on Build GRCh38
rs780174463 Chr.18: 2847849-2847849 on Build GRCh38
rs754048001 Chr.18: 2847955-2847955 on Build GRCh38
rs753909713 Chr.18: 2847851-2847851 on Build GRCh38
rs759243205 Chr.18: 2847793-2847793 on Build GRCh38
rs946207722 Chr.18: 2848030-2848030 on Build GRCh38
rs557441744 Chr.18: 2847960-2847960 on Build GRCh38
rs776263519 Chr.18: 2847790-2847790 on Build GRCh38
rs772466546 Chr.18: 2847776-2847776 on Build GRCh38
rs907637731 Chr.18: 2848102-2848102 on Build GRCh38
rs752789104 Chr.18: 2847968-2847968 on Build GRCh38
rs1018404227 Chr.18: 2848008-2848008 on Build GRCh38
rs956766659 Chr.18: 2848038-2848038 on Build GRCh38
rs964121860 Chr.18: 2848011-2848011 on Build GRCh38
rs779722942 Chr.18: 2847981-2847981 on Build GRCh38
rs559136331 Chr.18: 2848091-2848091 on Build GRCh38
rs573601386 Chr.18: 2848031-2848031 on Build GRCh38

View Full Product Details