Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 3497468-3497969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP1
Entrez Gene ID
9229
Gene Name
DLG associated protein 1
Gene Aliases
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1
UniGene
Hs.594640
Gene Chromosome Location
Chr.18: 3496032-4455307 on Build GRCh38
Transcripts
NM_001003809.2 NM_001242762.1 NM_001242763.1 NM_001242764.1 NM_001242766.1 NM_001308390.1 NM_004746.3 XM_005258171.2 XM_005258172.1 XM_005258173.3 XM_005258174.4 XM_006722367.3 XM_017026080.1 XM_017026081.1 XM_017026083.1

SNP Detail

SNP ID Chromosome Location
rs1047716598 Chr.18: 3497925-3497925 on Build GRCh38
rs142172904 Chr.18: 3497517-3497517 on Build GRCh38
rs938521581 Chr.18: 3497911-3497911 on Build GRCh38
rs1020216951 Chr.18: 3497628-3497628 on Build GRCh38
rs938610490 Chr.18: 3497727-3497727 on Build GRCh38
rs1028870940 Chr.18: 3497757-3497757 on Build GRCh38
rs191784529 Chr.18: 3497672-3497672 on Build GRCh38
rs238114 Chr.18: 3497854-3497854 on Build GRCh38
rs756830021 Chr.18: 3497719-3497719 on Build GRCh38
rs567831138 Chr.18: 3497695-3497695 on Build GRCh38
rs533565356 Chr.18: 3497688-3497688 on Build GRCh38
rs536410213 Chr.18: 3497867-3497867 on Build GRCh38
rs771764710 Chr.18: 3497864-3497864 on Build GRCh38
rs964786633 Chr.18: 3497747-3497747 on Build GRCh38
rs924578135 Chr.18: 3497511-3497511 on Build GRCh38
rs367718762 Chr.18: 3497720-3497720 on Build GRCh38
rs183640358 Chr.18: 3497791-3497791 on Build GRCh38
rs1044241961 Chr.18: 3497901-3497901 on Build GRCh38
rs955655558 Chr.18: 3497768-3497768 on Build GRCh38
rs748576566 Chr.18: 3497682-3497683 on Build GRCh38
rs934212968 Chr.18: 3497494-3497494 on Build GRCh38
rs907138404 Chr.18: 3497683-3497683 on Build GRCh38
rs986765380 Chr.18: 3497499-3497499 on Build GRCh38
rs188919572 Chr.18: 3497926-3497926 on Build GRCh38
rs10513868 Chr.18: 3497716-3497716 on Build GRCh38
rs778272493 Chr.18: 3497625-3497625 on Build GRCh38
rs914356831 Chr.18: 3497681-3497681 on Build GRCh38
rs146363500 Chr.18: 3497859-3497859 on Build GRCh38
rs75425207 Chr.18: 3497531-3497531 on Build GRCh38
rs553328378 Chr.18: 3497582-3497582 on Build GRCh38
rs975625360 Chr.18: 3497756-3497756 on Build GRCh38
rs778616922 Chr.18: 3497730-3497730 on Build GRCh38
rs956160591 Chr.18: 3497629-3497629 on Build GRCh38
rs746824570 Chr.18: 3497908-3497908 on Build GRCh38
rs1028312314 Chr.18: 3497534-3497534 on Build GRCh38
rs1025994781 Chr.18: 3497699-3497699 on Build GRCh38
rs1008459408 Chr.18: 3497601-3497601 on Build GRCh38
rs924861613 Chr.18: 3497792-3497792 on Build GRCh38
rs907702782 Chr.18: 3497741-3497741 on Build GRCh38
rs544353073 Chr.18: 3497837-3497837 on Build GRCh38
rs893205436 Chr.18: 3497581-3497581 on Build GRCh38

View Full Product Details