Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 3508485-3508981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP1
Entrez Gene ID
9229
Gene Name
DLG associated protein 1
Gene Aliases
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1
UniGene
Hs.594640
Gene Chromosome Location
Chr.18: 3496032-4455307 on Build GRCh38
Transcripts
NM_001003809.2 NM_001242761.1 NM_001242762.1 NM_001242763.1 NM_001242764.1 NM_001242765.1 NM_001242766.1 NM_001308390.1 NM_004746.3 XM_005258171.2 XM_005258172.1 XM_005258173.3 XM_005258174.4 XM_006722367.3 XM_017026080.1 XM_017026081.1 XM_017026082.1 XM_017026083.1 XM_017026084.1 XM_017026085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3891001 c.2486G>A p.G829E Chr.18: 3508655-3508655 on Build GRCh38
3525727 c.1609T>G p.C537G Chr.18: 3508584-3508584 on Build GRCh38
4677800 c.1615A>G p.I539V Chr.18: 3508650-3508650 on Build GRCh38
4827793 c.2532G>A p.Q844Q Chr.18: 3508609-3508609 on Build GRCh38
4867721 c.1625G>A p.G542D Chr.18: 3508634-3508634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765939102 Chr.18: 3508559-3508559 on Build GRCh38
rs112282558 Chr.18: 3508554-3508554 on Build GRCh38
rs187873892 Chr.18: 3508681-3508681 on Build GRCh38
rs114414304 Chr.18: 3508752-3508752 on Build GRCh38
rs150239405 Chr.18: 3508907-3508907 on Build GRCh38
rs538990113 Chr.18: 3508643-3508643 on Build GRCh38
rs553474012 Chr.18: 3508886-3508886 on Build GRCh38
rs769098308 Chr.18: 3508650-3508650 on Build GRCh38
rs775948897 Chr.18: 3508584-3508584 on Build GRCh38
rs770425612 Chr.18: 3508656-3508656 on Build GRCh38
rs200874819 Chr.18: 3508691-3508691 on Build GRCh38
rs1023500240 Chr.18: 3508829-3508829 on Build GRCh38
rs759554695 Chr.18: 3508604-3508604 on Build GRCh38
rs548021952 Chr.18: 3508764-3508764 on Build GRCh38
rs749712073 Chr.18: 3508534-3508534 on Build GRCh38
rs971007426 Chr.18: 3508850-3508850 on Build GRCh38
rs533864945 Chr.18: 3508883-3508883 on Build GRCh38
rs1051987274 Chr.18: 3508680-3508680 on Build GRCh38
rs1000713370 Chr.18: 3508509-3508509 on Build GRCh38
rs767422867 Chr.18: 3508911-3508911 on Build GRCh38
rs902054058 Chr.18: 3508881-3508881 on Build GRCh38
rs763643448 Chr.18: 3508550-3508552 on Build GRCh38
rs981950813 Chr.18: 3508882-3508882 on Build GRCh38
rs371204865 Chr.18: 3508671-3508671 on Build GRCh38
rs763961558 Chr.18: 3508703-3508703 on Build GRCh38
rs749763519 Chr.18: 3508647-3508647 on Build GRCh38
rs765335648 Chr.18: 3508606-3508606 on Build GRCh38
rs759179224 Chr.18: 3508689-3508689 on Build GRCh38
rs757361946 Chr.18: 3508641-3508641 on Build GRCh38
rs528036510 Chr.18: 3508653-3508653 on Build GRCh38
rs556451438 Chr.18: 3508535-3508535 on Build GRCh38
rs541412321 Chr.18: 3508655-3508655 on Build GRCh38
rs772959321 Chr.18: 3508541-3508541 on Build GRCh38
rs542499759 Chr.18: 3508624-3508624 on Build GRCh38
rs943526867 Chr.18: 3508820-3508820 on Build GRCh38
rs751826163 Chr.18: 3508633-3508633 on Build GRCh38
rs775970421 Chr.18: 3508533-3508533 on Build GRCh38
rs1042382410 Chr.18: 3508949-3508949 on Build GRCh38
rs568914787 Chr.18: 3508713-3508713 on Build GRCh38
rs369683779 Chr.18: 3508542-3508542 on Build GRCh38
rs909426293 Chr.18: 3508812-3508812 on Build GRCh38
rs781334454 Chr.18: 3508642-3508642 on Build GRCh38
rs149401099 Chr.18: 3508615-3508615 on Build GRCh38
rs138428792 Chr.18: 3508707-3508707 on Build GRCh38
rs756172283 Chr.18: 3508644-3508644 on Build GRCh38
rs893729029 Chr.18: 3508757-3508757 on Build GRCh38
rs770216761 Chr.18: 3508519-3508519 on Build GRCh38
rs972769165 Chr.18: 3508744-3508744 on Build GRCh38
rs752656401 Chr.18: 3508609-3508609 on Build GRCh38
rs918351711 Chr.18: 3508746-3508746 on Build GRCh38
rs562356088 Chr.18: 3508646-3508646 on Build GRCh38
rs983469242 Chr.18: 3508913-3508913 on Build GRCh38
rs570324784 Chr.18: 3508560-3508560 on Build GRCh38
rs1039297571 Chr.18: 3508824-3508824 on Build GRCh38
rs376173328 Chr.18: 3508693-3508693 on Build GRCh38
rs142991062 Chr.18: 3508822-3508822 on Build GRCh38

View Full Product Details