Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5143985-5144492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAIN1
Entrez Gene ID
642597
Gene Name
A kinase (PRKA) anchor inhibitor 1
Gene Aliases
C18orf42
UniGene
Hs.437160
Gene Chromosome Location
Chr.18: 5143673-5199997 on Build GRCh38
Transcripts
NM_001145194.1 XM_011525728.2

SNP Detail

SNP ID Chromosome Location
rs947577351 Chr.18: 5144284-5144285 on Build GRCh38
rs748650893 Chr.18: 5144359-5144359 on Build GRCh38
rs888359801 Chr.18: 5144213-5144213 on Build GRCh38
rs937379050 Chr.18: 5144033-5144033 on Build GRCh38
rs372993561 Chr.18: 5144314-5144314 on Build GRCh38
rs1017944138 Chr.18: 5144218-5144218 on Build GRCh38
rs926226305 Chr.18: 5144364-5144364 on Build GRCh38
rs903610496 Chr.18: 5144291-5144291 on Build GRCh38
rs984598544 Chr.18: 5144108-5144108 on Build GRCh38
rs1020033699 Chr.18: 5144275-5144275 on Build GRCh38
rs549132269 Chr.18: 5144149-5144149 on Build GRCh38
rs549606174 Chr.18: 5144285-5144285 on Build GRCh38
rs965396253 Chr.18: 5144255-5144255 on Build GRCh38
rs958993625 Chr.18: 5144367-5144367 on Build GRCh38
rs919322956 Chr.18: 5144451-5144451 on Build GRCh38
rs1857847 Chr.18: 5144271-5144271 on Build GRCh38
rs972476465 Chr.18: 5144210-5144210 on Build GRCh38
rs778090715 Chr.18: 5144371-5144371 on Build GRCh38
rs993946468 Chr.18: 5144180-5144180 on Build GRCh38
rs938967726 Chr.18: 5144261-5144261 on Build GRCh38
rs430364 Chr.18: 5144322-5144322 on Build GRCh38
rs567250629 Chr.18: 5144215-5144215 on Build GRCh38
rs897004229 Chr.18: 5144132-5144132 on Build GRCh38
rs755524755 Chr.18: 5144305-5144305 on Build GRCh38
rs766518672 Chr.18: 5144198-5144198 on Build GRCh38
rs950166620 Chr.18: 5144099-5144099 on Build GRCh38
rs997455076 Chr.18: 5144332-5144332 on Build GRCh38
rs537012746 Chr.18: 5144416-5144416 on Build GRCh38
rs756531930 Chr.18: 5144368-5144368 on Build GRCh38
rs1043180625 Chr.18: 5144288-5144288 on Build GRCh38
rs146085870 Chr.18: 5144110-5144110 on Build GRCh38
rs1052562532 Chr.18: 5144304-5144304 on Build GRCh38
rs1036432091 Chr.18: 5144437-5144437 on Build GRCh38
rs962476545 Chr.18: 5144428-5144428 on Build GRCh38
rs1016058662 Chr.18: 5144413-5144413 on Build GRCh38
rs1028931095 Chr.18: 5144339-5144339 on Build GRCh38
rs549915735 Chr.18: 5144044-5144044 on Build GRCh38
rs866101217 Chr.18: 5144128-5144128 on Build GRCh38
rs999246025 Chr.18: 5144293-5144293 on Build GRCh38
rs895769683 Chr.18: 5144073-5144073 on Build GRCh38
rs748983948 Chr.18: 5144446-5144446 on Build GRCh38
rs750732479 Chr.18: 5144139-5144139 on Build GRCh38
rs917274596 Chr.18: 5144372-5144372 on Build GRCh38
rs950062454 Chr.18: 5144430-5144430 on Build GRCh38
rs569766504 Chr.18: 5144087-5144087 on Build GRCh38
rs565919638 Chr.18: 5144270-5144270 on Build GRCh38
rs972167582 Chr.18: 5144434-5144434 on Build GRCh38
rs763976203 Chr.18: 5144060-5144060 on Build GRCh38

View Full Product Details