Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5145114-5145654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAIN1
Entrez Gene ID
642597
Gene Name
A kinase (PRKA) anchor inhibitor 1
Gene Aliases
C18orf42
UniGene
Hs.437160
Gene Chromosome Location
Chr.18: 5143673-5199997 on Build GRCh38
Transcripts
NM_001145194.1 XM_011525728.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4921621 c.153C>T p.N51N Chr.18: 5145619-5145619 on Build GRCh38
4774218 c.155G>A p.R52Q Chr.18: 5145617-5145617 on Build GRCh38
4638216 c.204G>T p.K68N Chr.18: 5145568-5145568 on Build GRCh38
4627597 c.198C>T p.H66H Chr.18: 5145574-5145574 on Build GRCh38
3821577 c.162C>A p.H54Q Chr.18: 5145610-5145610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972252151 Chr.18: 5145169-5145169 on Build GRCh38
rs142147429 Chr.18: 5145536-5145536 on Build GRCh38
rs984753179 Chr.18: 5145502-5145502 on Build GRCh38
rs553093719 Chr.18: 5145388-5145388 on Build GRCh38
rs767624362 Chr.18: 5145255-5145255 on Build GRCh38
rs965166326 Chr.18: 5145521-5145521 on Build GRCh38
rs745657091 Chr.18: 5145597-5145597 on Build GRCh38
rs753254899 Chr.18: 5145329-5145329 on Build GRCh38
rs894106435 Chr.18: 5145603-5145603 on Build GRCh38
rs1395063 Chr.18: 5145610-5145610 on Build GRCh38
rs113481842 Chr.18: 5145378-5145378 on Build GRCh38
rs933783719 Chr.18: 5145577-5145577 on Build GRCh38
rs150989410 Chr.18: 5145460-5145460 on Build GRCh38
rs1395064 Chr.18: 5145390-5145390 on Build GRCh38
rs895646791 Chr.18: 5145598-5145598 on Build GRCh38
rs993725993 Chr.18: 5145470-5145470 on Build GRCh38
rs1022948323 Chr.18: 5145363-5145363 on Build GRCh38
rs1028307771 Chr.18: 5145173-5145173 on Build GRCh38
rs148905078 Chr.18: 5145204-5145204 on Build GRCh38
rs952073396 Chr.18: 5145484-5145484 on Build GRCh38
rs933090460 Chr.18: 5145469-5145469 on Build GRCh38
rs770449987 Chr.18: 5145445-5145445 on Build GRCh38
rs935689481 Chr.18: 5145596-5145596 on Build GRCh38
rs1013117155 Chr.18: 5145435-5145435 on Build GRCh38
rs1026513127 Chr.18: 5145471-5145471 on Build GRCh38
rs977363812 Chr.18: 5145590-5145590 on Build GRCh38
rs775641340 Chr.18: 5145618-5145618 on Build GRCh38
rs929178332 Chr.18: 5145593-5145593 on Build GRCh38
rs554491696 Chr.18: 5145286-5145286 on Build GRCh38
rs370601324 Chr.18: 5145574-5145574 on Build GRCh38
rs897713968 Chr.18: 5145228-5145228 on Build GRCh38
rs948494552 Chr.18: 5145605-5145605 on Build GRCh38
rs952021136 Chr.18: 5145206-5145206 on Build GRCh38
rs888775743 Chr.18: 5145168-5145168 on Build GRCh38
rs1041632227 Chr.18: 5145359-5145359 on Build GRCh38
rs429311 Chr.18: 5145162-5145162 on Build GRCh38
rs551328645 Chr.18: 5145264-5145264 on Build GRCh38
rs933897796 Chr.18: 5145346-5145346 on Build GRCh38
rs1017723668 Chr.18: 5145503-5145503 on Build GRCh38
rs924501680 Chr.18: 5145595-5145595 on Build GRCh38
rs902863119 Chr.18: 5145374-5145374 on Build GRCh38
rs976949047 Chr.18: 5145428-5145428 on Build GRCh38
rs984871404 Chr.18: 5145230-5145230 on Build GRCh38
rs941970981 Chr.18: 5145198-5145198 on Build GRCh38
rs941724144 Chr.18: 5145573-5145573 on Build GRCh38
rs778145713 Chr.18: 5145459-5145459 on Build GRCh38
rs543765169 Chr.18: 5145364-5145364 on Build GRCh38
rs145900247 Chr.18: 5145585-5145585 on Build GRCh38

View Full Product Details