Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5145401-5145913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAIN1
Entrez Gene ID
642597
Gene Name
A kinase (PRKA) anchor inhibitor 1
Gene Aliases
C18orf42
UniGene
Hs.437160
Gene Chromosome Location
Chr.18: 5143673-5199997 on Build GRCh38
Transcripts
NM_001145194.1 XM_011525728.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4921621 c.153C>T p.N51N Chr.18: 5145619-5145619 on Build GRCh38
4774218 c.155G>A p.R52Q Chr.18: 5145617-5145617 on Build GRCh38
4638216 c.204G>T p.K68N Chr.18: 5145568-5145568 on Build GRCh38
4627597 c.198C>T p.H66H Chr.18: 5145574-5145574 on Build GRCh38
3821577 c.162C>A p.H54Q Chr.18: 5145610-5145610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531159940 Chr.18: 5145863-5145863 on Build GRCh38
rs142147429 Chr.18: 5145536-5145536 on Build GRCh38
rs977398965 Chr.18: 5145727-5145727 on Build GRCh38
rs1395063 Chr.18: 5145610-5145610 on Build GRCh38
rs1029864225 Chr.18: 5145759-5145759 on Build GRCh38
rs933783719 Chr.18: 5145577-5145577 on Build GRCh38
rs770449987 Chr.18: 5145445-5145445 on Build GRCh38
rs376977829 Chr.18: 5145645-5145645 on Build GRCh38
rs1013117155 Chr.18: 5145435-5145435 on Build GRCh38
rs367797718 Chr.18: 5145673-5145673 on Build GRCh38
rs924555473 Chr.18: 5145877-5145877 on Build GRCh38
rs1026513127 Chr.18: 5145471-5145471 on Build GRCh38
rs977363812 Chr.18: 5145590-5145590 on Build GRCh38
rs929178332 Chr.18: 5145593-5145593 on Build GRCh38
rs775641340 Chr.18: 5145618-5145618 on Build GRCh38
rs768068415 Chr.18: 5145695-5145695 on Build GRCh38
rs373261652 Chr.18: 5145644-5145644 on Build GRCh38
rs528582877 Chr.18: 5145778-5145778 on Build GRCh38
rs985603058 Chr.18: 5145799-5145799 on Build GRCh38
rs773755820 Chr.18: 5145712-5145712 on Build GRCh38
rs74408409 Chr.18: 5145841-5145841 on Build GRCh38
rs924501680 Chr.18: 5145595-5145595 on Build GRCh38
rs907412229 Chr.18: 5145650-5145650 on Build GRCh38
rs976949047 Chr.18: 5145428-5145428 on Build GRCh38
rs564863673 Chr.18: 5145866-5145866 on Build GRCh38
rs768553299 Chr.18: 5145662-5145662 on Build GRCh38
rs778145713 Chr.18: 5145459-5145459 on Build GRCh38
rs763453875 Chr.18: 5145795-5145795 on Build GRCh38
rs957280798 Chr.18: 5145879-5145879 on Build GRCh38
rs984753179 Chr.18: 5145502-5145502 on Build GRCh38
rs756282119 Chr.18: 5145685-5145686 on Build GRCh38
rs777946283 Chr.18: 5145739-5145740 on Build GRCh38
rs965166326 Chr.18: 5145521-5145521 on Build GRCh38
rs745657091 Chr.18: 5145597-5145597 on Build GRCh38
rs894106435 Chr.18: 5145603-5145603 on Build GRCh38
rs150989410 Chr.18: 5145460-5145460 on Build GRCh38
rs190365119 Chr.18: 5145805-5145805 on Build GRCh38
rs1030407910 Chr.18: 5145849-5145849 on Build GRCh38
rs879154642 Chr.18: 5145867-5145867 on Build GRCh38
rs895646791 Chr.18: 5145598-5145598 on Build GRCh38
rs757927146 Chr.18: 5145791-5145791 on Build GRCh38
rs993725993 Chr.18: 5145470-5145470 on Build GRCh38
rs1018026355 Chr.18: 5145752-5145752 on Build GRCh38
rs761094933 Chr.18: 5145754-5145754 on Build GRCh38
rs952073396 Chr.18: 5145484-5145484 on Build GRCh38
rs933090460 Chr.18: 5145469-5145469 on Build GRCh38
rs1026563889 Chr.18: 5145657-5145657 on Build GRCh38
rs935689481 Chr.18: 5145596-5145596 on Build GRCh38
rs138717602 Chr.18: 5145755-5145755 on Build GRCh38
rs752355934 Chr.18: 5145782-5145782 on Build GRCh38
rs904585599 Chr.18: 5145649-5145649 on Build GRCh38
rs370601324 Chr.18: 5145574-5145574 on Build GRCh38
rs948494552 Chr.18: 5145605-5145605 on Build GRCh38
rs771539300 Chr.18: 5145854-5145854 on Build GRCh38
rs762287832 Chr.18: 5145688-5145688 on Build GRCh38
rs1017723668 Chr.18: 5145503-5145503 on Build GRCh38
rs573697917 Chr.18: 5145667-5145667 on Build GRCh38
rs993610569 Chr.18: 5145656-5145656 on Build GRCh38
rs941724144 Chr.18: 5145573-5145573 on Build GRCh38
rs145900247 Chr.18: 5145585-5145585 on Build GRCh38

View Full Product Details