Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5396014-5396547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L3
Entrez Gene ID
23136
Gene Name
erythrocyte membrane protein band 4.1 like 3
Gene Aliases
4.1B, DAL-1, DAL1
UniGene
Hs.213394
Gene Chromosome Location
Chr.18: 5392381-5630666 on Build GRCh38
Transcripts
NM_001281533.1 NM_001281534.1 NM_001281535.1 NM_012307.3 XM_011525623.2 XM_011525626.2 XM_011525628.2 XM_011525632.2 XM_011525634.2 XM_011525637.2 XM_017025618.1 XM_017025619.1 XM_017025620.1 XM_017025621.1 XM_017025622.1 XM_017025623.1 XM_017025624.1 XM_017025625.1 XM_017025626.1 XM_017025627.1 XM_017025628.1 XM_017025629.1 XM_017025630.1 XM_017025631.1 XM_017025632.1 XM_017025633.1 XM_017025634.1 XM_017025635.1 XM_017025636.1 XM_017025637.1 XM_017025638.1 XM_017025639.1 XM_017025640.1 XM_017025641.1 XM_017025642.1 XM_017025643.1 XM_017025644.1 XM_017025645.1 XM_017025646.1 XM_017025647.1 XM_017025648.1 XM_017025649.1 XM_017025650.1 XM_017025651.1 XM_017025652.1 XM_017025653.1 XM_017025654.1 XM_017025655.1 XM_017025656.1 XM_017025657.1 XM_017025658.1 XM_017025659.1 XM_017025660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6340089 c.803C>A p.P268Q Chr.18: 5396286-5396286 on Build GRCh38
4931474 c.2960A>G p.Y987C Chr.18: 5396214-5396214 on Build GRCh38
989057 c.2875G>T p.G959C Chr.18: 5396299-5396299 on Build GRCh38
3526607 c.2352G>A p.T784T Chr.18: 5396315-5396315 on Build GRCh38
4275851 c.870C>A p.I290I Chr.18: 5396219-5396219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779517862 Chr.18: 5396207-5396207 on Build GRCh38
rs772420921 Chr.18: 5396213-5396213 on Build GRCh38
rs752012770 Chr.18: 5396495-5396496 on Build GRCh38
rs569678650 Chr.18: 5396384-5396384 on Build GRCh38
rs889360838 Chr.18: 5396070-5396070 on Build GRCh38
rs763721094 Chr.18: 5396118-5396118 on Build GRCh38
rs922269288 Chr.18: 5396078-5396078 on Build GRCh38
rs112187992 Chr.18: 5396045-5396046 on Build GRCh38
rs956341043 Chr.18: 5396287-5396287 on Build GRCh38
rs763191755 Chr.18: 5396188-5396188 on Build GRCh38
rs146947191 Chr.18: 5396059-5396059 on Build GRCh38
rs187762164 Chr.18: 5396072-5396072 on Build GRCh38
rs924777324 Chr.18: 5396099-5396099 on Build GRCh38
rs916048308 Chr.18: 5396449-5396449 on Build GRCh38
rs548283428 Chr.18: 5396084-5396084 on Build GRCh38
rs977669358 Chr.18: 5396082-5396082 on Build GRCh38
rs766204497 Chr.18: 5396505-5396507 on Build GRCh38
rs543291019 Chr.18: 5396060-5396060 on Build GRCh38
rs929311751 Chr.18: 5396430-5396430 on Build GRCh38
rs955030032 Chr.18: 5396391-5396391 on Build GRCh38
rs572428305 Chr.18: 5396401-5396401 on Build GRCh38
rs890599273 Chr.18: 5396491-5396491 on Build GRCh38
rs751101936 Chr.18: 5396520-5396520 on Build GRCh38
rs761006000 Chr.18: 5396306-5396306 on Build GRCh38
rs907165968 Chr.18: 5396183-5396183 on Build GRCh38
rs990304605 Chr.18: 5396442-5396442 on Build GRCh38
rs779642049 Chr.18: 5396345-5396345 on Build GRCh38
rs779573005 Chr.18: 5396324-5396324 on Build GRCh38
rs773650513 Chr.18: 5396108-5396108 on Build GRCh38
rs371591392 Chr.18: 5396233-5396233 on Build GRCh38
rs1013412227 Chr.18: 5396181-5396181 on Build GRCh38
rs936213491 Chr.18: 5396122-5396122 on Build GRCh38
rs1016725836 Chr.18: 5396054-5396054 on Build GRCh38
rs145344390 Chr.18: 5396273-5396273 on Build GRCh38
rs148045283 Chr.18: 5396315-5396315 on Build GRCh38
rs1046157942 Chr.18: 5396175-5396175 on Build GRCh38
rs979314894 Chr.18: 5396282-5396282 on Build GRCh38
rs57191955 Chr.18: 5396492-5396493 on Build GRCh38
rs112698199 Chr.18: 5396490-5396491 on Build GRCh38
rs750231606 Chr.18: 5396496-5396501 on Build GRCh38
rs928707605 Chr.18: 5396483-5396483 on Build GRCh38
rs767200554 Chr.18: 5396469-5396469 on Build GRCh38
rs748802311 Chr.18: 5396346-5396346 on Build GRCh38
rs768097979 Chr.18: 5396367-5396367 on Build GRCh38
rs781692643 Chr.18: 5396497-5396507 on Build GRCh38
rs948790432 Chr.18: 5396143-5396143 on Build GRCh38
rs201649249 Chr.18: 5396044-5396045 on Build GRCh38
rs138017302 Chr.18: 5396208-5396208 on Build GRCh38
rs887047678 Chr.18: 5396522-5396522 on Build GRCh38
rs138604700 Chr.18: 5396478-5396478 on Build GRCh38
rs778244742 Chr.18: 5396335-5396335 on Build GRCh38
rs115131748 Chr.18: 5396081-5396081 on Build GRCh38
rs368155284 Chr.18: 5396179-5396179 on Build GRCh38
rs929356049 Chr.18: 5396503-5396503 on Build GRCh38
rs755560965 Chr.18: 5396192-5396192 on Build GRCh38
rs570185440 Chr.18: 5396281-5396281 on Build GRCh38
rs150138682 Chr.18: 5396420-5396420 on Build GRCh38
rs770143511 Chr.18: 5396085-5396085 on Build GRCh38
rs993926303 Chr.18: 5396112-5396112 on Build GRCh38
rs747316632 Chr.18: 5396340-5396340 on Build GRCh38
rs79592897 Chr.18: 5396259-5396259 on Build GRCh38
rs556863431 Chr.18: 5396383-5396383 on Build GRCh38
rs750152997 Chr.18: 5396286-5396286 on Build GRCh38
rs1057450261 Chr.18: 5396441-5396441 on Build GRCh38
rs543820358 Chr.18: 5396120-5396120 on Build GRCh38
rs766462217 Chr.18: 5396309-5396309 on Build GRCh38
rs367547148 Chr.18: 5396046-5396046 on Build GRCh38
rs571003872 Chr.18: 5396278-5396278 on Build GRCh38
rs61735459 Chr.18: 5396325-5396325 on Build GRCh38
rs745604519 Chr.18: 5396234-5396234 on Build GRCh38
rs887015258 Chr.18: 5396253-5396253 on Build GRCh38
rs758476503 Chr.18: 5396503-5396503 on Build GRCh38
rs775033833 Chr.18: 5396239-5396239 on Build GRCh38
rs879796765 Chr.18: 5396397-5396397 on Build GRCh38
rs148372047 Chr.18: 5396363-5396364 on Build GRCh38
rs924902279 Chr.18: 5396415-5396415 on Build GRCh38
rs541104882 Chr.18: 5396499-5396499 on Build GRCh38
rs536788198 Chr.18: 5396133-5396133 on Build GRCh38
rs916027131 Chr.18: 5396139-5396139 on Build GRCh38
rs1016758548 Chr.18: 5396272-5396272 on Build GRCh38
rs773845261 Chr.18: 5396376-5396376 on Build GRCh38
rs750713407 Chr.18: 5396150-5396150 on Build GRCh38
rs375597366 Chr.18: 5396285-5396285 on Build GRCh38
rs769509413 Chr.18: 5396344-5396344 on Build GRCh38
rs780222810 Chr.18: 5396184-5396184 on Build GRCh38
rs878902748 Chr.18: 5396364-5396364 on Build GRCh38
rs755038593 Chr.18: 5396316-5396316 on Build GRCh38
rs527779795 Chr.18: 5396258-5396258 on Build GRCh38
rs58817821 Chr.18: 5396514-5396514 on Build GRCh38
rs535393984 Chr.18: 5396385-5396385 on Build GRCh38
rs988239687 Chr.18: 5396298-5396298 on Build GRCh38
rs375999728 Chr.18: 5396046-5396046 on Build GRCh38

View Full Product Details