Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 12673408-12673953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP76
Entrez Gene ID
79959
Gene Name
centrosomal protein 76
Gene Aliases
C18orf9, HsT1705
UniGene
Hs.236940
Gene Chromosome Location
Chr.18: 12661733-12702777 on Build GRCh38
Transcripts
NM_001271989.1 NM_024899.3 NR_073537.1 XM_005258149.4 XM_017025981.1 XM_017025982.1 XM_017025983.1 XM_017025984.1 XM_017025985.1 XM_017025986.1 XM_017025987.1 NM_147163.1

Gene Symbol
PSMG2
Entrez Gene ID
56984
Gene Name
proteasome assembly chaperone 2
Gene Aliases
CLAST3, HCCA3, HsT1707, MDS003, PAC2, TNFSF5IP1
UniGene
Hs.464652
Gene Chromosome Location
Chr.18: 12658739-12725740 on Build GRCh38
Transcripts
NM_001271989.1 NM_024899.3 NR_073537.1 XM_005258149.4 XM_017025981.1 XM_017025982.1 XM_017025983.1 XM_017025984.1 XM_017025985.1 XM_017025986.1 XM_017025987.1 NM_147163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5574153 c.1842-6_1842-5insTCTT p.? Chr.18: 12673508-12673509 on Build GRCh38
6003204 c.1900C>T p.R634C Chr.18: 12673445-12673445 on Build GRCh38
6218297 c.1929T>C p.S643S Chr.18: 12673416-12673416 on Build GRCh38
5548211 c.1842-10delT p.? Chr.18: 12673513-12673513 on Build GRCh38
6190007 c.1844C>T p.S615F Chr.18: 12673501-12673501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752598071 Chr.18: 12673508-12673508 on Build GRCh38
rs946934567 Chr.18: 12673793-12673793 on Build GRCh38
rs766404126 Chr.18: 12673512-12673513 on Build GRCh38
rs77719285 Chr.18: 12673513-12673513 on Build GRCh38
rs749875550 Chr.18: 12673796-12673796 on Build GRCh38
rs893047656 Chr.18: 12673768-12673768 on Build GRCh38
rs755939319 Chr.18: 12673541-12673541 on Build GRCh38
rs758604348 Chr.18: 12673512-12673512 on Build GRCh38
rs985012147 Chr.18: 12673859-12673859 on Build GRCh38
rs768282968 Chr.18: 12673439-12673439 on Build GRCh38
rs542652432 Chr.18: 12673828-12673828 on Build GRCh38
rs922206115 Chr.18: 12673651-12673651 on Build GRCh38
rs974863381 Chr.18: 12673838-12673838 on Build GRCh38
rs12959199 Chr.18: 12673575-12673575 on Build GRCh38
rs1050623946 Chr.18: 12673645-12673645 on Build GRCh38
rs1039663898 Chr.18: 12673646-12673646 on Build GRCh38
rs771629415 Chr.18: 12673460-12673460 on Build GRCh38
rs556470881 Chr.18: 12673774-12673774 on Build GRCh38
rs765300466 Chr.18: 12673499-12673499 on Build GRCh38
rs2542166 Chr.18: 12673849-12673849 on Build GRCh38
rs556525556 Chr.18: 12673738-12673738 on Build GRCh38
rs1001396261 Chr.18: 12673750-12673750 on Build GRCh38
rs761143650 Chr.18: 12673479-12673479 on Build GRCh38
rs372117728 Chr.18: 12673485-12673485 on Build GRCh38
rs185903279 Chr.18: 12673807-12673807 on Build GRCh38
rs922132685 Chr.18: 12673843-12673843 on Build GRCh38
rs993160878 Chr.18: 12673860-12673860 on Build GRCh38
rs912007348 Chr.18: 12673614-12673614 on Build GRCh38
rs547557605 Chr.18: 12673470-12673470 on Build GRCh38
rs45589535 Chr.18: 12673722-12673722 on Build GRCh38
rs1050596022 Chr.18: 12673794-12673794 on Build GRCh38
rs146390170 Chr.18: 12673708-12673708 on Build GRCh38
rs948115705 Chr.18: 12673524-12673524 on Build GRCh38
rs759520853 Chr.18: 12673498-12673498 on Build GRCh38
rs111451708 Chr.18: 12673540-12673540 on Build GRCh38
rs1008620959 Chr.18: 12673706-12673706 on Build GRCh38
rs182217907 Chr.18: 12673804-12673804 on Build GRCh38
rs1018596817 Chr.18: 12673445-12673445 on Build GRCh38
rs887716220 Chr.18: 12673880-12673880 on Build GRCh38
rs762653437 Chr.18: 12673522-12673522 on Build GRCh38
rs751437963 Chr.18: 12673513-12673513 on Build GRCh38
rs986605208 Chr.18: 12673606-12673606 on Build GRCh38
rs1005338330 Chr.18: 12673733-12673733 on Build GRCh38
rs915474736 Chr.18: 12673778-12673778 on Build GRCh38
rs549384161 Chr.18: 12673695-12673695 on Build GRCh38
rs528198617 Chr.18: 12673881-12673881 on Build GRCh38
rs376355586 Chr.18: 12673751-12673751 on Build GRCh38
rs982123695 Chr.18: 12673752-12673752 on Build GRCh38
rs764026270 Chr.18: 12673547-12673547 on Build GRCh38
rs762909227 Chr.18: 12673502-12673505 on Build GRCh38
rs957005197 Chr.18: 12673568-12673568 on Build GRCh38
rs912933116 Chr.18: 12673577-12673577 on Build GRCh38
rs1046210492 Chr.18: 12673885-12673885 on Build GRCh38
rs944899007 Chr.18: 12673642-12673642 on Build GRCh38
rs967109549 Chr.18: 12673588-12673588 on Build GRCh38
rs1019372506 Chr.18: 12673824-12673824 on Build GRCh38
rs368863361 Chr.18: 12673481-12673481 on Build GRCh38
rs1046171903 Chr.18: 12673704-12673704 on Build GRCh38
rs907269014 Chr.18: 12673892-12673892 on Build GRCh38
rs986919998 Chr.18: 12673816-12673816 on Build GRCh38
rs930920656 Chr.18: 12673663-12673663 on Build GRCh38
rs951995476 Chr.18: 12673741-12673741 on Build GRCh38
rs928006089 Chr.18: 12673755-12673755 on Build GRCh38
rs937454056 Chr.18: 12673894-12673894 on Build GRCh38
rs752722510 Chr.18: 12673506-12673506 on Build GRCh38
rs907848007 Chr.18: 12673868-12673868 on Build GRCh38
rs940439145 Chr.18: 12673739-12673739 on Build GRCh38
rs542820071 Chr.18: 12673819-12673819 on Build GRCh38
rs888883962 Chr.18: 12673688-12673688 on Build GRCh38
rs374138097 Chr.18: 12673456-12673456 on Build GRCh38
rs754240806 Chr.18: 12673581-12673581 on Build GRCh38
rs371344209 Chr.18: 12673440-12673440 on Build GRCh38
rs370698496 Chr.18: 12673533-12673535 on Build GRCh38
rs1055631053 Chr.18: 12673919-12673919 on Build GRCh38
rs991358894 Chr.18: 12673742-12673742 on Build GRCh38
rs915718080 Chr.18: 12673747-12673747 on Build GRCh38
rs1045824527 Chr.18: 12673737-12673737 on Build GRCh38

View Full Product Details