Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 12677936-12678437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP76
Entrez Gene ID
79959
Gene Name
centrosomal protein 76
Gene Aliases
C18orf9, HsT1705
UniGene
Hs.236940
Gene Chromosome Location
Chr.18: 12661733-12702777 on Build GRCh38
Transcripts
NM_001271989.1 NM_024899.3 NR_073537.1 XM_005258149.4 XM_017025981.1 XM_017025982.1 XM_017025983.1 XM_017025984.1 XM_017025985.1 XM_017025986.1 XM_017025987.1 NM_147163.1

Gene Symbol
PSMG2
Entrez Gene ID
56984
Gene Name
proteasome assembly chaperone 2
Gene Aliases
CLAST3, HCCA3, HsT1707, MDS003, PAC2, TNFSF5IP1
UniGene
Hs.464652
Gene Chromosome Location
Chr.18: 12658739-12725740 on Build GRCh38
Transcripts
NM_001271989.1 NM_024899.3 NR_073537.1 XM_005258149.4 XM_017025981.1 XM_017025982.1 XM_017025983.1 XM_017025984.1 XM_017025985.1 XM_017025986.1 XM_017025987.1 NM_147163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
365715 c.1601T>C p.L534P Chr.18: 12678131-12678131 on Build GRCh38
4839042 c.1301A>C p.K434T Chr.18: 12678431-12678431 on Build GRCh38
4808358 c.1529C>T p.P510L Chr.18: 12678203-12678203 on Build GRCh38
6627165 c.1450G>A p.D484N Chr.18: 12678282-12678282 on Build GRCh38
986653 c.1327G>A p.V443I Chr.18: 12678405-12678405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746207911 Chr.18: 12678100-12678100 on Build GRCh38
rs779390805 Chr.18: 12678391-12678391 on Build GRCh38
rs761369841 Chr.18: 12678316-12678316 on Build GRCh38
rs903278909 Chr.18: 12678246-12678246 on Build GRCh38
rs943087907 Chr.18: 12678169-12678169 on Build GRCh38
rs756920702 Chr.18: 12678083-12678083 on Build GRCh38
rs780891111 Chr.18: 12678192-12678192 on Build GRCh38
rs762889371 Chr.18: 12678288-12678288 on Build GRCh38
rs773472463 Chr.18: 12678239-12678239 on Build GRCh38
rs142832499 Chr.18: 12678132-12678132 on Build GRCh38
rs755686355 Chr.18: 12678336-12678336 on Build GRCh38
rs370425822 Chr.18: 12678149-12678149 on Build GRCh38
rs896346924 Chr.18: 12678218-12678218 on Build GRCh38
rs754179948 Chr.18: 12678371-12678371 on Build GRCh38
rs777350700 Chr.18: 12678074-12678074 on Build GRCh38
rs761904667 Chr.18: 12678137-12678137 on Build GRCh38
rs770120941 Chr.18: 12678103-12678103 on Build GRCh38
rs747134192 Chr.18: 12678407-12678407 on Build GRCh38
rs36092016 Chr.18: 12678334-12678335 on Build GRCh38
rs759114067 Chr.18: 12678411-12678411 on Build GRCh38
rs374370690 Chr.18: 12678089-12678089 on Build GRCh38
rs780672726 Chr.18: 12678071-12678073 on Build GRCh38
rs986592194 Chr.18: 12678118-12678118 on Build GRCh38
rs769159491 Chr.18: 12678129-12678129 on Build GRCh38
rs370679694 Chr.18: 12678347-12678347 on Build GRCh38
rs776340224 Chr.18: 12678321-12678321 on Build GRCh38
rs755375295 Chr.18: 12678377-12678377 on Build GRCh38
rs750097953 Chr.18: 12678193-12678193 on Build GRCh38
rs748498676 Chr.18: 12678366-12678366 on Build GRCh38
rs771036468 Chr.18: 12678408-12678408 on Build GRCh38
rs955716836 Chr.18: 12678115-12678115 on Build GRCh38
rs766318200 Chr.18: 12678314-12678314 on Build GRCh38
rs764882115 Chr.18: 12678170-12678170 on Build GRCh38
rs1051374949 Chr.18: 12678130-12678130 on Build GRCh38
rs199846298 Chr.18: 12678397-12678397 on Build GRCh38
rs751610398 Chr.18: 12678310-12678310 on Build GRCh38
rs570655668 Chr.18: 12678320-12678320 on Build GRCh38
rs921466436 Chr.18: 12678090-12678090 on Build GRCh38
rs749740623 Chr.18: 12678121-12678121 on Build GRCh38
rs766876816 Chr.18: 12678322-12678322 on Build GRCh38
rs151031205 Chr.18: 12678230-12678230 on Build GRCh38
rs752644871 Chr.18: 12678072-12678072 on Build GRCh38
rs530964720 Chr.18: 12678166-12678166 on Build GRCh38
rs370376860 Chr.18: 12678201-12678201 on Build GRCh38
rs1037424736 Chr.18: 12678184-12678184 on Build GRCh38
rs769163684 Chr.18: 12678190-12678191 on Build GRCh38
rs1024942795 Chr.18: 12678309-12678309 on Build GRCh38
rs999278452 Chr.18: 12677987-12677989 on Build GRCh38
rs369987338 Chr.18: 12678063-12678063 on Build GRCh38
rs372594882 Chr.18: 12678172-12678172 on Build GRCh38
rs141700810 Chr.18: 12678031-12678031 on Build GRCh38
rs747488126 Chr.18: 12678141-12678142 on Build GRCh38
rs200920017 Chr.18: 12678228-12678228 on Build GRCh38
rs769106251 Chr.18: 12678214-12678214 on Build GRCh38
rs1013817335 Chr.18: 12678219-12678219 on Build GRCh38
rs746548620 Chr.18: 12678082-12678082 on Build GRCh38
rs937966871 Chr.18: 12678189-12678189 on Build GRCh38
rs772924378 Chr.18: 12678365-12678365 on Build GRCh38
rs781703963 Chr.18: 12678214-12678216 on Build GRCh38
rs958174060 Chr.18: 12678343-12678343 on Build GRCh38
rs1025838504 Chr.18: 12678256-12678256 on Build GRCh38
rs756863527 Chr.18: 12678186-12678186 on Build GRCh38
rs749956570 Chr.18: 12678328-12678328 on Build GRCh38
rs771214847 Chr.18: 12678261-12678261 on Build GRCh38
rs1032983814 Chr.18: 12677997-12677999 on Build GRCh38
rs998906335 Chr.18: 12678257-12678257 on Build GRCh38
rs755808154 Chr.18: 12678194-12678194 on Build GRCh38
rs754777943 Chr.18: 12678062-12678062 on Build GRCh38
rs377176858 Chr.18: 12678198-12678198 on Build GRCh38
rs758422946 Chr.18: 12678073-12678073 on Build GRCh38
rs777672974 Chr.18: 12678405-12678405 on Build GRCh38
rs1013885052 Chr.18: 12678307-12678307 on Build GRCh38
rs370708558 Chr.18: 12678217-12678217 on Build GRCh38
rs377711940 Chr.18: 12678105-12678105 on Build GRCh38
rs767757265 Chr.18: 12678143-12678143 on Build GRCh38
rs764088609 Chr.18: 12678305-12678305 on Build GRCh38
rs751088597 Chr.18: 12678182-12678182 on Build GRCh38
rs997130485 Chr.18: 12678401-12678401 on Build GRCh38
rs772635325 Chr.18: 12678400-12678400 on Build GRCh38
rs760740251 Chr.18: 12678253-12678253 on Build GRCh38
rs764149719 Chr.18: 12678177-12678177 on Build GRCh38
rs139815657 Chr.18: 12678283-12678283 on Build GRCh38
rs752539145 Chr.18: 12678171-12678171 on Build GRCh38

View Full Product Details