Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13019002-13019501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP192
Entrez Gene ID
55125
Gene Name
centrosomal protein 192
Gene Aliases
PPP1R62
UniGene
Hs.100914
Gene Chromosome Location
Chr.18: 12991307-13125053 on Build GRCh38
Transcripts
NM_032142.3 XM_005258107.3 XM_005258109.3 XM_006722326.3 XM_006722327.3 XM_006722330.3 XM_011525673.2 XM_011525675.2 XM_017025803.1 XM_017025804.1 XM_017025806.1 XM_017025807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4983959 c.1042C>T p.L348F Chr.18: 13019198-13019198 on Build GRCh38
4792184 c.968G>A p.G323D Chr.18: 13019124-13019124 on Build GRCh38
7360878 c.932C>G p.S311C Chr.18: 13019088-13019088 on Build GRCh38
6307030 c.991A>G p.T331A Chr.18: 13019147-13019147 on Build GRCh38
3356992 c.983C>T p.S328F Chr.18: 13019139-13019139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774734734 Chr.18: 13019255-13019255 on Build GRCh38
rs534489071 Chr.18: 13019365-13019365 on Build GRCh38
rs149193512 Chr.18: 13019206-13019206 on Build GRCh38
rs974199526 Chr.18: 13019169-13019169 on Build GRCh38
rs747781590 Chr.18: 13019109-13019109 on Build GRCh38
rs746402954 Chr.18: 13019177-13019177 on Build GRCh38
rs1029824285 Chr.18: 13019437-13019437 on Build GRCh38
rs991830871 Chr.18: 13019244-13019244 on Build GRCh38
rs576968484 Chr.18: 13019372-13019372 on Build GRCh38
rs142378018 Chr.18: 13019264-13019264 on Build GRCh38
rs1010212951 Chr.18: 13019058-13019058 on Build GRCh38
rs1051041245 Chr.18: 13019422-13019422 on Build GRCh38
rs560112462 Chr.18: 13019032-13019032 on Build GRCh38
rs74420724 Chr.18: 13019103-13019103 on Build GRCh38
rs748529794 Chr.18: 13019357-13019357 on Build GRCh38
rs1043424383 Chr.18: 13019029-13019029 on Build GRCh38
rs186163226 Chr.18: 13019341-13019341 on Build GRCh38
rs768940114 Chr.18: 13019242-13019242 on Build GRCh38
rs946018648 Chr.18: 13019406-13019406 on Build GRCh38
rs751406029 Chr.18: 13019252-13019252 on Build GRCh38
rs753750160 Chr.18: 13019079-13019079 on Build GRCh38
rs758109660 Chr.18: 13019124-13019124 on Build GRCh38
rs8096860 Chr.18: 13019337-13019337 on Build GRCh38
rs781698192 Chr.18: 13019176-13019176 on Build GRCh38
rs182182159 Chr.18: 13019392-13019392 on Build GRCh38
rs531384100 Chr.18: 13019229-13019229 on Build GRCh38
rs962600742 Chr.18: 13019149-13019149 on Build GRCh38
rs998279151 Chr.18: 13019134-13019134 on Build GRCh38
rs775771818 Chr.18: 13019215-13019215 on Build GRCh38
rs56204439 Chr.18: 13019446-13019446 on Build GRCh38
rs191065107 Chr.18: 13019352-13019352 on Build GRCh38
rs747319531 Chr.18: 13019140-13019140 on Build GRCh38
rs980881905 Chr.18: 13019280-13019280 on Build GRCh38
rs551920435 Chr.18: 13019351-13019351 on Build GRCh38
rs1025859814 Chr.18: 13019104-13019104 on Build GRCh38
rs1018504082 Chr.18: 13019129-13019129 on Build GRCh38
rs186509602 Chr.18: 13019434-13019434 on Build GRCh38
rs939182620 Chr.18: 13019295-13019295 on Build GRCh38
rs375542687 Chr.18: 13019148-13019148 on Build GRCh38
rs777410032 Chr.18: 13019125-13019125 on Build GRCh38
rs555825426 Chr.18: 13019368-13019368 on Build GRCh38
rs964196728 Chr.18: 13019127-13019127 on Build GRCh38
rs927440693 Chr.18: 13019279-13019279 on Build GRCh38
rs768390672 Chr.18: 13019276-13019276 on Build GRCh38
rs913719709 Chr.18: 13019322-13019322 on Build GRCh38
rs868211851 Chr.18: 13019400-13019400 on Build GRCh38
rs1056343193 Chr.18: 13019297-13019297 on Build GRCh38
rs926670692 Chr.18: 13019289-13019289 on Build GRCh38
rs375162118 Chr.18: 13019416-13019416 on Build GRCh38
rs891886531 Chr.18: 13019402-13019402 on Build GRCh38
rs754478857 Chr.18: 13019082-13019082 on Build GRCh38
rs1029497131 Chr.18: 13019067-13019067 on Build GRCh38
rs1017729655 Chr.18: 13019126-13019126 on Build GRCh38
rs936198025 Chr.18: 13019312-13019312 on Build GRCh38
rs891769456 Chr.18: 13019030-13019030 on Build GRCh38

View Full Product Details