Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13277901-13278397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAD4
Entrez Gene ID
753
Gene Name
low density lipoprotein receptor class A domain containing 4
Gene Aliases
C18orf1
UniGene
Hs.731853
Gene Chromosome Location
Chr.18: 13217691-13652754 on Build GRCh38
Transcripts
NM_181481.4 NM_181482.4 XM_005258140.1 XM_006722353.1 XM_011525737.1 XM_011525738.1 XM_017025961.1 XM_017025962.1 XM_017025963.1 XM_017025964.1 XM_017025965.1 XM_017025966.1 XM_017025967.1 XM_017025968.1

SNP Detail

SNP ID Chromosome Location
rs182018895 Chr.18: 13278105-13278105 on Build GRCh38
rs568314488 Chr.18: 13278124-13278124 on Build GRCh38
rs971705454 Chr.18: 13277936-13277936 on Build GRCh38
rs960040908 Chr.18: 13278266-13278266 on Build GRCh38
rs919000407 Chr.18: 13278046-13278046 on Build GRCh38
rs895401523 Chr.18: 13277976-13277976 on Build GRCh38
rs528254865 Chr.18: 13278112-13278112 on Build GRCh38
rs930336553 Chr.18: 13277965-13277965 on Build GRCh38
rs62097323 Chr.18: 13277938-13277938 on Build GRCh38
rs780423849 Chr.18: 13278130-13278130 on Build GRCh38
rs141914726 Chr.18: 13278060-13278060 on Build GRCh38
rs777616900 Chr.18: 13277925-13277925 on Build GRCh38
rs948320335 Chr.18: 13278120-13278120 on Build GRCh38
rs936831346 Chr.18: 13278154-13278154 on Build GRCh38
rs112193886 Chr.18: 13277966-13277966 on Build GRCh38
rs889018223 Chr.18: 13278276-13278276 on Build GRCh38
rs71353260 Chr.18: 13278141-13278141 on Build GRCh38
rs1007492520 Chr.18: 13278245-13278245 on Build GRCh38
rs1018884694 Chr.18: 13278303-13278303 on Build GRCh38
rs971776060 Chr.18: 13278165-13278165 on Build GRCh38
rs570974728 Chr.18: 13278288-13278288 on Build GRCh38
rs1026093664 Chr.18: 13278249-13278249 on Build GRCh38
rs746397724 Chr.18: 13278106-13278106 on Build GRCh38
rs377386322 Chr.18: 13278304-13278304 on Build GRCh38
rs1027781854 Chr.18: 13278339-13278339 on Build GRCh38
rs191179380 Chr.18: 13278178-13278178 on Build GRCh38
rs892530142 Chr.18: 13278175-13278175 on Build GRCh38
rs768848460 Chr.18: 13278145-13278145 on Build GRCh38
rs182975715 Chr.18: 13278272-13278272 on Build GRCh38
rs1050488618 Chr.18: 13278254-13278254 on Build GRCh38
rs557806856 Chr.18: 13278247-13278247 on Build GRCh38
rs185902849 Chr.18: 13278116-13278116 on Build GRCh38
rs961847521 Chr.18: 13278332-13278332 on Build GRCh38
rs972301101 Chr.18: 13278248-13278248 on Build GRCh38
rs190930591 Chr.18: 13278056-13278056 on Build GRCh38
rs1030947929 Chr.18: 13278074-13278074 on Build GRCh38
rs1030353908 Chr.18: 13278215-13278215 on Build GRCh38
rs1052562961 Chr.18: 13278160-13278160 on Build GRCh38
rs150634858 Chr.18: 13278218-13278218 on Build GRCh38
rs920373333 Chr.18: 13278167-13278167 on Build GRCh38
rs1046533793 Chr.18: 13278241-13278241 on Build GRCh38
rs759552277 Chr.18: 13278292-13278292 on Build GRCh38
rs993331090 Chr.18: 13278337-13278337 on Build GRCh38
rs574504867 Chr.18: 13278006-13278006 on Build GRCh38
rs550586796 Chr.18: 13278142-13278142 on Build GRCh38
rs186163782 Chr.18: 13278009-13278009 on Build GRCh38
rs770073646 Chr.18: 13278114-13278114 on Build GRCh38
rs370043884 Chr.18: 13278042-13278042 on Build GRCh38
rs951864233 Chr.18: 13278372-13278372 on Build GRCh38
rs1016150863 Chr.18: 13278246-13278246 on Build GRCh38
rs916811753 Chr.18: 13278198-13278198 on Build GRCh38
rs1010926928 Chr.18: 13278118-13278118 on Build GRCh38

View Full Product Details