Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13387250-13387753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAD4
Entrez Gene ID
753
Gene Name
low density lipoprotein receptor class A domain containing 4
Gene Aliases
C18orf1
UniGene
Hs.731853
Gene Chromosome Location
Chr.18: 13217691-13652754 on Build GRCh38
Transcripts
NM_181481.4 NM_181482.4 XM_005258140.1 XM_006722353.1 XM_006722354.1 XM_011525737.1 XM_011525738.1 XM_017025961.1 XM_017025962.1 XM_017025963.1 XM_017025964.1 XM_017025965.1 XM_017025966.1 XM_017025967.1 XM_017025968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5440621 c.19C>T p.Q7* Chr.18: 13387741-13387741 on Build GRCh38
1198282 c.5C>T p.P2L Chr.18: 13387727-13387727 on Build GRCh38
1189649 c.21G>T p.Q7H Chr.18: 13387743-13387743 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs576947318 Chr.18: 13387573-13387573 on Build GRCh38
rs994010570 Chr.18: 13387489-13387489 on Build GRCh38
rs925636933 Chr.18: 13387296-13387296 on Build GRCh38
rs886953784 Chr.18: 13387504-13387504 on Build GRCh38
rs1025433867 Chr.18: 13387315-13387315 on Build GRCh38
rs955121578 Chr.18: 13387587-13387587 on Build GRCh38
rs964918383 Chr.18: 13387672-13387672 on Build GRCh38
rs939840883 Chr.18: 13387592-13387592 on Build GRCh38
rs374467516 Chr.18: 13387687-13387687 on Build GRCh38
rs1014103165 Chr.18: 13387554-13387554 on Build GRCh38
rs1002194877 Chr.18: 13387525-13387525 on Build GRCh38
rs921010140 Chr.18: 13387478-13387478 on Build GRCh38
rs867089629 Chr.18: 13387726-13387726 on Build GRCh38
rs933758243 Chr.18: 13387487-13387487 on Build GRCh38
rs775686974 Chr.18: 13387475-13387475 on Build GRCh38
rs113797780 Chr.18: 13387540-13387540 on Build GRCh38
rs370587696 Chr.18: 13387388-13387388 on Build GRCh38
rs967285086 Chr.18: 13387559-13387559 on Build GRCh38
rs1038295583 Chr.18: 13387512-13387512 on Build GRCh38
rs558352822 Chr.18: 13387563-13387563 on Build GRCh38
rs756905457 Chr.18: 13387728-13387728 on Build GRCh38
rs532724324 Chr.18: 13387381-13387381 on Build GRCh38
rs372160292 Chr.18: 13387574-13387574 on Build GRCh38
rs977104537 Chr.18: 13387295-13387295 on Build GRCh38
rs367734859 Chr.18: 13387694-13387694 on Build GRCh38
rs1033770126 Chr.18: 13387421-13387421 on Build GRCh38
rs898463818 Chr.18: 13387516-13387516 on Build GRCh38
rs759830764 Chr.18: 13387280-13387280 on Build GRCh38
rs975084103 Chr.18: 13387477-13387477 on Build GRCh38
rs898516185 Chr.18: 13387389-13387389 on Build GRCh38
rs367728790 Chr.18: 13387567-13387567 on Build GRCh38
rs920956972 Chr.18: 13387577-13387577 on Build GRCh38
rs1004565161 Chr.18: 13387368-13387368 on Build GRCh38
rs981797014 Chr.18: 13387527-13387527 on Build GRCh38
rs541001338 Chr.18: 13387586-13387586 on Build GRCh38
rs182912333 Chr.18: 13387545-13387545 on Build GRCh38
rs772840249 Chr.18: 13387562-13387562 on Build GRCh38
rs371083799 Chr.18: 13387680-13387680 on Build GRCh38
rs887012218 Chr.18: 13387367-13387367 on Build GRCh38
rs575423404 Chr.18: 13387629-13387629 on Build GRCh38
rs913472874 Chr.18: 13387648-13387648 on Build GRCh38
rs1002248662 Chr.18: 13387399-13387399 on Build GRCh38
rs902632245 Chr.18: 13387664-13387664 on Build GRCh38
rs966960803 Chr.18: 13387455-13387455 on Build GRCh38
rs187064537 Chr.18: 13387660-13387660 on Build GRCh38
rs78151564 Chr.18: 13387628-13387628 on Build GRCh38
rs1051408366 Chr.18: 13387362-13387362 on Build GRCh38
rs952407180 Chr.18: 13387344-13387344 on Build GRCh38
rs762212678 Chr.18: 13387572-13387572 on Build GRCh38
rs964970927 Chr.18: 13387569-13387569 on Build GRCh38
rs972560238 Chr.18: 13387309-13387309 on Build GRCh38
rs1021571375 Chr.18: 13387441-13387441 on Build GRCh38
rs992334284 Chr.18: 13387440-13387440 on Build GRCh38
rs1006878350 Chr.18: 13387476-13387476 on Build GRCh38
rs781427987 Chr.18: 13387350-13387350 on Build GRCh38
rs908331230 Chr.18: 13387591-13387591 on Build GRCh38
rs1019455802 Chr.18: 13387278-13387278 on Build GRCh38
rs764309263 Chr.18: 13387675-13387675 on Build GRCh38
rs979621580 Chr.18: 13387564-13387564 on Build GRCh38
rs143909561 Chr.18: 13387665-13387665 on Build GRCh38
rs933832541 Chr.18: 13387361-13387361 on Build GRCh38
rs372959054 Chr.18: 13387555-13387555 on Build GRCh38
rs866461324 Chr.18: 13387612-13387612 on Build GRCh38
rs899643325 Chr.18: 13387351-13387351 on Build GRCh38
rs765814801 Chr.18: 13387659-13387659 on Build GRCh38
rs928914131 Chr.18: 13387530-13387530 on Build GRCh38
rs935731016 Chr.18: 13387299-13387299 on Build GRCh38
rs774530664 Chr.18: 13387679-13387679 on Build GRCh38
rs1025443673 Chr.18: 13387490-13387490 on Build GRCh38
rs756981577 Chr.18: 13387685-13387686 on Build GRCh38
rs532733605 Chr.18: 13387667-13387667 on Build GRCh38
rs901088777 Chr.18: 13387565-13387565 on Build GRCh38
rs754870494 Chr.18: 13387688-13387688 on Build GRCh38
rs763347442 Chr.18: 13387674-13387674 on Build GRCh38
rs769727543 Chr.18: 13387402-13387402 on Build GRCh38
rs749124325 Chr.18: 13387424-13387424 on Build GRCh38
rs879645158 Chr.18: 13387425-13387425 on Build GRCh38
rs138001924 Chr.18: 13387619-13387619 on Build GRCh38
rs377305772 Chr.18: 13387666-13387666 on Build GRCh38
rs919836478 Chr.18: 13387610-13387610 on Build GRCh38
rs375595176 Chr.18: 13387589-13387589 on Build GRCh38
rs56828546 Chr.18: 13387491-13387491 on Build GRCh38
rs994061567 Chr.18: 13387566-13387566 on Build GRCh38
rs534213962 Chr.18: 13387531-13387531 on Build GRCh38
rs9957861 Chr.18: 13387627-13387627 on Build GRCh38
rs148609804 Chr.18: 13387712-13387712 on Build GRCh38
rs547823302 Chr.18: 13387705-13387705 on Build GRCh38
rs751024759 Chr.18: 13387714-13387714 on Build GRCh38
rs768661238 Chr.18: 13387467-13387467 on Build GRCh38
rs939929379 Chr.18: 13387511-13387511 on Build GRCh38
rs916229421 Chr.18: 13387400-13387400 on Build GRCh38
rs758303775 Chr.18: 13387709-13387709 on Build GRCh38
rs190626936 Chr.18: 13387273-13387273 on Build GRCh38
rs1038242796 Chr.18: 13387609-13387609 on Build GRCh38
rs565982986 Chr.18: 13387459-13387459 on Build GRCh38
rs938886569 Chr.18: 13387436-13387436 on Build GRCh38
rs1005279101 Chr.18: 13387492-13387492 on Build GRCh38

View Full Product Details