Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13438160-13438658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAD4
Entrez Gene ID
753
Gene Name
low density lipoprotein receptor class A domain containing 4
Gene Aliases
C18orf1
UniGene
Hs.731853
Gene Chromosome Location
Chr.18: 13217691-13652754 on Build GRCh38
Transcripts
NM_181481.4 NM_181482.4 XM_005258140.1 XM_006722353.1 XM_006722354.1 XM_011525737.1 XM_011525738.1 XM_017025961.1 XM_017025962.1 XM_017025963.1 XM_017025964.1 XM_017025965.1 XM_017025966.1 XM_017025967.1 XM_017025968.1 XM_017025970.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4071383 c.142C>G p.L48V Chr.18: 13438345-13438345 on Build GRCh38
73902 c.43T>A p.C15S Chr.18: 13438246-13438246 on Build GRCh38
1480230 c.99C>A p.N33K Chr.18: 13438302-13438302 on Build GRCh38
1387865 c.60C>A p.T20T Chr.18: 13438263-13438263 on Build GRCh38
4071382 c.130G>A p.E44K Chr.18: 13438333-13438333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368218936 Chr.18: 13438232-13438232 on Build GRCh38
rs756815419 Chr.18: 13438195-13438195 on Build GRCh38
rs543638427 Chr.18: 13438482-13438482 on Build GRCh38
rs771037002 Chr.18: 13438384-13438384 on Build GRCh38
rs755453642 Chr.18: 13438365-13438365 on Build GRCh38
rs889280358 Chr.18: 13438218-13438218 on Build GRCh38
rs780488637 Chr.18: 13438217-13438217 on Build GRCh38
rs200069437 Chr.18: 13438357-13438357 on Build GRCh38
rs994139167 Chr.18: 13438616-13438616 on Build GRCh38
rs898055539 Chr.18: 13438591-13438591 on Build GRCh38
rs760095311 Chr.18: 13438518-13438518 on Build GRCh38
rs748652072 Chr.18: 13438248-13438248 on Build GRCh38
rs760991562 Chr.18: 13438275-13438275 on Build GRCh38
rs1037881168 Chr.18: 13438187-13438187 on Build GRCh38
rs749605324 Chr.18: 13438219-13438219 on Build GRCh38
rs762801340 Chr.18: 13438284-13438284 on Build GRCh38
rs922406164 Chr.18: 13438460-13438460 on Build GRCh38
rs763860704 Chr.18: 13438289-13438289 on Build GRCh38
rs773138501 Chr.18: 13438271-13438271 on Build GRCh38
rs760171029 Chr.18: 13438415-13438415 on Build GRCh38
rs1007922 Chr.18: 13438544-13438544 on Build GRCh38
rs747994309 Chr.18: 13438396-13438396 on Build GRCh38
rs775279607 Chr.18: 13438280-13438280 on Build GRCh38
rs201120117 Chr.18: 13438392-13438392 on Build GRCh38
rs774092011 Chr.18: 13438398-13438398 on Build GRCh38
rs12232740 Chr.18: 13438632-13438632 on Build GRCh38
rs915214935 Chr.18: 13438512-13438512 on Build GRCh38
rs767902053 Chr.18: 13438190-13438190 on Build GRCh38
rs750130232 Chr.18: 13438202-13438202 on Build GRCh38
rs190877475 Chr.18: 13438325-13438325 on Build GRCh38
rs780541408 Chr.18: 13438198-13438198 on Build GRCh38
rs145523399 Chr.18: 13438332-13438332 on Build GRCh38
rs566414256 Chr.18: 13438560-13438560 on Build GRCh38
rs895942652 Chr.18: 13438579-13438579 on Build GRCh38
rs976564744 Chr.18: 13438448-13438448 on Build GRCh38
rs755802486 Chr.18: 13438356-13438356 on Build GRCh38
rs375478517 Chr.18: 13438203-13438203 on Build GRCh38
rs1040580164 Chr.18: 13438586-13438586 on Build GRCh38
rs1023373949 Chr.18: 13438422-13438422 on Build GRCh38
rs767297877 Chr.18: 13438409-13438409 on Build GRCh38
rs944824757 Chr.18: 13438584-13438584 on Build GRCh38
rs988134635 Chr.18: 13438473-13438473 on Build GRCh38
rs372796452 Chr.18: 13438299-13438299 on Build GRCh38
rs910642703 Chr.18: 13438572-13438572 on Build GRCh38
rs765901511 Chr.18: 13438433-13438433 on Build GRCh38
rs946734827 Chr.18: 13438526-13438526 on Build GRCh38
rs772243127 Chr.18: 13438260-13438260 on Build GRCh38
rs778415939 Chr.18: 13438335-13438336 on Build GRCh38
rs200627527 Chr.18: 13438397-13438397 on Build GRCh38
rs927869515 Chr.18: 13438461-13438461 on Build GRCh38
rs138826258 Chr.18: 13438360-13438360 on Build GRCh38
rs982906547 Chr.18: 13438493-13438493 on Build GRCh38
rs747005823 Chr.18: 13438371-13438371 on Build GRCh38
rs768543443 Chr.18: 13438306-13438306 on Build GRCh38
rs748709104 Chr.18: 13438369-13438369 on Build GRCh38
rs963902685 Chr.18: 13438438-13438438 on Build GRCh38
rs202231144 Chr.18: 13438370-13438370 on Build GRCh38
rs771564175 Chr.18: 13438435-13438435 on Build GRCh38
rs1025751980 Chr.18: 13438394-13438394 on Build GRCh38
rs779632232 Chr.18: 13438234-13438234 on Build GRCh38
rs771012364 Chr.18: 13438279-13438279 on Build GRCh38
rs16940538 Chr.18: 13438471-13438471 on Build GRCh38
rs753594371 Chr.18: 13438434-13438434 on Build GRCh38
rs183074583 Chr.18: 13438410-13438410 on Build GRCh38
rs898081484 Chr.18: 13438338-13438338 on Build GRCh38
rs749852696 Chr.18: 13438349-13438349 on Build GRCh38
rs761831578 Chr.18: 13438406-13438406 on Build GRCh38
rs909554413 Chr.18: 13438514-13438514 on Build GRCh38

View Full Product Details