Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13666240-13666752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM210A
Entrez Gene ID
125228
Gene Name
family with sequence similarity 210 member A
Gene Aliases
C18orf19, HsT2329
UniGene
Hs.13034
Gene Chromosome Location
Chr.18: 13663347-13726592 on Build GRCh38
Transcripts
NM_001098801.1 NM_152352.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
986748 c.702C>A p.D234E Chr.18: 13666597-13666597 on Build GRCh38
4854238 c.733G>A p.E245K Chr.18: 13666566-13666566 on Build GRCh38
986749 c.635C>G p.T212S Chr.18: 13666664-13666664 on Build GRCh38
377522 c.589G>T p.A197S Chr.18: 13666710-13666710 on Build GRCh38
4640148 c.738A>G p.K246K Chr.18: 13666561-13666561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773819952 Chr.18: 13666469-13666469 on Build GRCh38
rs757945137 Chr.18: 13666656-13666656 on Build GRCh38
rs765493182 Chr.18: 13666514-13666514 on Build GRCh38
rs967763800 Chr.18: 13666439-13666439 on Build GRCh38
rs1002936232 Chr.18: 13666310-13666310 on Build GRCh38
rs377046472 Chr.18: 13666413-13666413 on Build GRCh38
rs371575474 Chr.18: 13666530-13666530 on Build GRCh38
rs748991444 Chr.18: 13666448-13666448 on Build GRCh38
rs761744797 Chr.18: 13666528-13666528 on Build GRCh38
rs767364628 Chr.18: 13666635-13666635 on Build GRCh38
rs778201852 Chr.18: 13666658-13666658 on Build GRCh38
rs746502672 Chr.18: 13666607-13666608 on Build GRCh38
rs750055447 Chr.18: 13666453-13666454 on Build GRCh38
rs1053284335 Chr.18: 13666311-13666311 on Build GRCh38
rs928581359 Chr.18: 13666703-13666703 on Build GRCh38
rs779628499 Chr.18: 13666543-13666543 on Build GRCh38
rs775892022 Chr.18: 13666628-13666628 on Build GRCh38
rs753277415 Chr.18: 13666549-13666549 on Build GRCh38
rs761057336 Chr.18: 13666473-13666473 on Build GRCh38
rs200589135 Chr.18: 13666518-13666518 on Build GRCh38
rs777961735 Chr.18: 13666611-13666611 on Build GRCh38
rs1044516199 Chr.18: 13666294-13666294 on Build GRCh38
rs529431654 Chr.18: 13666690-13666690 on Build GRCh38
rs777894329 Chr.18: 13666459-13666460 on Build GRCh38
rs770018973 Chr.18: 13666694-13666694 on Build GRCh38
rs377031591 Chr.18: 13666501-13666501 on Build GRCh38
rs924336663 Chr.18: 13666353-13666353 on Build GRCh38
rs771076117 Chr.18: 13666478-13666478 on Build GRCh38
rs747275779 Chr.18: 13666680-13666680 on Build GRCh38
rs755685983 Chr.18: 13666438-13666438 on Build GRCh38
rs562311629 Chr.18: 13666617-13666617 on Build GRCh38
rs750302261 Chr.18: 13666433-13666433 on Build GRCh38
rs959542995 Chr.18: 13666609-13666609 on Build GRCh38
rs750403829 Chr.18: 13666531-13666531 on Build GRCh38
rs118166978 Chr.18: 13666329-13666329 on Build GRCh38
rs1054215584 Chr.18: 13666373-13666373 on Build GRCh38
rs949715012 Chr.18: 13666313-13666313 on Build GRCh38
rs35023957 Chr.18: 13666603-13666603 on Build GRCh38
rs907300889 Chr.18: 13666307-13666307 on Build GRCh38
rs763224814 Chr.18: 13666634-13666634 on Build GRCh38
rs764696749 Chr.18: 13666649-13666649 on Build GRCh38
rs773350410 Chr.18: 13666624-13666624 on Build GRCh38
rs1050996680 Chr.18: 13666416-13666416 on Build GRCh38
rs201929526 Chr.18: 13666456-13666456 on Build GRCh38
rs757624460 Chr.18: 13666689-13666689 on Build GRCh38
rs779389463 Chr.18: 13666444-13666444 on Build GRCh38
rs997103651 Chr.18: 13666382-13666382 on Build GRCh38
rs753761934 Chr.18: 13666637-13666637 on Build GRCh38
rs778595283 Chr.18: 13666591-13666591 on Build GRCh38
rs756994890 Chr.18: 13666448-13666449 on Build GRCh38
rs138091324 Chr.18: 13666595-13666595 on Build GRCh38
rs753744216 Chr.18: 13666447-13666448 on Build GRCh38
rs768238962 Chr.18: 13666451-13666451 on Build GRCh38
rs1022292114 Chr.18: 13666418-13666418 on Build GRCh38
rs775673770 Chr.18: 13666696-13666696 on Build GRCh38
rs374459586 Chr.18: 13666704-13666704 on Build GRCh38
rs754480188 Chr.18: 13666561-13666561 on Build GRCh38
rs760751853 Chr.18: 13666673-13666673 on Build GRCh38
rs760664524 Chr.18: 13666636-13666636 on Build GRCh38
rs149627810 Chr.18: 13666639-13666639 on Build GRCh38
rs576279471 Chr.18: 13666627-13666627 on Build GRCh38
rs59211767 Chr.18: 13666618-13666618 on Build GRCh38
rs572582049 Chr.18: 13666548-13666548 on Build GRCh38
rs539258590 Chr.18: 13666414-13666414 on Build GRCh38
rs748579173 Chr.18: 13666457-13666457 on Build GRCh38
rs752367715 Chr.18: 13666650-13666650 on Build GRCh38
rs779582741 Chr.18: 13666546-13666547 on Build GRCh38
rs778698722 Chr.18: 13666452-13666453 on Build GRCh38
rs755954474 Chr.18: 13666535-13666535 on Build GRCh38
rs772413284 Chr.18: 13666467-13666467 on Build GRCh38
rs571775136 Chr.18: 13666381-13666381 on Build GRCh38
rs965784338 Chr.18: 13666539-13666539 on Build GRCh38
rs774588845 Chr.18: 13666367-13666367 on Build GRCh38

View Full Product Details