Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13681606-13682118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM210A
Entrez Gene ID
125228
Gene Name
family with sequence similarity 210 member A
Gene Aliases
C18orf19, HsT2329
UniGene
Hs.13034
Gene Chromosome Location
Chr.18: 13663347-13726592 on Build GRCh38
Transcripts
NM_001098801.1 NM_152352.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
374264 c.384G>A p.K128K Chr.18: 13681694-13681694 on Build GRCh38
359171 c.333G>T p.E111D Chr.18: 13681745-13681745 on Build GRCh38
5812604 c.303A>T p.T101T Chr.18: 13681775-13681775 on Build GRCh38
5853940 c.392G>A p.R131K Chr.18: 13681686-13681686 on Build GRCh38
1387871 c.419T>C p.V140A Chr.18: 13681659-13681659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758549366 Chr.18: 13682030-13682030 on Build GRCh38
rs145075680 Chr.18: 13681877-13681877 on Build GRCh38
rs752763602 Chr.18: 13681752-13681752 on Build GRCh38
rs113181833 Chr.18: 13681744-13681744 on Build GRCh38
rs764961799 Chr.18: 13681751-13681752 on Build GRCh38
rs764167668 Chr.18: 13681918-13681918 on Build GRCh38
rs750097013 Chr.18: 13681722-13681722 on Build GRCh38
rs904849603 Chr.18: 13682089-13682089 on Build GRCh38
rs780936180 Chr.18: 13681999-13681999 on Build GRCh38
rs143865357 Chr.18: 13681900-13681900 on Build GRCh38
rs759972433 Chr.18: 13681844-13681844 on Build GRCh38
rs779671314 Chr.18: 13682065-13682065 on Build GRCh38
rs373098318 Chr.18: 13681812-13681812 on Build GRCh38
rs373346025 Chr.18: 13681848-13681848 on Build GRCh38
rs762179618 Chr.18: 13682084-13682084 on Build GRCh38
rs758038817 Chr.18: 13681789-13681789 on Build GRCh38
rs774888776 Chr.18: 13682079-13682079 on Build GRCh38
rs769622172 Chr.18: 13681774-13681774 on Build GRCh38
rs777433452 Chr.18: 13681981-13681981 on Build GRCh38
rs767475426 Chr.18: 13682021-13682021 on Build GRCh38
rs774144774 Chr.18: 13681818-13681818 on Build GRCh38
rs745483549 Chr.18: 13682002-13682002 on Build GRCh38
rs572460629 Chr.18: 13682051-13682051 on Build GRCh38
rs745718079 Chr.18: 13681643-13681643 on Build GRCh38
rs375228347 Chr.18: 13682027-13682027 on Build GRCh38
rs771730636 Chr.18: 13681897-13681897 on Build GRCh38
rs113115642 Chr.18: 13681990-13681990 on Build GRCh38
rs34798758 Chr.18: 13681751-13681751 on Build GRCh38
rs771679397 Chr.18: 13681831-13681831 on Build GRCh38
rs35493157 Chr.18: 13681963-13681963 on Build GRCh38
rs745561931 Chr.18: 13682058-13682058 on Build GRCh38
rs986155289 Chr.18: 13681917-13681917 on Build GRCh38
rs755471515 Chr.18: 13681746-13681746 on Build GRCh38
rs747729503 Chr.18: 13681826-13681826 on Build GRCh38
rs1049672748 Chr.18: 13682048-13682048 on Build GRCh38
rs752482708 Chr.18: 13682041-13682041 on Build GRCh38
rs552403778 Chr.18: 13682012-13682012 on Build GRCh38
rs766683697 Chr.18: 13681834-13681834 on Build GRCh38
rs116967198 Chr.18: 13681833-13681833 on Build GRCh38
rs769212391 Chr.18: 13682010-13682010 on Build GRCh38
rs200885719 Chr.18: 13681884-13681884 on Build GRCh38
rs147538636 Chr.18: 13681760-13681760 on Build GRCh38
rs144093105 Chr.18: 13681956-13681956 on Build GRCh38
rs776132977 Chr.18: 13681910-13681910 on Build GRCh38
rs775768207 Chr.18: 13682004-13682004 on Build GRCh38
rs932727900 Chr.18: 13681969-13681969 on Build GRCh38
rs548721066 Chr.18: 13681912-13681912 on Build GRCh38
rs779862422 Chr.18: 13681775-13681775 on Build GRCh38
rs770213503 Chr.18: 13682003-13682003 on Build GRCh38
rs368496533 Chr.18: 13681688-13681688 on Build GRCh38
rs758510315 Chr.18: 13681855-13681855 on Build GRCh38
rs140347489 Chr.18: 13681916-13681916 on Build GRCh38
rs968087505 Chr.18: 13681691-13681691 on Build GRCh38
rs770557083 Chr.18: 13681908-13681908 on Build GRCh38
rs767849715 Chr.18: 13681936-13681936 on Build GRCh38
rs552796973 Chr.18: 13682038-13682038 on Build GRCh38
rs371076318 Chr.18: 13682059-13682059 on Build GRCh38
rs377543711 Chr.18: 13681875-13681875 on Build GRCh38
rs201619883 Chr.18: 13681952-13681952 on Build GRCh38
rs756838446 Chr.18: 13682056-13682056 on Build GRCh38
rs768085305 Chr.18: 13681685-13681685 on Build GRCh38
rs748077437 Chr.18: 13681896-13681896 on Build GRCh38
rs763468795 Chr.18: 13682005-13682005 on Build GRCh38
rs779670880 Chr.18: 13682036-13682037 on Build GRCh38
rs910638126 Chr.18: 13681720-13681720 on Build GRCh38
rs778033376 Chr.18: 13681761-13681761 on Build GRCh38
rs753137291 Chr.18: 13681852-13681852 on Build GRCh38
rs767337769 Chr.18: 13681716-13681716 on Build GRCh38
rs760768896 Chr.18: 13682037-13682037 on Build GRCh38
rs150631358 Chr.18: 13681724-13681724 on Build GRCh38
rs772224892 Chr.18: 13682086-13682086 on Build GRCh38
rs929569552 Chr.18: 13681781-13681781 on Build GRCh38
rs775325514 Chr.18: 13681664-13681664 on Build GRCh38
rs375912922 Chr.18: 13681729-13681729 on Build GRCh38
rs987998599 Chr.18: 13681651-13681651 on Build GRCh38
rs769569086 Chr.18: 13681647-13681647 on Build GRCh38
rs200198941 Chr.18: 13682046-13682046 on Build GRCh38
rs369772010 Chr.18: 13681842-13681842 on Build GRCh38
rs749722333 Chr.18: 13682072-13682072 on Build GRCh38
rs779729816 Chr.18: 13681876-13681876 on Build GRCh38
rs189782979 Chr.18: 13681915-13681915 on Build GRCh38
rs768981281 Chr.18: 13682074-13682074 on Build GRCh38
rs751584591 Chr.18: 13681922-13681922 on Build GRCh38
rs140179532 Chr.18: 13681788-13681788 on Build GRCh38
rs757273055 Chr.18: 13681930-13681930 on Build GRCh38
rs758344817 Chr.18: 13681967-13681967 on Build GRCh38
rs201878744 Chr.18: 13681867-13681867 on Build GRCh38
rs201164603 Chr.18: 13681700-13681700 on Build GRCh38
rs151157304 Chr.18: 13681911-13681911 on Build GRCh38
rs181249004 Chr.18: 13682093-13682093 on Build GRCh38
rs768583559 Chr.18: 13681813-13681813 on Build GRCh38
rs878933292 Chr.18: 13681929-13681929 on Build GRCh38
rs754852546 Chr.18: 13681959-13681959 on Build GRCh38
rs148603152 Chr.18: 13681902-13681902 on Build GRCh38
rs770339833 Chr.18: 13681998-13681998 on Build GRCh38
rs754726735 Chr.18: 13681880-13681880 on Build GRCh38
rs577321430 Chr.18: 13681868-13681868 on Build GRCh38
rs199631091 Chr.18: 13681764-13681764 on Build GRCh38
rs142125636 Chr.18: 13681904-13681904 on Build GRCh38
rs765484785 Chr.18: 13681913-13681913 on Build GRCh38
rs762902773 Chr.18: 13681680-13681680 on Build GRCh38
rs1049307337 Chr.18: 13681787-13681787 on Build GRCh38
rs915255851 Chr.18: 13681662-13681662 on Build GRCh38
rs761995037 Chr.18: 13682019-13682019 on Build GRCh38
rs750189472 Chr.18: 13681788-13681788 on Build GRCh38
rs746574984 Chr.18: 13681995-13681995 on Build GRCh38

View Full Product Details