Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 21664034-21664540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD3
Entrez Gene ID
171586
Gene Name
abhydrolase domain containing 3
Gene Aliases
LABH3
UniGene
Hs.397978
Gene Chromosome Location
Chr.18: 21650897-21704805 on Build GRCh38
Transcripts
NM_001308256.1 NM_001308257.1 NM_138340.4 XM_011525847.2 XM_017025571.1 XM_017025572.1 XM_017025573.1 XM_017025574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
40803 c.556-4C>T p.? Chr.18: 21664234-21664234 on Build GRCh38
7353717 c.616C>A p.H206N Chr.18: 21664170-21664170 on Build GRCh38
708341 c.608A>T p.H203L Chr.18: 21664178-21664178 on Build GRCh38
5853958 c.664G>A p.G222R Chr.18: 21664122-21664122 on Build GRCh38
6465327 c.627C>T p.Y209Y Chr.18: 21664159-21664159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765214305 Chr.18: 21664174-21664174 on Build GRCh38
rs866121696 Chr.18: 21664173-21664173 on Build GRCh38
rs371887064 Chr.18: 21664091-21664091 on Build GRCh38
rs970582334 Chr.18: 21664085-21664085 on Build GRCh38
rs113792655 Chr.18: 21664517-21664517 on Build GRCh38
rs989562672 Chr.18: 21664200-21664200 on Build GRCh38
rs776301243 Chr.18: 21664281-21664281 on Build GRCh38
rs1003656149 Chr.18: 21664485-21664485 on Build GRCh38
rs759309957 Chr.18: 21664282-21664282 on Build GRCh38
rs369673843 Chr.18: 21664187-21664187 on Build GRCh38
rs564896151 Chr.18: 21664409-21664409 on Build GRCh38
rs560882202 Chr.18: 21664477-21664477 on Build GRCh38
rs756580657 Chr.18: 21664238-21664238 on Build GRCh38
rs375511804 Chr.18: 21664132-21664132 on Build GRCh38
rs188645707 Chr.18: 21664358-21664358 on Build GRCh38
rs780603209 Chr.18: 21664241-21664241 on Build GRCh38
rs527503115 Chr.18: 21664417-21664417 on Build GRCh38
rs760512536 Chr.18: 21664168-21664168 on Build GRCh38
rs563766435 Chr.18: 21664287-21664287 on Build GRCh38
rs764241281 Chr.18: 21664077-21664077 on Build GRCh38
rs763118290 Chr.18: 21664069-21664069 on Build GRCh38
rs765165491 Chr.18: 21664195-21664195 on Build GRCh38
rs770299120 Chr.18: 21664147-21664147 on Build GRCh38
rs746197455 Chr.18: 21664229-21664229 on Build GRCh38
rs774539067 Chr.18: 21664166-21664166 on Build GRCh38
rs200769908 Chr.18: 21664175-21664175 on Build GRCh38
rs543873722 Chr.18: 21664246-21664246 on Build GRCh38
rs546285547 Chr.18: 21664372-21664372 on Build GRCh38
rs1005433818 Chr.18: 21664075-21664075 on Build GRCh38
rs941258780 Chr.18: 21664431-21664431 on Build GRCh38
rs944727523 Chr.18: 21664355-21664355 on Build GRCh38
rs749321117 Chr.18: 21664159-21664159 on Build GRCh38
rs776445992 Chr.18: 21664184-21664184 on Build GRCh38
rs553885195 Chr.18: 21664475-21664475 on Build GRCh38
rs753696806 Chr.18: 21664113-21664113 on Build GRCh38
rs956373983 Chr.18: 21664186-21664186 on Build GRCh38
rs751238154 Chr.18: 21664226-21664226 on Build GRCh38
rs888493186 Chr.18: 21664447-21664447 on Build GRCh38
rs1048878659 Chr.18: 21664436-21664436 on Build GRCh38
rs377531480 Chr.18: 21664144-21664144 on Build GRCh38
rs750317676 Chr.18: 21664210-21664212 on Build GRCh38
rs768695536 Chr.18: 21664264-21664264 on Build GRCh38
rs752795385 Chr.18: 21664105-21664105 on Build GRCh38
rs888292903 Chr.18: 21664404-21664404 on Build GRCh38
rs374203565 Chr.18: 21664270-21664270 on Build GRCh38
rs1023470214 Chr.18: 21664063-21664063 on Build GRCh38
rs766130161 Chr.18: 21664170-21664170 on Build GRCh38
rs574865405 Chr.18: 21664227-21664227 on Build GRCh38
rs574604166 Chr.18: 21664245-21664245 on Build GRCh38
rs904857772 Chr.18: 21664453-21664453 on Build GRCh38
rs761711494 Chr.18: 21664148-21664148 on Build GRCh38
rs376883720 Chr.18: 21664165-21664165 on Build GRCh38
rs748399535 Chr.18: 21664272-21664272 on Build GRCh38
rs754555865 Chr.18: 21664118-21664118 on Build GRCh38
rs919369587 Chr.18: 21664391-21664391 on Build GRCh38
rs757978890 Chr.18: 21664221-21664221 on Build GRCh38
rs111917449 Chr.18: 21664392-21664392 on Build GRCh38
rs988755625 Chr.18: 21664194-21664194 on Build GRCh38
rs547265712 Chr.18: 21664427-21664427 on Build GRCh38
rs747445817 Chr.18: 21664139-21664139 on Build GRCh38
rs758868400 Chr.18: 21664511-21664511 on Build GRCh38
rs775456524 Chr.18: 21664149-21664149 on Build GRCh38
rs752199897 Chr.18: 21664125-21664125 on Build GRCh38
rs947626999 Chr.18: 21664214-21664214 on Build GRCh38
rs911378498 Chr.18: 21664231-21664231 on Build GRCh38
rs1050002696 Chr.18: 21664230-21664230 on Build GRCh38
rs757682905 Chr.18: 21664141-21664141 on Build GRCh38
rs572056143 Chr.18: 21664499-21664499 on Build GRCh38
rs781443913 Chr.18: 21664228-21664228 on Build GRCh38
rs772278628 Chr.18: 21664116-21664116 on Build GRCh38
rs1044987043 Chr.18: 21664440-21664440 on Build GRCh38
rs34831491 Chr.18: 21664201-21664201 on Build GRCh38
rs1047302746 Chr.18: 21664403-21664403 on Build GRCh38
rs777502367 Chr.18: 21664134-21664134 on Build GRCh38

View Full Product Details