Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 23519048-23519596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf8
Entrez Gene ID
29919
Gene Name
chromosome 18 open reading frame 8
Gene Aliases
HsT2591, MIC1, Mic-1
UniGene
Hs.464779
Gene Chromosome Location
Chr.18: 23503470-23531807 on Build GRCh38
Transcripts
NM_001276342.1 NM_001318707.1 NM_001318708.1 NM_001318709.1 NM_013326.4 XM_011525939.1 XM_017025717.1 XM_005258277.1 XM_005258279.2 XM_011526015.1

Gene Symbol
NPC1
Entrez Gene ID
4864
Gene Name
NPC intracellular cholesterol transporter 1
Gene Aliases
NPC
UniGene
Hs.464779
Gene Chromosome Location
Chr.18: 23506184-23586617 on Build GRCh38
Transcripts
NM_001276342.1 NM_001318707.1 NM_001318708.1 NM_001318709.1 NM_013326.4 XM_011525939.1 XM_017025717.1 XM_005258277.1 XM_005258279.2 XM_011526015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
987075 c.809T>C p.V270A Chr.18: 23519134-23519134 on Build GRCh38
987074 c.774G>C p.L258F Chr.18: 23519099-23519099 on Build GRCh38
6839622 c.847G>C p.E283Q Chr.18: 23519172-23519172 on Build GRCh38
987073 c.744-1G>C p.? Chr.18: 23519068-23519068 on Build GRCh38
6326226 c.828C>T p.V276V Chr.18: 23519153-23519153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000197680 Chr.18: 23519473-23519473 on Build GRCh38
rs372391248 Chr.18: 23519106-23519106 on Build GRCh38
rs774197072 Chr.18: 23519102-23519102 on Build GRCh38
rs780086506 Chr.18: 23519190-23519190 on Build GRCh38
rs558821240 Chr.18: 23519327-23519327 on Build GRCh38
rs746156517 Chr.18: 23519202-23519203 on Build GRCh38
rs758542214 Chr.18: 23519204-23519205 on Build GRCh38
rs1036249921 Chr.18: 23519391-23519391 on Build GRCh38
rs753119699 Chr.18: 23519138-23519138 on Build GRCh38
rs745865912 Chr.18: 23519188-23519188 on Build GRCh38
rs940607685 Chr.18: 23519384-23519384 on Build GRCh38
rs561102299 Chr.18: 23519365-23519365 on Build GRCh38
rs545767332 Chr.18: 23519497-23519497 on Build GRCh38
rs867666714 Chr.18: 23519125-23519125 on Build GRCh38
rs757383487 Chr.18: 23519154-23519154 on Build GRCh38
rs970885416 Chr.18: 23519451-23519451 on Build GRCh38
rs187677149 Chr.18: 23519151-23519151 on Build GRCh38
rs989425095 Chr.18: 23519520-23519520 on Build GRCh38
rs546959665 Chr.18: 23519304-23519304 on Build GRCh38
rs541251395 Chr.18: 23519351-23519351 on Build GRCh38
rs781241792 Chr.18: 23519160-23519160 on Build GRCh38
rs1008371415 Chr.18: 23519315-23519315 on Build GRCh38
rs929850913 Chr.18: 23519518-23519518 on Build GRCh38
rs1038685108 Chr.18: 23519256-23519256 on Build GRCh38
rs1029069574 Chr.18: 23519525-23519525 on Build GRCh38
rs193211425 Chr.18: 23519378-23519378 on Build GRCh38
rs956859465 Chr.18: 23519147-23519147 on Build GRCh38
rs898454216 Chr.18: 23519263-23519263 on Build GRCh38
rs767588971 Chr.18: 23519108-23519108 on Build GRCh38
rs958001134 Chr.18: 23519348-23519348 on Build GRCh38
rs757170764 Chr.18: 23519281-23519281 on Build GRCh38
rs953462156 Chr.18: 23519527-23519527 on Build GRCh38
rs1039851278 Chr.18: 23519532-23519532 on Build GRCh38
rs987536260 Chr.18: 23519534-23519534 on Build GRCh38
rs576407071 Chr.18: 23519245-23519245 on Build GRCh38
rs71373347 Chr.18: 23519237-23519238 on Build GRCh38
rs956912730 Chr.18: 23519382-23519382 on Build GRCh38
rs758649987 Chr.18: 23519144-23519144 on Build GRCh38
rs994598278 Chr.18: 23519211-23519211 on Build GRCh38
rs911854634 Chr.18: 23519379-23519379 on Build GRCh38
rs751873745 Chr.18: 23519413-23519413 on Build GRCh38
rs1000569329 Chr.18: 23519361-23519361 on Build GRCh38
rs1045166831 Chr.18: 23519239-23519239 on Build GRCh38
rs1047033185 Chr.18: 23519520-23519523 on Build GRCh38
rs114598708 Chr.18: 23519515-23519515 on Build GRCh38
rs757657866 Chr.18: 23519182-23519185 on Build GRCh38
rs779121093 Chr.18: 23519198-23519199 on Build GRCh38
rs1025259153 Chr.18: 23519277-23519277 on Build GRCh38
rs754656257 Chr.18: 23519127-23519127 on Build GRCh38
rs1012403761 Chr.18: 23519246-23519246 on Build GRCh38
rs911724699 Chr.18: 23519565-23519565 on Build GRCh38
rs534549693 Chr.18: 23519091-23519091 on Build GRCh38
rs532036425 Chr.18: 23519475-23519475 on Build GRCh38
rs202070090 Chr.18: 23519113-23519113 on Build GRCh38
rs765968224 Chr.18: 23519117-23519117 on Build GRCh38
rs994089589 Chr.18: 23519280-23519280 on Build GRCh38
rs1031228391 Chr.18: 23519489-23519489 on Build GRCh38
rs764730840 Chr.18: 23519133-23519133 on Build GRCh38
rs1055577425 Chr.18: 23519409-23519409 on Build GRCh38
rs777477860 Chr.18: 23519200-23519200 on Build GRCh38
rs894221586 Chr.18: 23519428-23519428 on Build GRCh38
rs1001023600 Chr.18: 23519535-23519535 on Build GRCh38
rs562603731 Chr.18: 23519329-23519329 on Build GRCh38
rs35759590 Chr.18: 23519520-23519520 on Build GRCh38
rs151253134 Chr.18: 23519120-23519120 on Build GRCh38
rs748010525 Chr.18: 23519208-23519208 on Build GRCh38
rs964038505 Chr.18: 23519336-23519340 on Build GRCh38
rs764268307 Chr.18: 23519375-23519375 on Build GRCh38
rs760305045 Chr.18: 23519114-23519114 on Build GRCh38
rs746086714 Chr.18: 23519086-23519086 on Build GRCh38
rs754302429 Chr.18: 23519100-23519106 on Build GRCh38
rs770064244 Chr.18: 23519189-23519189 on Build GRCh38
rs994482142 Chr.18: 23519407-23519407 on Build GRCh38
rs921999751 Chr.18: 23519371-23519371 on Build GRCh38
rs112508588 Chr.18: 23519289-23519289 on Build GRCh38
rs567107574 Chr.18: 23519251-23519251 on Build GRCh38
rs898846265 Chr.18: 23519404-23519404 on Build GRCh38
rs557283866 Chr.18: 23519243-23519243 on Build GRCh38
rs550875313 Chr.18: 23519153-23519153 on Build GRCh38
rs530033292 Chr.18: 23519450-23519450 on Build GRCh38
rs9967282 Chr.18: 23519443-23519443 on Build GRCh38
rs1018330418 Chr.18: 23519094-23519094 on Build GRCh38

View Full Product Details