Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 24143106-24143512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABYR
Entrez Gene ID
26256
Gene Name
calcium binding tyrosine phosphorylation regulated
Gene Aliases
CABYRa, CABYRc, CABYRc/d, CABYRe, CBP86, CT88, FSP-2, FSP2
UniGene
Hs.511983
Gene Chromosome Location
Chr.18: 24138956-24161600 on Build GRCh38
Transcripts
NM_001308231.1 NM_012189.3 NM_138643.2 NM_138644.2 NM_153768.2 NM_153769.2 NM_153770.2 XM_005258247.2 XM_017025706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6465346 c.46A>G p.T16A Chr.18: 24143160-24143160 on Build GRCh38
6200819 c.58G>A p.G20R Chr.18: 24143172-24143172 on Build GRCh38
6349706 c.78C>T p.L26L Chr.18: 24143192-24143192 on Build GRCh38
6232286 c.96C>A p.N32K Chr.18: 24143210-24143210 on Build GRCh38
6232285 c.96C>A p.N32K Chr.18: 24143210-24143210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754680737 Chr.18: 24143358-24143358 on Build GRCh38
rs778246347 Chr.18: 24143363-24143363 on Build GRCh38
rs1051076089 Chr.18: 24143330-24143330 on Build GRCh38
rs372627357 Chr.18: 24143446-24143446 on Build GRCh38
rs762740534 Chr.18: 24143449-24143451 on Build GRCh38
rs188072225 Chr.18: 24143134-24143134 on Build GRCh38
rs747350083 Chr.18: 24143271-24143271 on Build GRCh38
rs199535292 Chr.18: 24143337-24143337 on Build GRCh38
rs779796982 Chr.18: 24143133-24143133 on Build GRCh38
rs778498292 Chr.18: 24143231-24143231 on Build GRCh38
rs561360855 Chr.18: 24143325-24143325 on Build GRCh38
rs868264529 Chr.18: 24143383-24143383 on Build GRCh38
rs771382555 Chr.18: 24143276-24143276 on Build GRCh38
rs140250271 Chr.18: 24143225-24143225 on Build GRCh38
rs150516148 Chr.18: 24143204-24143204 on Build GRCh38
rs200976954 Chr.18: 24143137-24143137 on Build GRCh38
rs759518813 Chr.18: 24143185-24143185 on Build GRCh38
rs767540119 Chr.18: 24143327-24143327 on Build GRCh38
rs776653099 Chr.18: 24143278-24143278 on Build GRCh38
rs896757178 Chr.18: 24143165-24143165 on Build GRCh38
rs771107571 Chr.18: 24143172-24143172 on Build GRCh38
rs141984542 Chr.18: 24143192-24143192 on Build GRCh38
rs771145148 Chr.18: 24143395-24143395 on Build GRCh38
rs756022680 Chr.18: 24143213-24143213 on Build GRCh38
rs752897110 Chr.18: 24143443-24143444 on Build GRCh38
rs369752863 Chr.18: 24143429-24143429 on Build GRCh38
rs529839297 Chr.18: 24143371-24143371 on Build GRCh38
rs906905043 Chr.18: 24143461-24143461 on Build GRCh38
rs202110713 Chr.18: 24143162-24143162 on Build GRCh38
rs376839136 Chr.18: 24143347-24143347 on Build GRCh38
rs778288864 Chr.18: 24143266-24143266 on Build GRCh38
rs200524238 Chr.18: 24143326-24143326 on Build GRCh38
rs779561578 Chr.18: 24143219-24143219 on Build GRCh38
rs751398808 Chr.18: 24143189-24143189 on Build GRCh38
rs760841937 Chr.18: 24143169-24143169 on Build GRCh38
rs759611458 Chr.18: 24143219-24143221 on Build GRCh38
rs759820327 Chr.18: 24143283-24143283 on Build GRCh38
rs904873967 Chr.18: 24143203-24143203 on Build GRCh38
rs990468143 Chr.18: 24143197-24143197 on Build GRCh38
rs201103485 Chr.18: 24143341-24143341 on Build GRCh38
rs144855105 Chr.18: 24143141-24143141 on Build GRCh38
rs912584704 Chr.18: 24143333-24143333 on Build GRCh38
rs200672743 Chr.18: 24143355-24143355 on Build GRCh38
rs753893248 Chr.18: 24143140-24143141 on Build GRCh38
rs755863027 Chr.18: 24143130-24143130 on Build GRCh38
rs776957335 Chr.18: 24143177-24143177 on Build GRCh38
rs144317760 Chr.18: 24143230-24143230 on Build GRCh38
rs781639836 Chr.18: 24143424-24143424 on Build GRCh38
rs176090 Chr.18: 24143168-24143168 on Build GRCh38
rs774713937 Chr.18: 24143142-24143142 on Build GRCh38
rs11552048 Chr.18: 24143217-24143217 on Build GRCh38
rs765365778 Chr.18: 24143186-24143186 on Build GRCh38
rs748656978 Chr.18: 24143145-24143145 on Build GRCh38
rs374894551 Chr.18: 24143258-24143258 on Build GRCh38
rs940074977 Chr.18: 24143342-24143342 on Build GRCh38
rs752167605 Chr.18: 24143366-24143366 on Build GRCh38
rs1023909145 Chr.18: 24143449-24143449 on Build GRCh38
rs895372794 Chr.18: 24143433-24143433 on Build GRCh38
rs552158197 Chr.18: 24143344-24143344 on Build GRCh38
rs750213238 Chr.18: 24143211-24143211 on Build GRCh38
rs775904436 Chr.18: 24143450-24143450 on Build GRCh38
rs769716005 Chr.18: 24143307-24143307 on Build GRCh38
rs749366392 Chr.18: 24143454-24143454 on Build GRCh38
rs765362589 Chr.18: 24143282-24143284 on Build GRCh38
rs757949748 Chr.18: 24143373-24143373 on Build GRCh38
rs778237472 Chr.18: 24143392-24143392 on Build GRCh38
rs931459628 Chr.18: 24143413-24143413 on Build GRCh38
rs747579674 Chr.18: 24143394-24143394 on Build GRCh38

View Full Product Details