Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31070447-31070991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSC2
Entrez Gene ID
1824
Gene Name
desmocollin 2
Gene Aliases
ARVD11, CDHF2, DG2, DGII/III, DSC3
UniGene
Hs.95612
Gene Chromosome Location
Chr.18: 31065974-31102432 on Build GRCh38
Transcripts
NM_004949.4 NM_024422.4 XM_005258206.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6083169 c.2250G>T p.V750V Chr.18: 31070726-31070726 on Build GRCh38
6083167 c.2239G>A p.D747N Chr.18: 31070737-31070737 on Build GRCh38
3970516 c.2138C>T p.T713M Chr.18: 31070838-31070838 on Build GRCh38
5585058 c.2145C>A p.V715V Chr.18: 31070831-31070831 on Build GRCh38
6003379 c.2183T>A p.I728N Chr.18: 31070793-31070793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754918811 Chr.18: 31070729-31070729 on Build GRCh38
rs1051424082 Chr.18: 31070672-31070672 on Build GRCh38
rs780367614 Chr.18: 31070848-31070848 on Build GRCh38
rs936641955 Chr.18: 31070857-31070857 on Build GRCh38
rs981341211 Chr.18: 31070827-31070827 on Build GRCh38
rs746396691 Chr.18: 31070683-31070683 on Build GRCh38
rs768971007 Chr.18: 31070860-31070860 on Build GRCh38
rs549986406 Chr.18: 31070914-31070914 on Build GRCh38
rs768946883 Chr.18: 31070872-31070872 on Build GRCh38
rs563005744 Chr.18: 31070948-31070948 on Build GRCh38
rs773487167 Chr.18: 31070881-31070881 on Build GRCh38
rs902548278 Chr.18: 31070588-31070588 on Build GRCh38
rs760610120 Chr.18: 31070896-31070896 on Build GRCh38
rs767518318 Chr.18: 31070716-31070716 on Build GRCh38
rs906398422 Chr.18: 31070933-31070933 on Build GRCh38
rs897102270 Chr.18: 31070627-31070627 on Build GRCh38
rs759031428 Chr.18: 31070636-31070636 on Build GRCh38
rs753775769 Chr.18: 31070728-31070728 on Build GRCh38
rs1051022664 Chr.18: 31070902-31070902 on Build GRCh38
rs886039103 Chr.18: 31070732-31070732 on Build GRCh38
rs371950232 Chr.18: 31070770-31070770 on Build GRCh38
rs771061771 Chr.18: 31070897-31070897 on Build GRCh38
rs941066003 Chr.18: 31070566-31070566 on Build GRCh38
rs376355669 Chr.18: 31070826-31070826 on Build GRCh38
rs112532429 Chr.18: 31070837-31070837 on Build GRCh38
rs775526125 Chr.18: 31070901-31070901 on Build GRCh38
rs145831682 Chr.18: 31070850-31070850 on Build GRCh38
rs759513934 Chr.18: 31070814-31070814 on Build GRCh38
rs180863872 Chr.18: 31070838-31070838 on Build GRCh38
rs540366026 Chr.18: 31070834-31070834 on Build GRCh38
rs763152020 Chr.18: 31070700-31070700 on Build GRCh38
rs779297158 Chr.18: 31070870-31070870 on Build GRCh38
rs756785687 Chr.18: 31070760-31070760 on Build GRCh38
rs751228044 Chr.18: 31070830-31070830 on Build GRCh38
rs771736377 Chr.18: 31070673-31070673 on Build GRCh38
rs956665667 Chr.18: 31070759-31070759 on Build GRCh38
rs912788402 Chr.18: 31070536-31070536 on Build GRCh38
rs559941287 Chr.18: 31070557-31070557 on Build GRCh38
rs770166480 Chr.18: 31070689-31070689 on Build GRCh38
rs876657788 Chr.18: 31070802-31070802 on Build GRCh38
rs762055797 Chr.18: 31070785-31070785 on Build GRCh38
rs727504534 Chr.18: 31070768-31070768 on Build GRCh38
rs541592397 Chr.18: 31070502-31070502 on Build GRCh38
rs766601913 Chr.18: 31070850-31070850 on Build GRCh38
rs758269321 Chr.18: 31070745-31070745 on Build GRCh38
rs767611804 Chr.18: 31070798-31070798 on Build GRCh38
rs774561385 Chr.18: 31070712-31070712 on Build GRCh38
rs1037987784 Chr.18: 31070613-31070613 on Build GRCh38
rs763850631 Chr.18: 31070949-31070949 on Build GRCh38
rs574688294 Chr.18: 31070484-31070484 on Build GRCh38
rs376526973 Chr.18: 31070864-31070864 on Build GRCh38
rs752499852 Chr.18: 31070739-31070739 on Build GRCh38
rs775049004 Chr.18: 31070710-31070710 on Build GRCh38
rs146701735 Chr.18: 31070944-31070944 on Build GRCh38
rs749589324 Chr.18: 31070775-31070775 on Build GRCh38
rs773220437 Chr.18: 31070813-31070813 on Build GRCh38
rs201039801 Chr.18: 31070714-31070714 on Build GRCh38
rs985629599 Chr.18: 31070819-31070819 on Build GRCh38
rs979880174 Chr.18: 31070509-31070509 on Build GRCh38
rs746679441 Chr.18: 31070756-31070756 on Build GRCh38
rs151024019 Chr.18: 31070782-31070782 on Build GRCh38
rs775922060 Chr.18: 31070774-31070774 on Build GRCh38
rs929927530 Chr.18: 31070652-31070652 on Build GRCh38
rs895212019 Chr.18: 31070875-31070875 on Build GRCh38
rs765017458 Chr.18: 31070824-31070824 on Build GRCh38
rs780610767 Chr.18: 31070761-31070761 on Build GRCh38
rs777143396 Chr.18: 31070677-31070677 on Build GRCh38
rs1046220549 Chr.18: 31070894-31070894 on Build GRCh38
rs775771127 Chr.18: 31070692-31070692 on Build GRCh38
rs886053695 Chr.18: 31070778-31070778 on Build GRCh38
rs760915248 Chr.18: 31070824-31070824 on Build GRCh38
rs921649870 Chr.18: 31070801-31070801 on Build GRCh38
rs765067237 Chr.18: 31070731-31070731 on Build GRCh38
rs730880076 Chr.18: 31070779-31070779 on Build GRCh38
rs752691135 Chr.18: 31070825-31070825 on Build GRCh38
rs769022411 Chr.18: 31070776-31070776 on Build GRCh38
rs748354683 Chr.18: 31070878-31070878 on Build GRCh38
rs142807209 Chr.18: 31070750-31070750 on Build GRCh38
rs764008863 Chr.18: 31070829-31070829 on Build GRCh38
rs886053694 Chr.18: 31070771-31070771 on Build GRCh38
rs776449710 Chr.18: 31070523-31070523 on Build GRCh38

View Full Product Details