Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31071605-31072127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSC2
Entrez Gene ID
1824
Gene Name
desmocollin 2
Gene Aliases
ARVD11, CDHF2, DG2, DGII/III, DSC3
UniGene
Hs.95612
Gene Chromosome Location
Chr.18: 31065974-31102432 on Build GRCh38
Transcripts
NM_004949.4 NM_024422.4 XM_005258206.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3524862 c.1912C>T p.Q638* Chr.18: 31071818-31071818 on Build GRCh38
7311571 c.2030G>C p.C677S Chr.18: 31071700-31071700 on Build GRCh38
6225418 c.2118G>A p.L706L Chr.18: 31071612-31071612 on Build GRCh38
3821305 c.2020G>A p.E674K Chr.18: 31071710-31071710 on Build GRCh38
1725116 c.2059G>A p.G687S Chr.18: 31071671-31071671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113126164 Chr.18: 31071954-31071954 on Build GRCh38
rs745879343 Chr.18: 31071751-31071751 on Build GRCh38
rs758646191 Chr.18: 31071734-31071734 on Build GRCh38
rs542585921 Chr.18: 31071838-31071838 on Build GRCh38
rs963749078 Chr.18: 31071919-31071919 on Build GRCh38
rs754488475 Chr.18: 31071699-31071699 on Build GRCh38
rs562928418 Chr.18: 31071777-31071777 on Build GRCh38
rs889049693 Chr.18: 31071861-31071861 on Build GRCh38
rs138904684 Chr.18: 31071925-31071925 on Build GRCh38
rs899415985 Chr.18: 31071865-31071865 on Build GRCh38
rs762974426 Chr.18: 31071658-31071658 on Build GRCh38
rs753266534 Chr.18: 31071713-31071713 on Build GRCh38
rs145710244 Chr.18: 31071694-31071694 on Build GRCh38
rs869025387 Chr.18: 31071717-31071718 on Build GRCh38
rs201541042 Chr.18: 31071844-31071844 on Build GRCh38
rs781511528 Chr.18: 31071854-31071854 on Build GRCh38
rs147742157 Chr.18: 31071816-31071816 on Build GRCh38
rs200067684 Chr.18: 31071870-31071870 on Build GRCh38
rs756223772 Chr.18: 31071873-31071873 on Build GRCh38
rs975331812 Chr.18: 31071690-31071690 on Build GRCh38
rs374002111 Chr.18: 31071691-31071691 on Build GRCh38
rs998591665 Chr.18: 31071899-31071899 on Build GRCh38
rs868527749 Chr.18: 31072030-31072030 on Build GRCh38
rs186201750 Chr.18: 31072002-31072002 on Build GRCh38
rs770010850 Chr.18: 31071753-31071753 on Build GRCh38
rs776903280 Chr.18: 31071657-31071657 on Build GRCh38
rs397517395 Chr.18: 31071792-31071792 on Build GRCh38
rs146248554 Chr.18: 31071924-31071924 on Build GRCh38
rs926725037 Chr.18: 31071959-31071959 on Build GRCh38
rs199616533 Chr.18: 31071671-31071671 on Build GRCh38
rs963715928 Chr.18: 31071646-31071646 on Build GRCh38
rs773026094 Chr.18: 31071884-31071884 on Build GRCh38
rs746610878 Chr.18: 31072091-31072091 on Build GRCh38
rs747173696 Chr.18: 31071740-31071740 on Build GRCh38
rs776278152 Chr.18: 31071804-31071804 on Build GRCh38
rs949214007 Chr.18: 31071942-31071942 on Build GRCh38
rs912230070 Chr.18: 31071643-31071643 on Build GRCh38
rs181237366 Chr.18: 31071963-31071963 on Build GRCh38
rs200475862 Chr.18: 31071829-31071829 on Build GRCh38
rs267605146 Chr.18: 31071812-31071812 on Build GRCh38
rs778236763 Chr.18: 31071736-31071736 on Build GRCh38
rs745357999 Chr.18: 31071923-31071923 on Build GRCh38
rs57830743 Chr.18: 31071934-31071934 on Build GRCh38
rs1008404256 Chr.18: 31071896-31071896 on Build GRCh38
rs201469817 Chr.18: 31071711-31071711 on Build GRCh38
rs878855084 Chr.18: 31071795-31071795 on Build GRCh38
rs772112403 Chr.18: 31071880-31071880 on Build GRCh38
rs1009067550 Chr.18: 31072064-31072064 on Build GRCh38
rs765948147 Chr.18: 31071784-31071784 on Build GRCh38
rs781419827 Chr.18: 31071749-31071749 on Build GRCh38
rs899446910 Chr.18: 31072018-31072018 on Build GRCh38
rs1019323358 Chr.18: 31071900-31071900 on Build GRCh38
rs771735507 Chr.18: 31071778-31071778 on Build GRCh38
rs369281243 Chr.18: 31071814-31071814 on Build GRCh38
rs375842356 Chr.18: 31071716-31071716 on Build GRCh38
rs868423764 Chr.18: 31071650-31071650 on Build GRCh38
rs929577916 Chr.18: 31072024-31072024 on Build GRCh38
rs794728062 Chr.18: 31071697-31071697 on Build GRCh38
rs764355417 Chr.18: 31071823-31071823 on Build GRCh38
rs902914213 Chr.18: 31071888-31071888 on Build GRCh38
rs757253682 Chr.18: 31071692-31071692 on Build GRCh38
rs987960431 Chr.18: 31071701-31071701 on Build GRCh38
rs758990451 Chr.18: 31071820-31071820 on Build GRCh38
rs957922794 Chr.18: 31071926-31071926 on Build GRCh38
rs74374471 Chr.18: 31071877-31071877 on Build GRCh38
rs760238296 Chr.18: 31071782-31071782 on Build GRCh38
rs768784624 Chr.18: 31071662-31071662 on Build GRCh38
rs397517396 Chr.18: 31071759-31071759 on Build GRCh38
rs368353349 Chr.18: 31071806-31071806 on Build GRCh38
rs373329428 Chr.18: 31072029-31072029 on Build GRCh38
rs565645256 Chr.18: 31072050-31072050 on Build GRCh38
rs553599693 Chr.18: 31072014-31072014 on Build GRCh38
rs757613258 Chr.18: 31071744-31071744 on Build GRCh38
rs761458001 Chr.18: 31071673-31071673 on Build GRCh38
rs780224772 Chr.18: 31071874-31071874 on Build GRCh38
rs989615497 Chr.18: 31071931-31071931 on Build GRCh38
rs913738245 Chr.18: 31071940-31071940 on Build GRCh38
rs397517397 Chr.18: 31071648-31071648 on Build GRCh38
rs746747801 Chr.18: 31071889-31071889 on Build GRCh38
rs759893001 Chr.18: 31071850-31071853 on Build GRCh38
rs770883208 Chr.18: 31071651-31071651 on Build GRCh38
rs773110364 Chr.18: 31071779-31071779 on Build GRCh38

View Full Product Details