Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31622848-31623358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT6
Entrez Gene ID
9331
Gene Name
beta-1,4-galactosyltransferase 6
Gene Aliases
B4Gal-T6, beta4Gal-T6
UniGene
Hs.591063
Gene Chromosome Location
Chr.18: 31622246-31724670 on Build GRCh38
Transcripts
NM_004775.3 XM_005258387.4 XM_006722578.3 XM_006722579.3 XM_017026088.1 XM_017026089.1 XM_017026090.1 XM_017026091.1 XM_017026092.1

SNP Detail

SNP ID Chromosome Location
rs961808558 Chr.18: 31623149-31623149 on Build GRCh38
rs34022043 Chr.18: 31623066-31623067 on Build GRCh38
rs1037299451 Chr.18: 31623092-31623092 on Build GRCh38
rs1049861687 Chr.18: 31622907-31622907 on Build GRCh38
rs185106118 Chr.18: 31623039-31623039 on Build GRCh38
rs543436137 Chr.18: 31623178-31623178 on Build GRCh38
rs986281277 Chr.18: 31622965-31622965 on Build GRCh38
rs558258759 Chr.18: 31623316-31623319 on Build GRCh38
rs549070624 Chr.18: 31622964-31622964 on Build GRCh38
rs569628554 Chr.18: 31622952-31622952 on Build GRCh38
rs1023430334 Chr.18: 31623111-31623111 on Build GRCh38
rs551738913 Chr.18: 31622926-31622926 on Build GRCh38
rs977608419 Chr.18: 31623257-31623257 on Build GRCh38
rs920197669 Chr.18: 31623110-31623110 on Build GRCh38
rs938785327 Chr.18: 31623298-31623298 on Build GRCh38
rs796444617 Chr.18: 31623056-31623056 on Build GRCh38
rs193170324 Chr.18: 31622887-31622887 on Build GRCh38
rs902542501 Chr.18: 31623274-31623274 on Build GRCh38
rs938647432 Chr.18: 31623024-31623024 on Build GRCh38
rs923522289 Chr.18: 31622927-31622927 on Build GRCh38
rs915901939 Chr.18: 31623324-31623324 on Build GRCh38
rs1057196682 Chr.18: 31623047-31623047 on Build GRCh38
rs373662988 Chr.18: 31623162-31623162 on Build GRCh38
rs1029960620 Chr.18: 31623204-31623204 on Build GRCh38
rs970948553 Chr.18: 31623318-31623318 on Build GRCh38
rs924689337 Chr.18: 31623285-31623285 on Build GRCh38
rs547190829 Chr.18: 31623000-31623000 on Build GRCh38
rs951797393 Chr.18: 31622879-31622879 on Build GRCh38
rs1012587824 Chr.18: 31623108-31623108 on Build GRCh38
rs984950228 Chr.18: 31622891-31622891 on Build GRCh38
rs1014521497 Chr.18: 31622901-31622901 on Build GRCh38
rs772818932 Chr.18: 31623016-31623016 on Build GRCh38
rs115186394 Chr.18: 31622962-31622962 on Build GRCh38
rs997130338 Chr.18: 31623195-31623195 on Build GRCh38
rs975749453 Chr.18: 31622914-31622914 on Build GRCh38
rs903294657 Chr.18: 31623011-31623011 on Build GRCh38
rs17803165 Chr.18: 31622988-31622988 on Build GRCh38
rs369764215 Chr.18: 31623082-31623082 on Build GRCh38
rs983250228 Chr.18: 31623333-31623333 on Build GRCh38
rs959282805 Chr.18: 31623302-31623302 on Build GRCh38
rs76992272 Chr.18: 31623314-31623314 on Build GRCh38
rs183757867 Chr.18: 31623230-31623230 on Build GRCh38
rs140359087 Chr.18: 31623008-31623008 on Build GRCh38
rs1001289141 Chr.18: 31623291-31623291 on Build GRCh38
rs961701014 Chr.18: 31622912-31622912 on Build GRCh38
rs953616121 Chr.18: 31623215-31623215 on Build GRCh38
rs1008694387 Chr.18: 31622959-31622959 on Build GRCh38
rs568107199 Chr.18: 31623296-31623296 on Build GRCh38
rs555409623 Chr.18: 31623247-31623247 on Build GRCh38
rs778366215 Chr.18: 31623264-31623264 on Build GRCh38
rs887627200 Chr.18: 31623113-31623113 on Build GRCh38
rs189431887 Chr.18: 31623124-31623124 on Build GRCh38
rs768030775 Chr.18: 31623085-31623085 on Build GRCh38
rs143416107 Chr.18: 31622911-31622911 on Build GRCh38
rs1014615225 Chr.18: 31623140-31623140 on Build GRCh38
rs867966973 Chr.18: 31623231-31623231 on Build GRCh38
rs192477492 Chr.18: 31623137-31623137 on Build GRCh38
rs147177441 Chr.18: 31622995-31622995 on Build GRCh38

View Full Product Details