Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 32974468-32974965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC178
Entrez Gene ID
374864
Gene Name
coiled-coil domain containing 178
Gene Aliases
C18orf34
UniGene
Hs.115461
Gene Chromosome Location
Chr.18: 32937402-33441040 on Build GRCh38
Transcripts
NM_001105528.1 NM_001308126.1 NM_198995.2 XM_011525948.1 XM_011525951.1 XM_017025721.1 XM_017025722.1 XM_017025723.1 XM_017025724.1 XM_017025725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4930353 c.2435T>A p.F812Y Chr.18: 32974635-32974635 on Build GRCh38
1303693 c.2349G>A p.M783I Chr.18: 32974607-32974607 on Build GRCh38
4930354 c.2321T>A p.F774Y Chr.18: 32974635-32974635 on Build GRCh38
4923861 c.2399T>A p.L800Q Chr.18: 32974671-32974671 on Build GRCh38
4923862 c.2285T>A p.L762Q Chr.18: 32974671-32974671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763424125 Chr.18: 32974876-32974876 on Build GRCh38
rs371552745 Chr.18: 32974589-32974589 on Build GRCh38
rs756461860 Chr.18: 32974538-32974538 on Build GRCh38
rs754219224 Chr.18: 32974662-32974662 on Build GRCh38
rs1003717980 Chr.18: 32974832-32974832 on Build GRCh38
rs61747484 Chr.18: 32974779-32974779 on Build GRCh38
rs749479987 Chr.18: 32974572-32974572 on Build GRCh38
rs533623752 Chr.18: 32974746-32974746 on Build GRCh38
rs746955163 Chr.18: 32974503-32974503 on Build GRCh38
rs200388828 Chr.18: 32974600-32974600 on Build GRCh38
rs1016183680 Chr.18: 32974940-32974940 on Build GRCh38
rs906788977 Chr.18: 32974806-32974806 on Build GRCh38
rs935672830 Chr.18: 32974766-32974766 on Build GRCh38
rs542659548 Chr.18: 32974627-32974627 on Build GRCh38
rs761287196 Chr.18: 32974615-32974615 on Build GRCh38
rs763605372 Chr.18: 32974526-32974526 on Build GRCh38
rs776072184 Chr.18: 32974809-32974809 on Build GRCh38
rs751086764 Chr.18: 32974681-32974681 on Build GRCh38
rs766669256 Chr.18: 32974641-32974641 on Build GRCh38
rs756218203 Chr.18: 32974495-32974495 on Build GRCh38
rs965871391 Chr.18: 32974938-32974938 on Build GRCh38
rs780429501 Chr.18: 32974552-32974552 on Build GRCh38
rs560840106 Chr.18: 32974634-32974634 on Build GRCh38
rs1027719269 Chr.18: 32974861-32974861 on Build GRCh38
rs375762929 Chr.18: 32974517-32974517 on Build GRCh38
rs1033678024 Chr.18: 32974551-32974551 on Build GRCh38
rs983296085 Chr.18: 32974887-32974887 on Build GRCh38
rs907488229 Chr.18: 32974899-32974899 on Build GRCh38
rs771954203 Chr.18: 32974803-32974803 on Build GRCh38
rs1004373742 Chr.18: 32974936-32974936 on Build GRCh38
rs767977316 Chr.18: 32974535-32974535 on Build GRCh38
rs995273552 Chr.18: 32974846-32974846 on Build GRCh38
rs770605714 Chr.18: 32974605-32974606 on Build GRCh38
rs935758401 Chr.18: 32974823-32974823 on Build GRCh38
rs201647936 Chr.18: 32974558-32974558 on Build GRCh38
rs199823753 Chr.18: 32974506-32974506 on Build GRCh38
rs776633813 Chr.18: 32974509-32974509 on Build GRCh38
rs145798402 Chr.18: 32974708-32974708 on Build GRCh38
rs557095493 Chr.18: 32974595-32974595 on Build GRCh38
rs758581618 Chr.18: 32974697-32974697 on Build GRCh38
rs750844147 Chr.18: 32974536-32974536 on Build GRCh38
rs780760125 Chr.18: 32974647-32974648 on Build GRCh38
rs1009720388 Chr.18: 32974800-32974800 on Build GRCh38
rs924031750 Chr.18: 32974675-32974675 on Build GRCh38
rs950901757 Chr.18: 32974882-32974882 on Build GRCh38
rs777841251 Chr.18: 32974701-32974701 on Build GRCh38
rs138659028 Chr.18: 32974891-32974891 on Build GRCh38
rs891568802 Chr.18: 32974790-32974790 on Build GRCh38
rs759220363 Chr.18: 32974513-32974513 on Build GRCh38
rs959335674 Chr.18: 32974845-32974845 on Build GRCh38
rs147677442 Chr.18: 32974576-32974576 on Build GRCh38
rs372732510 Chr.18: 32974703-32974703 on Build GRCh38
rs1042991986 Chr.18: 32974804-32974804 on Build GRCh38
rs777650739 Chr.18: 32974512-32974512 on Build GRCh38
rs376925410 Chr.18: 32974645-32974645 on Build GRCh38
rs572263200 Chr.18: 32974579-32974579 on Build GRCh38
rs752873388 Chr.18: 32974694-32974694 on Build GRCh38
rs1033897423 Chr.18: 32974834-32974834 on Build GRCh38
rs189685071 Chr.18: 32974939-32974939 on Build GRCh38
rs201959675 Chr.18: 32974611-32974611 on Build GRCh38
rs779776918 Chr.18: 32974725-32974725 on Build GRCh38
rs771033497 Chr.18: 32974584-32974584 on Build GRCh38
rs761181378 Chr.18: 32974640-32974640 on Build GRCh38
rs757825168 Chr.18: 32974533-32974533 on Build GRCh38
rs745778282 Chr.18: 32974591-32974591 on Build GRCh38
rs749402102 Chr.18: 32974592-32974592 on Build GRCh38
rs115131430 Chr.18: 32974688-32974688 on Build GRCh38
rs745649516 Chr.18: 32974657-32974658 on Build GRCh38
rs751052765 Chr.18: 32974527-32974527 on Build GRCh38
rs368888946 Chr.18: 32974520-32974520 on Build GRCh38
rs142622043 Chr.18: 32974609-32974609 on Build GRCh38

View Full Product Details