Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 33215308-33215808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC178
Entrez Gene ID
374864
Gene Name
coiled-coil domain containing 178
Gene Aliases
C18orf34
UniGene
Hs.115461
Gene Chromosome Location
Chr.18: 32937402-33441040 on Build GRCh38
Transcripts
NM_001105528.1 NM_001308126.1 NM_198995.2 XM_011525948.1 XM_011525951.1 XM_017025721.1 XM_017025722.1 XM_017025723.1 XM_017025724.1 XM_017025725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1388460 c.2075T>G p.L692* Chr.18: 33215553-33215553 on Build GRCh38
5040987 c.2002A>G p.I668V Chr.18: 33215626-33215626 on Build GRCh38
5690369 c.1946A>T p.Q649L Chr.18: 33215568-33215568 on Build GRCh38
5040988 c.1888A>G p.I630V Chr.18: 33215626-33215626 on Build GRCh38
6621865 c.1933-4_1933-3insT p.? Chr.18: 33215698-33215699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027001133 Chr.18: 33215401-33215401 on Build GRCh38
rs190537483 Chr.18: 33215606-33215606 on Build GRCh38
rs368398499 Chr.18: 33215530-33215530 on Build GRCh38
rs752191887 Chr.18: 33215694-33215694 on Build GRCh38
rs773423783 Chr.18: 33215699-33215699 on Build GRCh38
rs753440368 Chr.18: 33215600-33215600 on Build GRCh38
rs141556619 Chr.18: 33215659-33215659 on Build GRCh38
rs766656505 Chr.18: 33215714-33215715 on Build GRCh38
rs766152063 Chr.18: 33215685-33215685 on Build GRCh38
rs757522678 Chr.18: 33215620-33215620 on Build GRCh38
rs781290469 Chr.18: 33215623-33215623 on Build GRCh38
rs753286855 Chr.18: 33215501-33215501 on Build GRCh38
rs542630209 Chr.18: 33215669-33215669 on Build GRCh38
rs188071651 Chr.18: 33215736-33215736 on Build GRCh38
rs768473871 Chr.18: 33215577-33215581 on Build GRCh38
rs1018233877 Chr.18: 33215632-33215632 on Build GRCh38
rs780350905 Chr.18: 33215735-33215735 on Build GRCh38
rs755551397 Chr.18: 33215507-33215507 on Build GRCh38
rs754470875 Chr.18: 33215602-33215602 on Build GRCh38
rs564832471 Chr.18: 33215622-33215622 on Build GRCh38
rs542672484 Chr.18: 33215751-33215751 on Build GRCh38
rs148600773 Chr.18: 33215711-33215711 on Build GRCh38
rs753207695 Chr.18: 33215605-33215605 on Build GRCh38
rs547653738 Chr.18: 33215437-33215437 on Build GRCh38
rs767429656 Chr.18: 33215590-33215590 on Build GRCh38
rs769053961 Chr.18: 33215431-33215431 on Build GRCh38
rs951500656 Chr.18: 33215453-33215453 on Build GRCh38
rs760813652 Chr.18: 33215698-33215699 on Build GRCh38
rs370176857 Chr.18: 33215647-33215647 on Build GRCh38
rs925213643 Chr.18: 33215520-33215520 on Build GRCh38
rs536715092 Chr.18: 33215532-33215532 on Build GRCh38
rs746086356 Chr.18: 33215712-33215712 on Build GRCh38
rs376508906 Chr.18: 33215698-33215698 on Build GRCh38
rs199585390 Chr.18: 33215702-33215702 on Build GRCh38
rs942781348 Chr.18: 33215350-33215350 on Build GRCh38
rs769859990 Chr.18: 33215587-33215587 on Build GRCh38
rs760474921 Chr.18: 33215596-33215596 on Build GRCh38
rs761771379 Chr.18: 33215589-33215589 on Build GRCh38
rs773185121 Chr.18: 33215668-33215668 on Build GRCh38
rs769766212 Chr.18: 33215633-33215633 on Build GRCh38
rs768688851 Chr.18: 33215547-33215548 on Build GRCh38
rs1052777509 Chr.18: 33215495-33215498 on Build GRCh38
rs554440136 Chr.18: 33215577-33215577 on Build GRCh38
rs773131720 Chr.18: 33215591-33215591 on Build GRCh38
rs985442655 Chr.18: 33215419-33215420 on Build GRCh38
rs760993717 Chr.18: 33215626-33215626 on Build GRCh38
rs764766573 Chr.18: 33215603-33215603 on Build GRCh38
rs934416946 Chr.18: 33215486-33215486 on Build GRCh38
rs548726292 Chr.18: 33215550-33215550 on Build GRCh38
rs752012433 Chr.18: 33215703-33215703 on Build GRCh38
rs559325540 Chr.18: 33215454-33215454 on Build GRCh38
rs535810036 Chr.18: 33215568-33215568 on Build GRCh38
rs1019673291 Chr.18: 33215732-33215732 on Build GRCh38
rs374381010 Chr.18: 33215536-33215536 on Build GRCh38
rs776180170 Chr.18: 33215688-33215688 on Build GRCh38
rs1057361150 Chr.18: 33215750-33215750 on Build GRCh38
rs138991378 Chr.18: 33215628-33215628 on Build GRCh38
rs577108818 Chr.18: 33215351-33215351 on Build GRCh38
rs576629806 Chr.18: 33215658-33215658 on Build GRCh38
rs755868769 Chr.18: 33215519-33215522 on Build GRCh38
rs117839804 Chr.18: 33215480-33215480 on Build GRCh38
rs757965966 Chr.18: 33215543-33215546 on Build GRCh38
rs898725913 Chr.18: 33215416-33215416 on Build GRCh38
rs867565267 Chr.18: 33215642-33215642 on Build GRCh38
rs779988464 Chr.18: 33215522-33215523 on Build GRCh38
rs1019619497 Chr.18: 33215466-33215466 on Build GRCh38
rs1025632329 Chr.18: 33215436-33215436 on Build GRCh38
rs573968133 Chr.18: 33215759-33215759 on Build GRCh38
rs200395184 Chr.18: 33215613-33215613 on Build GRCh38
rs774183656 Chr.18: 33215588-33215588 on Build GRCh38
rs748218897 Chr.18: 33215585-33215587 on Build GRCh38
rs551114960 Chr.18: 33215519-33215519 on Build GRCh38
rs747342331 Chr.18: 33215617-33215617 on Build GRCh38
rs374324334 Chr.18: 33215716-33215716 on Build GRCh38
rs185692505 Chr.18: 33215565-33215565 on Build GRCh38
rs569423926 Chr.18: 33215523-33215523 on Build GRCh38
rs748658191 Chr.18: 33215482-33215482 on Build GRCh38
rs765918268 Chr.18: 33215599-33215599 on Build GRCh38
rs1001669801 Chr.18: 33215342-33215342 on Build GRCh38
rs372954179 Chr.18: 33215661-33215661 on Build GRCh38
rs370362165 Chr.18: 33215709-33215709 on Build GRCh38
rs113751117 Chr.18: 33215531-33215531 on Build GRCh38
rs914304772 Chr.18: 33215504-33215507 on Build GRCh38
rs774108139 Chr.18: 33215578-33215578 on Build GRCh38
rs183451981 Chr.18: 33215410-33215410 on Build GRCh38
rs150710440 Chr.18: 33215689-33215689 on Build GRCh38
rs994220093 Chr.18: 33215399-33215399 on Build GRCh38
rs772064858 Chr.18: 33215634-33215635 on Build GRCh38

View Full Product Details