Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 33731882-33732377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASXL3
Entrez Gene ID
80816
Gene Name
additional sex combs like 3, transcriptional regulator
Gene Aliases
BRPS, KIAA1713
UniGene
Hs.464876
Gene Chromosome Location
Chr.18: 33578577-33751195 on Build GRCh38
Transcripts
NM_030632.2 XM_005258356.1 XM_011526205.2 XM_011526206.2 XM_011526209.1 XM_011526210.1 XM_011526212.1 XM_011526213.1 XM_017026012.1 XM_017026013.1 XM_017026014.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183846 c.896T>G p.I299S Chr.18: 33731984-33731984 on Build GRCh38
1183847 c.17T>G p.I6S Chr.18: 33731984-33731984 on Build GRCh38
4395996 c.976G>A p.G326R Chr.18: 33732064-33732064 on Build GRCh38
2810251 c.68C>A p.A23E Chr.18: 33732035-33732035 on Build GRCh38
987886 c.952G>T p.G318W Chr.18: 33732040-33732040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052889905 Chr.18: 33732299-33732299 on Build GRCh38
rs980113090 Chr.18: 33732183-33732183 on Build GRCh38
rs778077409 Chr.18: 33732095-33732095 on Build GRCh38
rs372250848 Chr.18: 33732052-33732052 on Build GRCh38
rs778378012 Chr.18: 33732087-33732088 on Build GRCh38
rs769172263 Chr.18: 33732270-33732270 on Build GRCh38
rs997289376 Chr.18: 33732280-33732280 on Build GRCh38
rs757363000 Chr.18: 33732109-33732109 on Build GRCh38
rs578145074 Chr.18: 33732162-33732162 on Build GRCh38
rs547955930 Chr.18: 33732088-33732088 on Build GRCh38
rs911481540 Chr.18: 33732241-33732241 on Build GRCh38
rs371504132 Chr.18: 33732281-33732283 on Build GRCh38
rs867317785 Chr.18: 33732042-33732042 on Build GRCh38
rs1050261678 Chr.18: 33732351-33732351 on Build GRCh38
rs529895456 Chr.18: 33732114-33732114 on Build GRCh38
rs139327492 Chr.18: 33731908-33731908 on Build GRCh38
rs760311379 Chr.18: 33732054-33732054 on Build GRCh38
rs149979379 Chr.18: 33732105-33732105 on Build GRCh38
rs749998326 Chr.18: 33732053-33732053 on Build GRCh38
rs770948375 Chr.18: 33731975-33731975 on Build GRCh38
rs755530224 Chr.18: 33732080-33732080 on Build GRCh38
rs758650050 Chr.18: 33732094-33732094 on Build GRCh38
rs760725458 Chr.18: 33732268-33732268 on Build GRCh38
rs1021553717 Chr.18: 33732153-33732153 on Build GRCh38
rs778639580 Chr.18: 33732038-33732038 on Build GRCh38
rs113843395 Chr.18: 33732316-33732316 on Build GRCh38
rs960145790 Chr.18: 33732185-33732185 on Build GRCh38
rs781478563 Chr.18: 33732317-33732317 on Build GRCh38
rs775451402 Chr.18: 33731982-33731982 on Build GRCh38
rs779333335 Chr.18: 33731931-33731931 on Build GRCh38
rs972859572 Chr.18: 33732193-33732193 on Build GRCh38
rs193195531 Chr.18: 33731952-33731952 on Build GRCh38
rs1049088798 Chr.18: 33732234-33732234 on Build GRCh38
rs768502867 Chr.18: 33731990-33731990 on Build GRCh38
rs762844143 Chr.18: 33731983-33731983 on Build GRCh38
rs758867575 Chr.18: 33732081-33732082 on Build GRCh38
rs535736347 Chr.18: 33732262-33732262 on Build GRCh38
rs781242006 Chr.18: 33732113-33732113 on Build GRCh38
rs943013563 Chr.18: 33732242-33732242 on Build GRCh38
rs1021960580 Chr.18: 33732320-33732320 on Build GRCh38
rs761451645 Chr.18: 33732049-33732049 on Build GRCh38
rs748492487 Chr.18: 33732069-33732070 on Build GRCh38
rs747123290 Chr.18: 33731958-33731958 on Build GRCh38
rs777789527 Chr.18: 33731953-33731953 on Build GRCh38
rs549258822 Chr.18: 33732333-33732333 on Build GRCh38
rs969995625 Chr.18: 33732175-33732175 on Build GRCh38
rs779328462 Chr.18: 33731930-33731930 on Build GRCh38
rs754465235 Chr.18: 33732069-33732069 on Build GRCh38
rs10432168 Chr.18: 33732142-33732142 on Build GRCh38
rs866283297 Chr.18: 33732045-33732045 on Build GRCh38
rs779228191 Chr.18: 33732084-33732084 on Build GRCh38
rs527674539 Chr.18: 33732056-33732056 on Build GRCh38
rs7504560 Chr.18: 33732184-33732184 on Build GRCh38
rs931512004 Chr.18: 33732225-33732225 on Build GRCh38
rs918727107 Chr.18: 33732224-33732224 on Build GRCh38
rs901631238 Chr.18: 33732012-33732012 on Build GRCh38
rs774211783 Chr.18: 33732024-33732024 on Build GRCh38
rs541393351 Chr.18: 33732117-33732117 on Build GRCh38
rs748377649 Chr.18: 33731940-33731940 on Build GRCh38
rs550081271 Chr.18: 33732157-33732157 on Build GRCh38

View Full Product Details