Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 37244927-37245437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF4
Entrez Gene ID
56853
Gene Name
CUGBP, Elav-like family member 4
Gene Aliases
BRUNOL4, CELF-4
UniGene
Hs.435976
Gene Chromosome Location
Chr.18: 37243040-37566052 on Build GRCh38
Transcripts
NM_001025087.1 NM_001025088.1 NM_001025089.1 NM_020180.3 XM_005258308.3 XM_011526079.2 XM_011526081.2 XM_011526083.2 XM_011526084.2 XM_011526085.2 XM_011526086.2 XM_011526087.2 XM_011526090.2 XM_011526091.2 XM_011526092.2 XM_017025840.1 XM_017025841.1 XM_017025842.1 XM_017025843.1 XM_017025844.1 XM_017025845.1 XM_017025846.1 XM_017025847.1 XM_017025848.1 XM_017025849.1 XM_017025850.1 XM_017025851.1 XM_017025852.1 XM_017025853.1 XM_017025854.1 XM_017025855.1 XM_017025856.1 XM_017025858.1 XM_017025859.1 XM_017025860.1 XM_017025861.1 XM_017025862.1 XM_017025863.1 XM_017025864.1 XM_017025865.1 XM_017025866.1 XM_017025867.1 XM_017025868.1 XM_017025869.1 XM_017025870.1 XM_017025871.1 XM_017025872.1

SNP Detail

SNP ID Chromosome Location
rs927836922 Chr.18: 37244984-37244984 on Build GRCh38
rs1043855017 Chr.18: 37245101-37245101 on Build GRCh38
rs914067694 Chr.18: 37245028-37245028 on Build GRCh38
rs929225033 Chr.18: 37244952-37244953 on Build GRCh38
rs1030082745 Chr.18: 37245373-37245373 on Build GRCh38
rs540754161 Chr.18: 37245376-37245376 on Build GRCh38
rs565279263 Chr.18: 37245059-37245059 on Build GRCh38
rs1043055543 Chr.18: 37245033-37245033 on Build GRCh38
rs982376688 Chr.18: 37244980-37244980 on Build GRCh38
rs372623807 Chr.18: 37245339-37245339 on Build GRCh38
rs572135479 Chr.18: 37245361-37245361 on Build GRCh38
rs903463398 Chr.18: 37245077-37245077 on Build GRCh38
rs1052425449 Chr.18: 37245108-37245108 on Build GRCh38
rs564942697 Chr.18: 37244960-37244962 on Build GRCh38
rs922627690 Chr.18: 37244992-37244992 on Build GRCh38
rs187152953 Chr.18: 37245095-37245095 on Build GRCh38
rs895449163 Chr.18: 37245164-37245164 on Build GRCh38
rs751027779 Chr.18: 37245155-37245155 on Build GRCh38
rs886840256 Chr.18: 37245264-37245264 on Build GRCh38
rs919199515 Chr.18: 37244952-37244952 on Build GRCh38
rs1007690468 Chr.18: 37245149-37245149 on Build GRCh38
rs556865150 Chr.18: 37245011-37245011 on Build GRCh38
rs1016772303 Chr.18: 37245357-37245357 on Build GRCh38
rs768139289 Chr.18: 37245098-37245099 on Build GRCh38
rs544977802 Chr.18: 37245308-37245308 on Build GRCh38
rs576119645 Chr.18: 37245032-37245032 on Build GRCh38
rs368538277 Chr.18: 37245065-37245065 on Build GRCh38
rs1005419363 Chr.18: 37245299-37245299 on Build GRCh38
rs996734232 Chr.18: 37245371-37245371 on Build GRCh38
rs890644011 Chr.18: 37245114-37245114 on Build GRCh38
rs1014434895 Chr.18: 37245183-37245183 on Build GRCh38
rs754502679 Chr.18: 37245221-37245221 on Build GRCh38
rs994938824 Chr.18: 37245182-37245182 on Build GRCh38
rs955643693 Chr.18: 37245010-37245010 on Build GRCh38
rs118119532 Chr.18: 37245153-37245153 on Build GRCh38
rs937817675 Chr.18: 37244987-37244987 on Build GRCh38
rs926905695 Chr.18: 37245111-37245111 on Build GRCh38
rs982063872 Chr.18: 37245274-37245274 on Build GRCh38

View Full Product Details