Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 45840122-45840618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPG5
Entrez Gene ID
57724
Gene Name
ectopic P-granules autophagy protein 5 homolog
Gene Aliases
HEEW1, KIAA1632, VICIS
UniGene
Hs.514843
Gene Chromosome Location
Chr.18: 45800590-45967339 on Build GRCh38
Transcripts
XM_017025890.1 NM_213602.2

Gene Symbol
SIGLEC15
Entrez Gene ID
284266
Gene Name
sialic acid binding Ig like lectin 15
Gene Aliases
CD33L3, HsT1361, SIGLEC-15
UniGene
Hs.287692
Gene Chromosome Location
Chr.18: 45825580-45842556 on Build GRCh38
Transcripts
XM_017025890.1 NM_213602.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6148581 c.902C>A p.P301Q Chr.18: 45840238-45840238 on Build GRCh38
4000405 c.876G>A p.E292E Chr.18: 45840212-45840212 on Build GRCh38
6778404 c.891G>A p.P297P Chr.18: 45840227-45840227 on Build GRCh38
6778403 c.904C>T p.R302W Chr.18: 45840240-45840240 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs895597129 Chr.18: 45840424-45840424 on Build GRCh38
rs952360131 Chr.18: 45840358-45840358 on Build GRCh38
rs775641948 Chr.18: 45840168-45840168 on Build GRCh38
rs1036680770 Chr.18: 45840405-45840405 on Build GRCh38
rs750729197 Chr.18: 45840218-45840226 on Build GRCh38
rs568729219 Chr.18: 45840414-45840415 on Build GRCh38
rs894049351 Chr.18: 45840473-45840473 on Build GRCh38
rs765753007 Chr.18: 45840213-45840213 on Build GRCh38
rs1014026740 Chr.18: 45840439-45840439 on Build GRCh38
rs753415708 Chr.18: 45840215-45840215 on Build GRCh38
rs1018973197 Chr.18: 45840511-45840511 on Build GRCh38
rs750961639 Chr.18: 45840209-45840209 on Build GRCh38
rs769829923 Chr.18: 45840162-45840162 on Build GRCh38
rs1033419033 Chr.18: 45840232-45840232 on Build GRCh38
rs903718421 Chr.18: 45840451-45840451 on Build GRCh38
rs768832104 Chr.18: 45840277-45840277 on Build GRCh38
rs201873132 Chr.18: 45840257-45840257 on Build GRCh38
rs1002750734 Chr.18: 45840459-45840459 on Build GRCh38
rs187274750 Chr.18: 45840487-45840487 on Build GRCh38
rs767987359 Chr.18: 45840198-45840198 on Build GRCh38
rs554881390 Chr.18: 45840226-45840226 on Build GRCh38
rs777759596 Chr.18: 45840242-45840242 on Build GRCh38
rs369068735 Chr.18: 45840303-45840303 on Build GRCh38
rs1048313955 Chr.18: 45840362-45840362 on Build GRCh38
rs191761840 Chr.18: 45840500-45840500 on Build GRCh38
rs531777091 Chr.18: 45840493-45840493 on Build GRCh38
rs541176226 Chr.18: 45840368-45840368 on Build GRCh38
rs532250568 Chr.18: 45840337-45840337 on Build GRCh38
rs747664544 Chr.18: 45840565-45840565 on Build GRCh38
rs61104666 Chr.18: 45840212-45840212 on Build GRCh38
rs150566568 Chr.18: 45840443-45840443 on Build GRCh38
rs921113005 Chr.18: 45840310-45840310 on Build GRCh38
rs749412361 Chr.18: 45840274-45840274 on Build GRCh38
rs370574702 Chr.18: 45840151-45840151 on Build GRCh38
rs756423080 Chr.18: 45840217-45840218 on Build GRCh38
rs142753118 Chr.18: 45840574-45840574 on Build GRCh38
rs140370595 Chr.18: 45840517-45840517 on Build GRCh38
rs774602373 Chr.18: 45840283-45840283 on Build GRCh38
rs777418141 Chr.18: 45840564-45840564 on Build GRCh38
rs767733687 Chr.18: 45840204-45840207 on Build GRCh38
rs984344340 Chr.18: 45840562-45840562 on Build GRCh38
rs559382780 Chr.18: 45840435-45840435 on Build GRCh38
rs932436030 Chr.18: 45840361-45840361 on Build GRCh38
rs375172698 Chr.18: 45840190-45840190 on Build GRCh38
rs953933297 Chr.18: 45840555-45840555 on Build GRCh38
rs758328612 Chr.18: 45840241-45840241 on Build GRCh38
rs772358700 Chr.18: 45840287-45840287 on Build GRCh38
rs778749999 Chr.18: 45840227-45840227 on Build GRCh38
rs909780887 Chr.18: 45840365-45840365 on Build GRCh38
rs752519661 Chr.18: 45840240-45840240 on Build GRCh38
rs939925000 Chr.18: 45840585-45840585 on Build GRCh38
rs1026483087 Chr.18: 45840331-45840331 on Build GRCh38
rs114831310 Chr.18: 45840158-45840158 on Build GRCh38
rs368718179 Chr.18: 45840169-45840169 on Build GRCh38
rs991800942 Chr.18: 45840408-45840408 on Build GRCh38
rs896311408 Chr.18: 45840197-45840197 on Build GRCh38
rs554889394 Chr.18: 45840461-45840461 on Build GRCh38
rs565719173 Chr.18: 45840550-45840550 on Build GRCh38
rs901886461 Chr.18: 45840537-45840537 on Build GRCh38
rs571359969 Chr.18: 45840512-45840512 on Build GRCh38
rs117658632 Chr.18: 45840256-45840256 on Build GRCh38
rs199951548 Chr.18: 45840284-45840284 on Build GRCh38
rs536272942 Chr.18: 45840551-45840551 on Build GRCh38
rs572049329 Chr.18: 45840584-45840584 on Build GRCh38
rs1056167918 Chr.18: 45840170-45840170 on Build GRCh38
rs1019067949 Chr.18: 45840265-45840265 on Build GRCh38
rs1028799085 Chr.18: 45840522-45840522 on Build GRCh38
rs780535470 Chr.18: 45840223-45840223 on Build GRCh38
rs750315264 Chr.18: 45840155-45840155 on Build GRCh38
rs997212225 Chr.18: 45840231-45840231 on Build GRCh38
rs529750082 Chr.18: 45840442-45840442 on Build GRCh38
rs200018273 Chr.18: 45840185-45840185 on Build GRCh38
rs1013784994 Chr.18: 45840152-45840152 on Build GRCh38
rs553886336 Chr.18: 45840546-45840546 on Build GRCh38
rs547933374 Chr.18: 45840463-45840463 on Build GRCh38
rs1042579665 Chr.18: 45840448-45840448 on Build GRCh38

View Full Product Details