Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 46262798-46263345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf25
Entrez Gene ID
147339
Gene Name
chromosome 18 open reading frame 25
Gene Aliases
ARKL1, RNF111L1
UniGene
Hs.116486
Gene Chromosome Location
Chr.18: 46173553-46266992 on Build GRCh38
Transcripts
NM_001008239.2 NM_145055.3 XM_011525822.1 XM_017025555.1

SNP Detail

SNP ID Chromosome Location
rs945591367 Chr.18: 46262986-46262986 on Build GRCh38
rs768607066 Chr.18: 46263089-46263089 on Build GRCh38
rs895507161 Chr.18: 46262843-46262843 on Build GRCh38
rs764454031 Chr.18: 46263165-46263165 on Build GRCh38
rs758783585 Chr.18: 46263003-46263003 on Build GRCh38
rs1019274008 Chr.18: 46262829-46262829 on Build GRCh38
rs182863218 Chr.18: 46263022-46263022 on Build GRCh38
rs370825575 Chr.18: 46263019-46263019 on Build GRCh38
rs775254764 Chr.18: 46263038-46263038 on Build GRCh38
rs765562079 Chr.18: 46262992-46262992 on Build GRCh38
rs758549278 Chr.18: 46263164-46263164 on Build GRCh38
rs376437618 Chr.18: 46263268-46263268 on Build GRCh38
rs774998171 Chr.18: 46262915-46262915 on Build GRCh38
rs756783626 Chr.18: 46262905-46262905 on Build GRCh38
rs774456845 Chr.18: 46262969-46262969 on Build GRCh38
rs770459580 Chr.18: 46262891-46262891 on Build GRCh38
rs374621291 Chr.18: 46262985-46262985 on Build GRCh38
rs140741823 Chr.18: 46263203-46263203 on Build GRCh38
rs533497718 Chr.18: 46263304-46263304 on Build GRCh38
rs774224443 Chr.18: 46263090-46263090 on Build GRCh38
rs553223006 Chr.18: 46263295-46263295 on Build GRCh38
rs377662492 Chr.18: 46262920-46262920 on Build GRCh38
rs773021738 Chr.18: 46263100-46263100 on Build GRCh38
rs993809031 Chr.18: 46263219-46263219 on Build GRCh38
rs538332653 Chr.18: 46262899-46262899 on Build GRCh38
rs769541807 Chr.18: 46262913-46262913 on Build GRCh38
rs889526384 Chr.18: 46263057-46263059 on Build GRCh38
rs931113817 Chr.18: 46262876-46262876 on Build GRCh38
rs202185890 Chr.18: 46262947-46262947 on Build GRCh38
rs755001541 Chr.18: 46262991-46262991 on Build GRCh38
rs535879718 Chr.18: 46262877-46262877 on Build GRCh38
rs913995758 Chr.18: 46263046-46263046 on Build GRCh38
rs544868826 Chr.18: 46263261-46263261 on Build GRCh38
rs762103346 Chr.18: 46262995-46262996 on Build GRCh38
rs952664941 Chr.18: 46263259-46263259 on Build GRCh38
rs753491164 Chr.18: 46263144-46263144 on Build GRCh38
rs961275831 Chr.18: 46262830-46262830 on Build GRCh38
rs762843128 Chr.18: 46262925-46262925 on Build GRCh38
rs1048280906 Chr.18: 46263218-46263218 on Build GRCh38
rs551882332 Chr.18: 46263308-46263308 on Build GRCh38
rs754664124 Chr.18: 46263232-46263232 on Build GRCh38
rs938114764 Chr.18: 46262904-46262904 on Build GRCh38
rs763955763 Chr.18: 46262943-46262943 on Build GRCh38
rs761859081 Chr.18: 46263094-46263094 on Build GRCh38
rs765209941 Chr.18: 46263102-46263102 on Build GRCh38
rs749514967 Chr.18: 46262831-46262831 on Build GRCh38
rs753899195 Chr.18: 46262990-46262990 on Build GRCh38
rs1013938001 Chr.18: 46262868-46262868 on Build GRCh38
rs766974541 Chr.18: 46263130-46263131 on Build GRCh38
rs745518551 Chr.18: 46262901-46262901 on Build GRCh38
rs201781548 Chr.18: 46263123-46263123 on Build GRCh38
rs932599334 Chr.18: 46263204-46263204 on Build GRCh38
rs759702161 Chr.18: 46263101-46263101 on Build GRCh38
rs747518245 Chr.18: 46263011-46263011 on Build GRCh38
rs748847364 Chr.18: 46263067-46263067 on Build GRCh38
rs368297854 Chr.18: 46263008-46263008 on Build GRCh38
rs769152032 Chr.18: 46263032-46263032 on Build GRCh38
rs778072024 Chr.18: 46263006-46263006 on Build GRCh38
rs888316709 Chr.18: 46263225-46263225 on Build GRCh38
rs758209304 Chr.18: 46263240-46263240 on Build GRCh38
rs553380894 Chr.18: 46262982-46262982 on Build GRCh38
rs767621876 Chr.18: 46263096-46263096 on Build GRCh38
rs745447991 Chr.18: 46263028-46263028 on Build GRCh38
rs1028498976 Chr.18: 46263040-46263040 on Build GRCh38
rs150922345 Chr.18: 46262851-46262851 on Build GRCh38

View Full Product Details