Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 46264391-46264893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf25
Entrez Gene ID
147339
Gene Name
chromosome 18 open reading frame 25
Gene Aliases
ARKL1, RNF111L1
UniGene
Hs.116486
Gene Chromosome Location
Chr.18: 46173553-46266992 on Build GRCh38
Transcripts
NM_001008239.2 NM_145055.3 XM_011525822.1 XM_017025555.1

SNP Detail

SNP ID Chromosome Location
rs76109390 Chr.18: 46264751-46264751 on Build GRCh38
rs1051504665 Chr.18: 46264766-46264766 on Build GRCh38
rs578222433 Chr.18: 46264671-46264671 on Build GRCh38
rs949935049 Chr.18: 46264738-46264738 on Build GRCh38
rs1005660305 Chr.18: 46264611-46264611 on Build GRCh38
rs8086610 Chr.18: 46264695-46264695 on Build GRCh38
rs554586980 Chr.18: 46264470-46264470 on Build GRCh38
rs556859054 Chr.18: 46264660-46264660 on Build GRCh38
rs11405758 Chr.18: 46264735-46264736 on Build GRCh38
rs199524693 Chr.18: 46264708-46264709 on Build GRCh38
rs9959526 Chr.18: 46264771-46264771 on Build GRCh38
rs766153763 Chr.18: 46264567-46264567 on Build GRCh38
rs971314623 Chr.18: 46264696-46264696 on Build GRCh38
rs142440469 Chr.18: 46264451-46264456 on Build GRCh38
rs57160009 Chr.18: 46264605-46264605 on Build GRCh38
rs976993885 Chr.18: 46264630-46264630 on Build GRCh38
rs554069131 Chr.18: 46264770-46264770 on Build GRCh38
rs1055456900 Chr.18: 46264669-46264669 on Build GRCh38
rs901436092 Chr.18: 46264543-46264543 on Build GRCh38
rs895506976 Chr.18: 46264736-46264742 on Build GRCh38
rs990286800 Chr.18: 46264423-46264423 on Build GRCh38
rs35509991 Chr.18: 46264736-46264737 on Build GRCh38
rs758028287 Chr.18: 46264866-46264866 on Build GRCh38
rs937094054 Chr.18: 46264665-46264665 on Build GRCh38
rs957428635 Chr.18: 46264786-46264786 on Build GRCh38
rs1018267225 Chr.18: 46264612-46264612 on Build GRCh38
rs760648256 Chr.18: 46264459-46264459 on Build GRCh38
rs934222942 Chr.18: 46264545-46264545 on Build GRCh38
rs893885084 Chr.18: 46264853-46264853 on Build GRCh38
rs374510215 Chr.18: 46264465-46264470 on Build GRCh38
rs895701845 Chr.18: 46264427-46264427 on Build GRCh38
rs776497941 Chr.18: 46264673-46264673 on Build GRCh38
rs1000451243 Chr.18: 46264582-46264582 on Build GRCh38
rs200571596 Chr.18: 46264752-46264752 on Build GRCh38
rs965595486 Chr.18: 46264625-46264625 on Build GRCh38
rs768965632 Chr.18: 46264720-46264720 on Build GRCh38
rs1017434927 Chr.18: 46264661-46264661 on Build GRCh38
rs918741109 Chr.18: 46264653-46264653 on Build GRCh38
rs980001477 Chr.18: 46264711-46264711 on Build GRCh38
rs192305564 Chr.18: 46264590-46264590 on Build GRCh38
rs1034207105 Chr.18: 46264776-46264776 on Build GRCh38
rs774434935 Chr.18: 46264735-46264735 on Build GRCh38
rs1054706223 Chr.18: 46264838-46264838 on Build GRCh38
rs945695846 Chr.18: 46264481-46264481 on Build GRCh38
rs763566006 Chr.18: 46264456-46264466 on Build GRCh38
rs958367391 Chr.18: 46264605-46264605 on Build GRCh38
rs139249539 Chr.18: 46264482-46264482 on Build GRCh38
rs115931036 Chr.18: 46264788-46264788 on Build GRCh38
rs989949988 Chr.18: 46264852-46264852 on Build GRCh38

View Full Product Details