Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 65858835-65859344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH7
Entrez Gene ID
1005
Gene Name
cadherin 7
Gene Aliases
CDH7L1
UniGene
Hs.541003
Gene Chromosome Location
Chr.18: 65750252-65885136 on Build GRCh38
Transcripts
NM_001317214.1 NM_004361.3 NM_033646.2 XM_017025523.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7360556 c.1472G>C p.C491S Chr.18: 65859024-65859024 on Build GRCh38
1523521 c.1467C>T p.T489T Chr.18: 65859019-65859019 on Build GRCh38
5524727 c.1474G>A p.E492K Chr.18: 65859026-65859026 on Build GRCh38
5854391 c.1391G>A p.G464E Chr.18: 65858943-65858943 on Build GRCh38
1247877 c.1417C>A p.L473I Chr.18: 65858969-65858969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016829639 Chr.18: 65859264-65859264 on Build GRCh38
rs375888840 Chr.18: 65858992-65858992 on Build GRCh38
rs758119044 Chr.18: 65859091-65859091 on Build GRCh38
rs989469995 Chr.18: 65859229-65859229 on Build GRCh38
rs147764044 Chr.18: 65859019-65859019 on Build GRCh38
rs767181274 Chr.18: 65859037-65859037 on Build GRCh38
rs568863911 Chr.18: 65859078-65859078 on Build GRCh38
rs770479958 Chr.18: 65858969-65858969 on Build GRCh38
rs771231129 Chr.18: 65858963-65858963 on Build GRCh38
rs939088711 Chr.18: 65858873-65858873 on Build GRCh38
rs145272015 Chr.18: 65859040-65859040 on Build GRCh38
rs573289567 Chr.18: 65859274-65859274 on Build GRCh38
rs746337755 Chr.18: 65858966-65858966 on Build GRCh38
rs779529611 Chr.18: 65858921-65858921 on Build GRCh38
rs367817697 Chr.18: 65858882-65858882 on Build GRCh38
rs750355372 Chr.18: 65858889-65858889 on Build GRCh38
rs144893792 Chr.18: 65859249-65859249 on Build GRCh38
rs778588459 Chr.18: 65859080-65859080 on Build GRCh38
rs117028776 Chr.18: 65859208-65859208 on Build GRCh38
rs747724928 Chr.18: 65859082-65859082 on Build GRCh38
rs781396727 Chr.18: 65859095-65859095 on Build GRCh38
rs1016755844 Chr.18: 65859158-65859158 on Build GRCh38
rs149210579 Chr.18: 65859252-65859252 on Build GRCh38
rs756045305 Chr.18: 65859043-65859043 on Build GRCh38
rs370320169 Chr.18: 65859297-65859297 on Build GRCh38
rs747350932 Chr.18: 65858960-65858960 on Build GRCh38
rs753289706 Chr.18: 65859068-65859068 on Build GRCh38
rs765064010 Chr.18: 65859179-65859179 on Build GRCh38
rs761625164 Chr.18: 65859020-65859020 on Build GRCh38
rs371669828 Chr.18: 65858953-65858953 on Build GRCh38
rs962613641 Chr.18: 65859279-65859279 on Build GRCh38
rs781132344 Chr.18: 65858888-65858888 on Build GRCh38
rs886445090 Chr.18: 65859070-65859070 on Build GRCh38
rs756096853 Chr.18: 65858908-65858908 on Build GRCh38
rs768233043 Chr.18: 65858952-65858952 on Build GRCh38
rs749777659 Chr.18: 65858987-65858987 on Build GRCh38
rs759202083 Chr.18: 65859047-65859047 on Build GRCh38
rs112245860 Chr.18: 65859142-65859142 on Build GRCh38
rs777094089 Chr.18: 65858964-65858964 on Build GRCh38
rs748867401 Chr.18: 65858933-65858933 on Build GRCh38
rs368578868 Chr.18: 65859054-65859054 on Build GRCh38
rs757013274 Chr.18: 65858887-65858887 on Build GRCh38
rs116715722 Chr.18: 65859161-65859161 on Build GRCh38
rs945105506 Chr.18: 65858871-65858871 on Build GRCh38
rs141136047 Chr.18: 65859039-65859039 on Build GRCh38
rs918230455 Chr.18: 65859307-65859307 on Build GRCh38
rs997989143 Chr.18: 65859182-65859182 on Build GRCh38
rs916716569 Chr.18: 65858946-65858946 on Build GRCh38
rs367829639 Chr.18: 65858989-65858989 on Build GRCh38
rs557663186 Chr.18: 65859168-65859168 on Build GRCh38
rs190087257 Chr.18: 65859301-65859301 on Build GRCh38
rs775706195 Chr.18: 65858986-65858986 on Build GRCh38
rs945035749 Chr.18: 65859289-65859289 on Build GRCh38
rs748321894 Chr.18: 65859040-65859040 on Build GRCh38
rs1006254247 Chr.18: 65859130-65859130 on Build GRCh38
rs953981906 Chr.18: 65859226-65859226 on Build GRCh38
rs776661561 Chr.18: 65858988-65858988 on Build GRCh38

View Full Product Details