Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 66501051-66501588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH19
Entrez Gene ID
28513
Gene Name
cadherin 19
Gene Aliases
CDH7, CDH7L2
UniGene
Hs.42771
Gene Chromosome Location
Chr.18: 66501187-66604241 on Build GRCh38
Transcripts
NM_001271028.1 NM_021153.3 NR_073130.1 XM_011525931.2 XM_017025711.1

SNP Detail

SNP ID Chromosome Location
rs775188720 Chr.18: 66501517-66501517 on Build GRCh38
rs537317135 Chr.18: 66501281-66501281 on Build GRCh38
rs909713737 Chr.18: 66501305-66501305 on Build GRCh38
rs922243748 Chr.18: 66501263-66501263 on Build GRCh38
rs554845508 Chr.18: 66501255-66501255 on Build GRCh38
rs79236222 Chr.18: 66501288-66501288 on Build GRCh38
rs925228878 Chr.18: 66501200-66501200 on Build GRCh38
rs149840246 Chr.18: 66501366-66501366 on Build GRCh38
rs978763566 Chr.18: 66501459-66501459 on Build GRCh38
rs778346473 Chr.18: 66501352-66501352 on Build GRCh38
rs895981921 Chr.18: 66501412-66501412 on Build GRCh38
rs553880345 Chr.18: 66501415-66501415 on Build GRCh38
rs935237764 Chr.18: 66501207-66501207 on Build GRCh38
rs909798017 Chr.18: 66501536-66501536 on Build GRCh38
rs746899099 Chr.18: 66501523-66501523 on Build GRCh38
rs1022901807 Chr.18: 66501132-66501132 on Build GRCh38
rs113883158 Chr.18: 66501560-66501560 on Build GRCh38
rs1007666330 Chr.18: 66501455-66501455 on Build GRCh38
rs1050603446 Chr.18: 66501286-66501286 on Build GRCh38
rs542844527 Chr.18: 66501445-66501445 on Build GRCh38
rs929902330 Chr.18: 66501284-66501284 on Build GRCh38
rs994168522 Chr.18: 66501372-66501372 on Build GRCh38
rs367928074 Chr.18: 66501405-66501405 on Build GRCh38
rs1029746954 Chr.18: 66501479-66501479 on Build GRCh38
rs748900500 Chr.18: 66501249-66501249 on Build GRCh38
rs988465022 Chr.18: 66501552-66501552 on Build GRCh38
rs867222298 Chr.18: 66501325-66501325 on Build GRCh38
rs976374423 Chr.18: 66501252-66501252 on Build GRCh38
rs939359409 Chr.18: 66501367-66501367 on Build GRCh38
rs866597567 Chr.18: 66501489-66501489 on Build GRCh38
rs1031647402 Chr.18: 66501467-66501467 on Build GRCh38
rs781583908 Chr.18: 66501098-66501098 on Build GRCh38
rs561476529 Chr.18: 66501246-66501246 on Build GRCh38
rs747655170 Chr.18: 66501353-66501353 on Build GRCh38
rs570026706 Chr.18: 66501272-66501272 on Build GRCh38
rs186615186 Chr.18: 66501424-66501424 on Build GRCh38
rs978732421 Chr.18: 66501131-66501131 on Build GRCh38
rs754603188 Chr.18: 66501314-66501314 on Build GRCh38
rs866310219 Chr.18: 66501413-66501413 on Build GRCh38
rs988048041 Chr.18: 66501225-66501225 on Build GRCh38
rs968211505 Chr.18: 66501144-66501144 on Build GRCh38
rs976448061 Chr.18: 66501479-66501481 on Build GRCh38
rs1018007354 Chr.18: 66501465-66501465 on Build GRCh38
rs1056490146 Chr.18: 66501368-66501368 on Build GRCh38
rs1009791295 Chr.18: 66501126-66501126 on Build GRCh38
rs566822189 Chr.18: 66501404-66501404 on Build GRCh38
rs951045911 Chr.18: 66501496-66501498 on Build GRCh38
rs1035293367 Chr.18: 66501118-66501118 on Build GRCh38
rs566036796 Chr.18: 66501119-66501119 on Build GRCh38
rs948820 Chr.18: 66501356-66501356 on Build GRCh38
rs773112755 Chr.18: 66501485-66501485 on Build GRCh38

View Full Product Details