Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 66508830-66509328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH19
Entrez Gene ID
28513
Gene Name
cadherin 19
Gene Aliases
CDH7, CDH7L2
UniGene
Hs.42771
Gene Chromosome Location
Chr.18: 66501187-66604241 on Build GRCh38
Transcripts
NM_001271028.1 NM_021153.3 NR_073130.1 XM_011525931.2 XM_017025711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5815699 c.1577-2A>T p.? Chr.18: 66509248-66509248 on Build GRCh38
6493200 c.1630C>A p.P544T Chr.18: 66509193-66509193 on Build GRCh38
3821685 c.1631C>A p.P544H Chr.18: 66509192-66509192 on Build GRCh38
5783308 c.1806C>A p.L602L Chr.18: 66509017-66509017 on Build GRCh38
4529606 c.1627G>A p.E543K Chr.18: 66509196-66509196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780716221 Chr.18: 66509051-66509051 on Build GRCh38
rs755850405 Chr.18: 66509065-66509065 on Build GRCh38
rs970272972 Chr.18: 66508873-66508873 on Build GRCh38
rs752965631 Chr.18: 66509138-66509138 on Build GRCh38
rs777146093 Chr.18: 66508958-66508958 on Build GRCh38
rs570028831 Chr.18: 66509121-66509121 on Build GRCh38
rs980666266 Chr.18: 66508899-66508899 on Build GRCh38
rs771959922 Chr.18: 66509296-66509296 on Build GRCh38
rs760264043 Chr.18: 66509208-66509208 on Build GRCh38
rs752556700 Chr.18: 66508952-66508952 on Build GRCh38
rs758420270 Chr.18: 66508955-66508955 on Build GRCh38
rs747560102 Chr.18: 66509083-66509083 on Build GRCh38
rs773302376 Chr.18: 66509192-66509193 on Build GRCh38
rs769856415 Chr.18: 66509164-66509164 on Build GRCh38
rs765120513 Chr.18: 66508944-66508944 on Build GRCh38
rs535975922 Chr.18: 66508857-66508857 on Build GRCh38
rs745955503 Chr.18: 66509161-66509161 on Build GRCh38
rs750000043 Chr.18: 66509126-66509126 on Build GRCh38
rs926119294 Chr.18: 66509302-66509302 on Build GRCh38
rs752606398 Chr.18: 66509026-66509026 on Build GRCh38
rs933971650 Chr.18: 66509025-66509025 on Build GRCh38
rs772189050 Chr.18: 66509191-66509192 on Build GRCh38
rs200384608 Chr.18: 66509247-66509247 on Build GRCh38
rs763894852 Chr.18: 66509117-66509117 on Build GRCh38
rs749512374 Chr.18: 66509284-66509284 on Build GRCh38
rs774189883 Chr.18: 66509193-66509193 on Build GRCh38
rs917844326 Chr.18: 66508869-66508869 on Build GRCh38
rs756844149 Chr.18: 66508967-66508967 on Build GRCh38
rs763120390 Chr.18: 66509178-66509178 on Build GRCh38
rs761787994 Chr.18: 66509120-66509120 on Build GRCh38
rs150398178 Chr.18: 66509019-66509019 on Build GRCh38
rs150353589 Chr.18: 66509149-66509149 on Build GRCh38
rs765847179 Chr.18: 66509214-66509214 on Build GRCh38
rs201875592 Chr.18: 66509082-66509082 on Build GRCh38
rs775087702 Chr.18: 66509100-66509100 on Build GRCh38
rs776911833 Chr.18: 66509087-66509087 on Build GRCh38
rs776337568 Chr.18: 66508870-66508870 on Build GRCh38
rs771636477 Chr.18: 66509150-66509150 on Build GRCh38
rs759837003 Chr.18: 66509092-66509092 on Build GRCh38
rs746532503 Chr.18: 66508961-66508961 on Build GRCh38
rs970730738 Chr.18: 66509264-66509264 on Build GRCh38
rs747949472 Chr.18: 66509289-66509289 on Build GRCh38
rs1011621976 Chr.18: 66509130-66509130 on Build GRCh38
rs750051171 Chr.18: 66509061-66509061 on Build GRCh38
rs775809410 Chr.18: 66509177-66509177 on Build GRCh38
rs745892548 Chr.18: 66509267-66509267 on Build GRCh38
rs913243965 Chr.18: 66509001-66509001 on Build GRCh38
rs756183591 Chr.18: 66509279-66509279 on Build GRCh38
rs936104626 Chr.18: 66508937-66508937 on Build GRCh38
rs780844802 Chr.18: 66508969-66508969 on Build GRCh38
rs758200517 Chr.18: 66509027-66509027 on Build GRCh38
rs3764472 Chr.18: 66509032-66509032 on Build GRCh38
rs553634152 Chr.18: 66509281-66509281 on Build GRCh38
rs768019916 Chr.18: 66508939-66508988 on Build GRCh38
rs757783009 Chr.18: 66509266-66509266 on Build GRCh38
rs572151385 Chr.18: 66509306-66509306 on Build GRCh38
rs547614576 Chr.18: 66508977-66508977 on Build GRCh38
rs752575939 Chr.18: 66509040-66509040 on Build GRCh38
rs1013855720 Chr.18: 66509181-66509181 on Build GRCh38
rs781356507 Chr.18: 66509156-66509156 on Build GRCh38
rs151229169 Chr.18: 66508974-66508974 on Build GRCh38
rs768475225 Chr.18: 66509184-66509184 on Build GRCh38
rs755653539 Chr.18: 66509127-66509127 on Build GRCh38
rs146826932 Chr.18: 66509029-66509029 on Build GRCh38
rs770062227 Chr.18: 66509094-66509094 on Build GRCh38
rs547476906 Chr.18: 66509300-66509300 on Build GRCh38
rs766672388 Chr.18: 66509007-66509007 on Build GRCh38
rs1031984943 Chr.18: 66509124-66509124 on Build GRCh38
rs1055335 Chr.18: 66509128-66509128 on Build GRCh38
rs376977357 Chr.18: 66509203-66509203 on Build GRCh38
rs764104518 Chr.18: 66509028-66509028 on Build GRCh38
rs767445363 Chr.18: 66509196-66509196 on Build GRCh38
rs780042579 Chr.18: 66509280-66509280 on Build GRCh38
rs1039297084 Chr.18: 66509052-66509052 on Build GRCh38
rs748587721 Chr.18: 66508984-66508984 on Build GRCh38
rs748560534 Chr.18: 66509078-66509078 on Build GRCh38
rs571116499 Chr.18: 66509189-66509189 on Build GRCh38
rs1026009837 Chr.18: 66509158-66509158 on Build GRCh38
rs749889296 Chr.18: 66509205-66509205 on Build GRCh38
rs760863253 Chr.18: 66508993-66508993 on Build GRCh38
rs751463316 Chr.18: 66509119-66509119 on Build GRCh38
rs758759719 Chr.18: 66509144-66509144 on Build GRCh38
rs778926316 Chr.18: 66508945-66508946 on Build GRCh38
rs764552620 Chr.18: 66509230-66509230 on Build GRCh38
rs565084237 Chr.18: 66508901-66508901 on Build GRCh38
rs559793081 Chr.18: 66509017-66509017 on Build GRCh38
rs774065192 Chr.18: 66509287-66509287 on Build GRCh38
rs768932423 Chr.18: 66509285-66509285 on Build GRCh38
rs771117251 Chr.18: 66509084-66509084 on Build GRCh38
rs761733099 Chr.18: 66509195-66509195 on Build GRCh38
rs774823923 Chr.18: 66508981-66508981 on Build GRCh38
rs1024539391 Chr.18: 66509246-66509246 on Build GRCh38
rs779138174 Chr.18: 66509068-66509068 on Build GRCh38
rs772411740 Chr.18: 66509081-66509081 on Build GRCh38
rs555362958 Chr.18: 66508973-66508973 on Build GRCh38
rs764929174 Chr.18: 66509020-66509020 on Build GRCh38
rs772639440 Chr.18: 66508990-66508990 on Build GRCh38
rs1051722402 Chr.18: 66509031-66509031 on Build GRCh38
rs762825177 Chr.18: 66509109-66509109 on Build GRCh38
rs754648930 Chr.18: 66509225-66509225 on Build GRCh38
rs936787184 Chr.18: 66508954-66508954 on Build GRCh38
rs538620894 Chr.18: 66509224-66509224 on Build GRCh38
rs748196865 Chr.18: 66509061-66509061 on Build GRCh38
rs902323350 Chr.18: 66508902-66508902 on Build GRCh38
rs140371685 Chr.18: 66509048-66509048 on Build GRCh38

View Full Product Details