Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 69956649-69957162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD226
Entrez Gene ID
10666
Gene Name
CD226 molecule
Gene Aliases
DNAM-1, DNAM1, PTA1, TLiSA1
UniGene
Hs.660130
Gene Chromosome Location
Chr.18: 69860228-69962086 on Build GRCh38
Transcripts
NM_001303619.1 NM_006566.3 XM_005266642.3 XM_006722374.3

SNP Detail

SNP ID Chromosome Location
rs112067947 Chr.18: 69956675-69956675 on Build GRCh38
rs1025776982 Chr.18: 69956825-69956825 on Build GRCh38
rs796323862 Chr.18: 69956889-69956889 on Build GRCh38
rs980413077 Chr.18: 69956876-69956876 on Build GRCh38
rs554579615 Chr.18: 69956716-69956716 on Build GRCh38
rs537130264 Chr.18: 69956812-69956812 on Build GRCh38
rs887680412 Chr.18: 69956810-69956810 on Build GRCh38
rs4891383 Chr.18: 69956937-69956937 on Build GRCh38
rs759353052 Chr.18: 69956912-69956912 on Build GRCh38
rs766923619 Chr.18: 69956672-69956673 on Build GRCh38
rs67532422 Chr.18: 69956675-69956681 on Build GRCh38
rs776436487 Chr.18: 69956783-69956783 on Build GRCh38
rs753311815 Chr.18: 69956844-69956844 on Build GRCh38
rs951664410 Chr.18: 69957131-69957131 on Build GRCh38
rs766144448 Chr.18: 69956776-69956776 on Build GRCh38
rs914855384 Chr.18: 69956873-69956873 on Build GRCh38
rs995551008 Chr.18: 69957034-69957034 on Build GRCh38
rs149499885 Chr.18: 69956678-69956678 on Build GRCh38
rs891159820 Chr.18: 69956791-69956791 on Build GRCh38
rs951540719 Chr.18: 69956835-69956835 on Build GRCh38
rs1018242844 Chr.18: 69956963-69956963 on Build GRCh38
rs1006884708 Chr.18: 69956799-69956799 on Build GRCh38
rs1022597124 Chr.18: 69956818-69956818 on Build GRCh38
rs576235269 Chr.18: 69956800-69956800 on Build GRCh38
rs985687815 Chr.18: 69956893-69956893 on Build GRCh38
rs757347658 Chr.18: 69957109-69957109 on Build GRCh38
rs563757064 Chr.18: 69957124-69957125 on Build GRCh38
rs542633396 Chr.18: 69956944-69956944 on Build GRCh38
rs889763161 Chr.18: 69956688-69956688 on Build GRCh38
rs981599952 Chr.18: 69956833-69956833 on Build GRCh38
rs892273947 Chr.18: 69956768-69956768 on Build GRCh38
rs1052284540 Chr.18: 69956742-69956742 on Build GRCh38
rs551603932 Chr.18: 69956849-69956849 on Build GRCh38
rs1052150057 Chr.18: 69956927-69956927 on Build GRCh38
rs118053430 Chr.18: 69956936-69956936 on Build GRCh38
rs188992378 Chr.18: 69956924-69956924 on Build GRCh38
rs1045253625 Chr.18: 69956689-69956689 on Build GRCh38
rs993338409 Chr.18: 69956784-69956784 on Build GRCh38
rs1047538735 Chr.18: 69957014-69957014 on Build GRCh38
rs185294450 Chr.18: 69956923-69956923 on Build GRCh38
rs34070253 Chr.18: 69956753-69956754 on Build GRCh38
rs867342236 Chr.18: 69956709-69956709 on Build GRCh38
rs918679693 Chr.18: 69956955-69956955 on Build GRCh38
rs72064680 Chr.18: 69956673-69956679 on Build GRCh38
rs918170507 Chr.18: 69956863-69956863 on Build GRCh38
rs905533648 Chr.18: 69957095-69957095 on Build GRCh38
rs115493081 Chr.18: 69956821-69956821 on Build GRCh38
rs947743000 Chr.18: 69956870-69956870 on Build GRCh38
rs1006110959 Chr.18: 69957029-69957029 on Build GRCh38
rs929157288 Chr.18: 69956973-69956973 on Build GRCh38
rs1044252307 Chr.18: 69957061-69957061 on Build GRCh38
rs988976594 Chr.18: 69956854-69956854 on Build GRCh38
rs1030207815 Chr.18: 69956789-69956789 on Build GRCh38
rs1041096999 Chr.18: 69956711-69956711 on Build GRCh38
rs560923348 Chr.18: 69957028-69957028 on Build GRCh38
rs1049744527 Chr.18: 69956672-69956672 on Build GRCh38
rs765632551 Chr.18: 69956677-69956683 on Build GRCh38
rs566435642 Chr.18: 69956881-69956881 on Build GRCh38
rs956439014 Chr.18: 69956846-69956846 on Build GRCh38
rs1025680323 Chr.18: 69957112-69957112 on Build GRCh38
rs903595140 Chr.18: 69956694-69956694 on Build GRCh38
rs576962849 Chr.18: 69956899-69956899 on Build GRCh38

View Full Product Details