Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 72537358-72537867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLN2
Entrez Gene ID
147381
Gene Name
cerebellin 2 precursor
Gene Aliases
-
UniGene
Hs.569851
Gene Chromosome Location
Chr.18: 72533720-72638485 on Build GRCh38
Transcripts
NM_182511.3 XM_006722394.3 XM_011525824.2 XM_017025559.1 XM_017025560.1

SNP Detail

SNP ID Chromosome Location
rs768022821 Chr.18: 72537711-72537711 on Build GRCh38
rs616371 Chr.18: 72537462-72537462 on Build GRCh38
rs967874583 Chr.18: 72537767-72537767 on Build GRCh38
rs533678268 Chr.18: 72537513-72537513 on Build GRCh38
rs774893377 Chr.18: 72537598-72537598 on Build GRCh38
rs1028156335 Chr.18: 72537545-72537545 on Build GRCh38
rs1022186393 Chr.18: 72537759-72537759 on Build GRCh38
rs902662053 Chr.18: 72537793-72537793 on Build GRCh38
rs564851585 Chr.18: 72537425-72537425 on Build GRCh38
rs996616637 Chr.18: 72537842-72537842 on Build GRCh38
rs937635806 Chr.18: 72537709-72537709 on Build GRCh38
rs955286267 Chr.18: 72537563-72537563 on Build GRCh38
rs763294951 Chr.18: 72537551-72537551 on Build GRCh38
rs1015754273 Chr.18: 72537392-72537392 on Build GRCh38
rs967864742 Chr.18: 72537485-72537485 on Build GRCh38
rs1022176541 Chr.18: 72537483-72537483 on Build GRCh38
rs187322565 Chr.18: 72537738-72537738 on Build GRCh38
rs900501572 Chr.18: 72537837-72537837 on Build GRCh38
rs867169483 Chr.18: 72537495-72537495 on Build GRCh38
rs760415741 Chr.18: 72537780-72537780 on Build GRCh38
rs1042572383 Chr.18: 72537429-72537429 on Build GRCh38
rs775450068 Chr.18: 72537443-72537443 on Build GRCh38
rs896827579 Chr.18: 72537415-72537415 on Build GRCh38
rs553546398 Chr.18: 72537561-72537561 on Build GRCh38
rs986627960 Chr.18: 72537585-72537585 on Build GRCh38
rs145556323 Chr.18: 72537613-72537613 on Build GRCh38
rs764078682 Chr.18: 72537812-72537812 on Build GRCh38
rs940830154 Chr.18: 72537675-72537675 on Build GRCh38
rs571237213 Chr.18: 72537466-72537466 on Build GRCh38
rs1013801416 Chr.18: 72537420-72537420 on Build GRCh38
rs556199014 Chr.18: 72537663-72537663 on Build GRCh38
rs909421367 Chr.18: 72537669-72537669 on Build GRCh38
rs66925706 Chr.18: 72537456-72537456 on Build GRCh38
rs918207159 Chr.18: 72537744-72537744 on Build GRCh38
rs919610932 Chr.18: 72537706-72537706 on Build GRCh38
rs948328516 Chr.18: 72537662-72537662 on Build GRCh38
rs542473254 Chr.18: 72537661-72537661 on Build GRCh38
rs996781819 Chr.18: 72537523-72537523 on Build GRCh38
rs530590637 Chr.18: 72537453-72537453 on Build GRCh38
rs974204705 Chr.18: 72537651-72537651 on Build GRCh38
rs73966255 Chr.18: 72537729-72537729 on Build GRCh38
rs949605912 Chr.18: 72537758-72537758 on Build GRCh38
rs992538274 Chr.18: 72537634-72537634 on Build GRCh38
rs773641162 Chr.18: 72537733-72537733 on Build GRCh38
rs1049301261 Chr.18: 72537667-72537667 on Build GRCh38
rs565321983 Chr.18: 72537766-72537766 on Build GRCh38
rs1036787447 Chr.18: 72537676-72537676 on Build GRCh38
rs145307033 Chr.18: 72537836-72537836 on Build GRCh38
rs527784292 Chr.18: 72537828-72537828 on Build GRCh38
rs896874255 Chr.18: 72537700-72537700 on Build GRCh38

View Full Product Details