Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 72537755-72538250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLN2
Entrez Gene ID
147381
Gene Name
cerebellin 2 precursor
Gene Aliases
-
UniGene
Hs.569851
Gene Chromosome Location
Chr.18: 72533720-72638485 on Build GRCh38
Transcripts
NM_182511.3 XM_006722394.3 XM_011525824.2 XM_017025559.1 XM_017025560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1389626 c.614G>A p.G205D Chr.18: 72538237-72538237 on Build GRCh38
1389625 c.650C>T p.S217L Chr.18: 72538201-72538201 on Build GRCh38
4894220 c.670C>T p.L224L Chr.18: 72538181-72538181 on Build GRCh38
1389624 c.668C>A p.P223H Chr.18: 72538183-72538183 on Build GRCh38
6620951 c.609G>C p.E203D Chr.18: 72538242-72538242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992234578 Chr.18: 72537845-72537845 on Build GRCh38
rs755777666 Chr.18: 72538191-72538191 on Build GRCh38
rs780827068 Chr.18: 72538176-72538176 on Build GRCh38
rs1036909589 Chr.18: 72537937-72537937 on Build GRCh38
rs766041524 Chr.18: 72537858-72537858 on Build GRCh38
rs370758001 Chr.18: 72538134-72538134 on Build GRCh38
rs73478206 Chr.18: 72538203-72538203 on Build GRCh38
rs567089320 Chr.18: 72538104-72538104 on Build GRCh38
rs1015398557 Chr.18: 72537887-72537887 on Build GRCh38
rs900616251 Chr.18: 72538086-72538086 on Build GRCh38
rs747489925 Chr.18: 72538206-72538206 on Build GRCh38
rs147631667 Chr.18: 72538048-72538048 on Build GRCh38
rs902662053 Chr.18: 72537793-72537793 on Build GRCh38
rs996616637 Chr.18: 72537842-72537842 on Build GRCh38
rs923451326 Chr.18: 72537945-72537945 on Build GRCh38
rs368019104 Chr.18: 72538173-72538173 on Build GRCh38
rs1053953562 Chr.18: 72538071-72538071 on Build GRCh38
rs772498335 Chr.18: 72538127-72538127 on Build GRCh38
rs897068810 Chr.18: 72538155-72538155 on Build GRCh38
rs923473797 Chr.18: 72538178-72538178 on Build GRCh38
rs771463589 Chr.18: 72538224-72538224 on Build GRCh38
rs903742263 Chr.18: 72538020-72538020 on Build GRCh38
rs199631826 Chr.18: 72538143-72538143 on Build GRCh38
rs754478664 Chr.18: 72537901-72537901 on Build GRCh38
rs34699127 Chr.18: 72538167-72538167 on Build GRCh38
rs551079563 Chr.18: 72538026-72538026 on Build GRCh38
rs779514783 Chr.18: 72538194-72538194 on Build GRCh38
rs917573348 Chr.18: 72538044-72538044 on Build GRCh38
rs990921162 Chr.18: 72538032-72538032 on Build GRCh38
rs757433037 Chr.18: 72538061-72538062 on Build GRCh38
rs1050856978 Chr.18: 72537981-72537981 on Build GRCh38
rs900501572 Chr.18: 72537837-72537837 on Build GRCh38
rs760415741 Chr.18: 72537780-72537780 on Build GRCh38
rs752655925 Chr.18: 72538102-72538102 on Build GRCh38
rs867406493 Chr.18: 72538183-72538183 on Build GRCh38
rs371750538 Chr.18: 72538188-72538188 on Build GRCh38
rs752528591 Chr.18: 72538163-72538163 on Build GRCh38
rs561429387 Chr.18: 72537928-72537928 on Build GRCh38
rs374962749 Chr.18: 72538200-72538200 on Build GRCh38
rs550424923 Chr.18: 72537987-72537987 on Build GRCh38
rs970349935 Chr.18: 72538212-72538212 on Build GRCh38
rs553458535 Chr.18: 72538099-72538099 on Build GRCh38
rs533594528 Chr.18: 72538070-72538070 on Build GRCh38
rs764078682 Chr.18: 72537812-72537812 on Build GRCh38
rs978311837 Chr.18: 72538057-72538057 on Build GRCh38
rs961473771 Chr.18: 72537888-72537888 on Build GRCh38
rs776738382 Chr.18: 72538152-72538152 on Build GRCh38
rs558634017 Chr.18: 72538169-72538169 on Build GRCh38
rs751503806 Chr.18: 72538172-72538172 on Build GRCh38
rs969355241 Chr.18: 72537850-72537850 on Build GRCh38
rs868142587 Chr.18: 72538184-72538184 on Build GRCh38
rs932069827 Chr.18: 72538097-72538097 on Build GRCh38
rs770946224 Chr.18: 72538150-72538150 on Build GRCh38
rs777330942 Chr.18: 72538163-72538163 on Build GRCh38
rs924115858 Chr.18: 72538061-72538061 on Build GRCh38
rs538653467 Chr.18: 72538078-72538078 on Build GRCh38
rs145290226 Chr.18: 72538179-72538179 on Build GRCh38
rs759769062 Chr.18: 72538158-72538158 on Build GRCh38
rs761199634 Chr.18: 72538145-72538145 on Build GRCh38
rs368883601 Chr.18: 72538125-72538125 on Build GRCh38
rs570228659 Chr.18: 72537990-72537990 on Build GRCh38
rs1030279898 Chr.18: 72537849-72537849 on Build GRCh38
rs1023643995 Chr.18: 72538075-72538075 on Build GRCh38
rs59751882 Chr.18: 72538166-72538166 on Build GRCh38
rs985017420 Chr.18: 72537856-72537856 on Build GRCh38
rs1015426534 Chr.18: 72538135-72538135 on Build GRCh38
rs746751951 Chr.18: 72538159-72538159 on Build GRCh38
rs141549773 Chr.18: 72538124-72538124 on Build GRCh38
rs145307033 Chr.18: 72537836-72537836 on Build GRCh38
rs374299629 Chr.18: 72537999-72537999 on Build GRCh38
rs940882464 Chr.18: 72537919-72537919 on Build GRCh38
rs889462907 Chr.18: 72537986-72537986 on Build GRCh38
rs193130187 Chr.18: 72538014-72538014 on Build GRCh38
rs949636725 Chr.18: 72538050-72538050 on Build GRCh38
rs527784292 Chr.18: 72537828-72537828 on Build GRCh38
rs572397653 Chr.18: 72537952-72537952 on Build GRCh38
rs633092 Chr.18: 72537870-72537870 on Build GRCh38

View Full Product Details