Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40301574-40302078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A2
Entrez Gene ID
1298
Gene Name
collagen type IX alpha 2
Gene Aliases
DJ39G22.4, EDM2, MED, STL5
UniGene
Hs.418012
Gene Chromosome Location
Chr.1: 40300487-40317653 on Build GRCh38
Transcripts
NM_001852.3 XM_006710365.3 XM_011540715.2 XM_011540716.2 XM_011540717.2 XM_017000332.1 XM_017000333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5846676 c.1832G>A p.R611Q Chr.1: 40301850-40301850 on Build GRCh38
909431 c.1837C>T p.H613Y Chr.1: 40301845-40301845 on Build GRCh38
3934725 c.1824A>C p.E608D Chr.1: 40301858-40301858 on Build GRCh38
3489542 c.1793-1G>A p.? Chr.1: 40301890-40301890 on Build GRCh38
909432 c.1826T>C p.V609A Chr.1: 40301856-40301856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776104044 Chr.1: 40301917-40301917 on Build GRCh38
rs372342860 Chr.1: 40301807-40301807 on Build GRCh38
rs746562272 Chr.1: 40301912-40301912 on Build GRCh38
rs544478587 Chr.1: 40301727-40301727 on Build GRCh38
rs984663279 Chr.1: 40301711-40301711 on Build GRCh38
rs371875492 Chr.1: 40301784-40301784 on Build GRCh38
rs534134155 Chr.1: 40302042-40302042 on Build GRCh38
rs1008151090 Chr.1: 40301647-40301647 on Build GRCh38
rs541441413 Chr.1: 40301656-40301656 on Build GRCh38
rs1013441541 Chr.1: 40302023-40302023 on Build GRCh38
rs771660985 Chr.1: 40301825-40301825 on Build GRCh38
rs541456378 Chr.1: 40301861-40301861 on Build GRCh38
rs1048397045 Chr.1: 40301936-40301936 on Build GRCh38
rs777618371 Chr.1: 40301805-40301805 on Build GRCh38
rs75924208 Chr.1: 40301754-40301754 on Build GRCh38
rs775570119 Chr.1: 40301883-40301883 on Build GRCh38
rs757079626 Chr.1: 40301889-40301889 on Build GRCh38
rs868625575 Chr.1: 40301866-40301866 on Build GRCh38
rs752785723 Chr.1: 40301855-40301855 on Build GRCh38
rs959177145 Chr.1: 40302001-40302001 on Build GRCh38
rs759578875 Chr.1: 40301824-40301824 on Build GRCh38
rs375951731 Chr.1: 40302052-40302052 on Build GRCh38
rs755487302 Chr.1: 40301850-40301850 on Build GRCh38
rs961463613 Chr.1: 40301659-40301659 on Build GRCh38
rs138949427 Chr.1: 40301858-40301858 on Build GRCh38
rs760311046 Chr.1: 40301839-40301839 on Build GRCh38
rs1008455322 Chr.1: 40301775-40301775 on Build GRCh38
rs1027482569 Chr.1: 40301795-40301795 on Build GRCh38
rs954034778 Chr.1: 40301666-40301666 on Build GRCh38
rs770501503 Chr.1: 40301797-40301797 on Build GRCh38
rs118134553 Chr.1: 40302000-40302000 on Build GRCh38
rs150450056 Chr.1: 40301693-40301693 on Build GRCh38
rs745744307 Chr.1: 40301899-40301899 on Build GRCh38
rs375875745 Chr.1: 40301843-40301843 on Build GRCh38
rs139929272 Chr.1: 40301803-40301803 on Build GRCh38
rs149823112 Chr.1: 40301665-40301665 on Build GRCh38
rs1049863222 Chr.1: 40301880-40301880 on Build GRCh38
rs951954434 Chr.1: 40301701-40301701 on Build GRCh38
rs74541133 Chr.1: 40301720-40301720 on Build GRCh38
rs748341610 Chr.1: 40301792-40301792 on Build GRCh38
rs368106966 Chr.1: 40301768-40301768 on Build GRCh38
rs751603033 Chr.1: 40301785-40301785 on Build GRCh38
rs896696018 Chr.1: 40301622-40301622 on Build GRCh38
rs6674856 Chr.1: 40301906-40301906 on Build GRCh38
rs972995750 Chr.1: 40301983-40301983 on Build GRCh38
rs531953673 Chr.1: 40301851-40301851 on Build GRCh38
rs774879246 Chr.1: 40301827-40301827 on Build GRCh38
rs992498924 Chr.1: 40301833-40301833 on Build GRCh38
rs760683637 Chr.1: 40301896-40301896 on Build GRCh38
rs770970103 Chr.1: 40301816-40301816 on Build GRCh38
rs560573323 Chr.1: 40301756-40301756 on Build GRCh38
rs569248644 Chr.1: 40301965-40301965 on Build GRCh38
rs938336387 Chr.1: 40302009-40302009 on Build GRCh38
rs368763623 Chr.1: 40301757-40301757 on Build GRCh38
rs938325154 Chr.1: 40301782-40301783 on Build GRCh38
rs972557776 Chr.1: 40301742-40301742 on Build GRCh38
rs766082169 Chr.1: 40302008-40302008 on Build GRCh38
rs768336173 Chr.1: 40301914-40301914 on Build GRCh38
rs1042516104 Chr.1: 40301753-40301753 on Build GRCh38
rs142151614 Chr.1: 40301848-40301848 on Build GRCh38
rs368623893 Chr.1: 40301806-40301806 on Build GRCh38
rs774546392 Chr.1: 40301695-40301695 on Build GRCh38
rs1056718072 Chr.1: 40301603-40301603 on Build GRCh38
rs187442430 Chr.1: 40301728-40301728 on Build GRCh38
rs183518528 Chr.1: 40301628-40301628 on Build GRCh38
rs746507014 Chr.1: 40301642-40301642 on Build GRCh38
rs1027099993 Chr.1: 40301669-40301669 on Build GRCh38
rs1001637084 Chr.1: 40301986-40301986 on Build GRCh38
rs936838151 Chr.1: 40301755-40301755 on Build GRCh38
rs572258426 Chr.1: 40302001-40302003 on Build GRCh38
rs763724733 Chr.1: 40301840-40301840 on Build GRCh38
rs756634762 Chr.1: 40301664-40301664 on Build GRCh38
rs776614013 Chr.1: 40301964-40301964 on Build GRCh38
rs756345160 Chr.1: 40301856-40301856 on Build GRCh38
rs766668857 Chr.1: 40301935-40301935 on Build GRCh38
rs961226125 Chr.1: 40301732-40301732 on Build GRCh38
rs1026790340 Chr.1: 40301990-40301990 on Build GRCh38
rs868108157 Chr.1: 40301865-40301865 on Build GRCh38
rs778874959 Chr.1: 40301898-40301898 on Build GRCh38
rs781268566 Chr.1: 40301787-40301787 on Build GRCh38
rs917591985 Chr.1: 40301736-40301736 on Build GRCh38
rs749078666 Chr.1: 40301884-40301884 on Build GRCh38
rs766758379 Chr.1: 40301846-40301846 on Build GRCh38
rs761473011 Chr.1: 40301928-40301928 on Build GRCh38
rs774470944 Chr.1: 40301823-40301823 on Build GRCh38
rs750760762 Chr.1: 40301764-40301764 on Build GRCh38
rs775064718 Chr.1: 40301909-40301909 on Build GRCh38
rs1020037987 Chr.1: 40301658-40301658 on Build GRCh38
rs144792323 Chr.1: 40301743-40301743 on Build GRCh38
rs992456020 Chr.1: 40302029-40302029 on Build GRCh38

View Full Product Details